Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209782366-209782865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf74
Entrez Gene ID
148304
Gene Name
chromosome 1 open reading frame 74
Gene Aliases
URLC4
UniGene
Hs.497642
Gene Chromosome Location
Chr.1: 209779208-209784730 on Build GRCh38
Transcripts
NM_152485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1929232 c.804C>T p.A268A Chr.1: 209782831-209782831 on Build GRCh38
425215 c.795G>C p.P265P Chr.1: 209782840-209782840 on Build GRCh38
5969131 c.794_795insCC p.A266fs*>5 Chr.1: 209782840-209782841 on Build GRCh38
6396778 c.775T>C p.S259P Chr.1: 209782860-209782860 on Build GRCh38
7336178 c.773C>T p.S258F Chr.1: 209782862-209782862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568607543 Chr.1: 209782396-209782396 on Build GRCh38
rs891067549 Chr.1: 209782630-209782630 on Build GRCh38
rs947747327 Chr.1: 209782544-209782544 on Build GRCh38
rs930887926 Chr.1: 209782723-209782723 on Build GRCh38
rs893051251 Chr.1: 209782520-209782520 on Build GRCh38
rs761822377 Chr.1: 209782819-209782819 on Build GRCh38
rs981170451 Chr.1: 209782608-209782608 on Build GRCh38
rs996012394 Chr.1: 209782496-209782496 on Build GRCh38
rs148900804 Chr.1: 209782823-209782823 on Build GRCh38
rs542021007 Chr.1: 209782719-209782719 on Build GRCh38
rs747958511 Chr.1: 209782760-209782760 on Build GRCh38
rs769459706 Chr.1: 209782777-209782777 on Build GRCh38
rs755050983 Chr.1: 209782755-209782755 on Build GRCh38
rs983643975 Chr.1: 209782471-209782471 on Build GRCh38
rs751511633 Chr.1: 209782811-209782811 on Build GRCh38
rs771325415 Chr.1: 209782839-209782839 on Build GRCh38
rs145674628 Chr.1: 209782622-209782622 on Build GRCh38
rs150951869 Chr.1: 209782841-209782841 on Build GRCh38
rs72741033 Chr.1: 209782472-209782472 on Build GRCh38
rs760354082 Chr.1: 209782821-209782822 on Build GRCh38
rs778281874 Chr.1: 209782805-209782805 on Build GRCh38
rs1019917098 Chr.1: 209782558-209782558 on Build GRCh38
rs952058708 Chr.1: 209782449-209782449 on Build GRCh38
rs916850696 Chr.1: 209782678-209782678 on Build GRCh38
rs763659756 Chr.1: 209782806-209782806 on Build GRCh38
rs762377924 Chr.1: 209782804-209782804 on Build GRCh38
rs562702510 Chr.1: 209782649-209782649 on Build GRCh38
rs748764138 Chr.1: 209782510-209782510 on Build GRCh38
rs1057068190 Chr.1: 209782507-209782507 on Build GRCh38
rs546192494 Chr.1: 209782536-209782536 on Build GRCh38
rs775150702 Chr.1: 209782803-209782803 on Build GRCh38
rs1011232035 Chr.1: 209782546-209782546 on Build GRCh38
rs963918068 Chr.1: 209782690-209782690 on Build GRCh38
rs1019017139 Chr.1: 209782391-209782391 on Build GRCh38
rs558213359 Chr.1: 209782588-209782588 on Build GRCh38
rs996185896 Chr.1: 209782747-209782747 on Build GRCh38
rs796855616 Chr.1: 209782705-209782705 on Build GRCh38
rs759854681 Chr.1: 209782774-209782774 on Build GRCh38
rs868040997 Chr.1: 209782783-209782783 on Build GRCh38
rs536050310 Chr.1: 209782419-209782419 on Build GRCh38
rs750059595 Chr.1: 209782409-209782409 on Build GRCh38
rs1018484329 Chr.1: 209782655-209782655 on Build GRCh38
rs529807748 Chr.1: 209782676-209782676 on Build GRCh38
rs144888845 Chr.1: 209782392-209782392 on Build GRCh38
rs753849030 Chr.1: 209782394-209782394 on Build GRCh38
rs114937059 Chr.1: 209782598-209782598 on Build GRCh38
rs1040460852 Chr.1: 209782426-209782426 on Build GRCh38
rs651141 Chr.1: 209782503-209782503 on Build GRCh38
rs886403612 Chr.1: 209782413-209782413 on Build GRCh38

View Full Product Details