Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212565242-212565742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF3
Entrez Gene ID
467
Gene Name
activating transcription factor 3
Gene Aliases
-
UniGene
Hs.460
Gene Chromosome Location
Chr.1: 212565334-212620777 on Build GRCh38
Transcripts
NM_001030287.3

SNP Detail

SNP ID Chromosome Location
rs774352146 Chr.1: 212565528-212565528 on Build GRCh38
rs550363172 Chr.1: 212565387-212565387 on Build GRCh38
rs903921581 Chr.1: 212565637-212565637 on Build GRCh38
rs767277600 Chr.1: 212565608-212565608 on Build GRCh38
rs984917541 Chr.1: 212565381-212565381 on Build GRCh38
rs895299155 Chr.1: 212565340-212565340 on Build GRCh38
rs113082637 Chr.1: 212565363-212565363 on Build GRCh38
rs754754158 Chr.1: 212565584-212565584 on Build GRCh38
rs1055646134 Chr.1: 212565338-212565338 on Build GRCh38
rs146798629 Chr.1: 212565694-212565694 on Build GRCh38
rs1006621624 Chr.1: 212565434-212565434 on Build GRCh38
rs772837203 Chr.1: 212565489-212565489 on Build GRCh38
rs1195472 Chr.1: 212565567-212565567 on Build GRCh38
rs752527276 Chr.1: 212565691-212565691 on Build GRCh38
rs141171464 Chr.1: 212565570-212565570 on Build GRCh38
rs890793785 Chr.1: 212565679-212565679 on Build GRCh38
rs999735887 Chr.1: 212565398-212565398 on Build GRCh38
rs1013765894 Chr.1: 212565681-212565681 on Build GRCh38
rs562507030 Chr.1: 212565391-212565391 on Build GRCh38
rs1007001347 Chr.1: 212565702-212565702 on Build GRCh38
rs890801072 Chr.1: 212565433-212565433 on Build GRCh38
rs1043950407 Chr.1: 212565350-212565350 on Build GRCh38
rs532247853 Chr.1: 212565382-212565382 on Build GRCh38
rs3806460 Chr.1: 212565493-212565493 on Build GRCh38
rs143587293 Chr.1: 212565655-212565655 on Build GRCh38
rs964756068 Chr.1: 212565406-212565406 on Build GRCh38
rs951250799 Chr.1: 212565530-212565530 on Build GRCh38
rs1013711970 Chr.1: 212565337-212565337 on Build GRCh38
rs182058164 Chr.1: 212565522-212565522 on Build GRCh38
rs1029261010 Chr.1: 212565432-212565432 on Build GRCh38
rs866758041 Chr.1: 212565707-212565707 on Build GRCh38
rs570514574 Chr.1: 212565650-212565650 on Build GRCh38
rs896744248 Chr.1: 212565680-212565680 on Build GRCh38
rs974432940 Chr.1: 212565452-212565452 on Build GRCh38
rs973867619 Chr.1: 212565477-212565477 on Build GRCh38
rs780119096 Chr.1: 212565280-212565280 on Build GRCh38
rs1044229616 Chr.1: 212565619-212565619 on Build GRCh38
rs1051318233 Chr.1: 212565669-212565669 on Build GRCh38
rs1055097609 Chr.1: 212565675-212565675 on Build GRCh38
rs961015936 Chr.1: 212565573-212565573 on Build GRCh38
rs41305114 Chr.1: 212565341-212565341 on Build GRCh38
rs551969947 Chr.1: 212565593-212565593 on Build GRCh38
rs41314256 Chr.1: 212565261-212565261 on Build GRCh38
rs960935258 Chr.1: 212565336-212565336 on Build GRCh38
rs962567101 Chr.1: 212565469-212565469 on Build GRCh38

View Full Product Details