Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212618930-212619433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF3
Entrez Gene ID
467
Gene Name
activating transcription factor 3
Gene Aliases
-
UniGene
Hs.460
Gene Chromosome Location
Chr.1: 212565334-212620777 on Build GRCh38
Transcripts
NM_001030287.3 NM_001040619.2 NM_001206484.2 NM_001206486.2 NM_001206488.2 NM_001674.3 XM_005273146.1 XM_011509579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522162 c.370C>T p.Q124* Chr.1: 212619067-212619067 on Build GRCh38
1338673 c.399A>G p.S133S Chr.1: 212619096-212619096 on Build GRCh38
3740993 c.396T>C p.I132I Chr.1: 212619405-212619405 on Build GRCh38
5368435 c.353C>T p.S118L Chr.1: 212619362-212619362 on Build GRCh38
3483147 c.412G>A p.E138K Chr.1: 212619421-212619421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201880223 Chr.1: 212619147-212619147 on Build GRCh38
rs780270474 Chr.1: 212619363-212619363 on Build GRCh38
rs752098584 Chr.1: 212619340-212619340 on Build GRCh38
rs766927717 Chr.1: 212619155-212619155 on Build GRCh38
rs924742366 Chr.1: 212619161-212619161 on Build GRCh38
rs943604526 Chr.1: 212619197-212619197 on Build GRCh38
rs550567614 Chr.1: 212619011-212619011 on Build GRCh38
rs758923412 Chr.1: 212619127-212619127 on Build GRCh38
rs771217741 Chr.1: 212619025-212619025 on Build GRCh38
rs375790706 Chr.1: 212619042-212619042 on Build GRCh38
rs1040722195 Chr.1: 212619285-212619285 on Build GRCh38
rs545110478 Chr.1: 212619209-212619209 on Build GRCh38
rs1052746055 Chr.1: 212619306-212619306 on Build GRCh38
rs11571553 Chr.1: 212619238-212619238 on Build GRCh38
rs775404705 Chr.1: 212619064-212619064 on Build GRCh38
rs1037999839 Chr.1: 212619205-212619205 on Build GRCh38
rs770096732 Chr.1: 212619144-212619144 on Build GRCh38
rs538902269 Chr.1: 212618988-212618988 on Build GRCh38
rs933377505 Chr.1: 212619171-212619171 on Build GRCh38
rs768439148 Chr.1: 212619374-212619374 on Build GRCh38
rs753140735 Chr.1: 212619122-212619122 on Build GRCh38
rs369897861 Chr.1: 212619316-212619316 on Build GRCh38
rs893599892 Chr.1: 212619023-212619023 on Build GRCh38
rs747016690 Chr.1: 212619138-212619138 on Build GRCh38
rs759869143 Chr.1: 212619046-212619046 on Build GRCh38
rs764050940 Chr.1: 212619078-212619078 on Build GRCh38
rs372541088 Chr.1: 212619330-212619330 on Build GRCh38
rs753051033 Chr.1: 212619319-212619319 on Build GRCh38
rs1051948035 Chr.1: 212619174-212619174 on Build GRCh38
rs967010475 Chr.1: 212619404-212619404 on Build GRCh38
rs751256772 Chr.1: 212619377-212619378 on Build GRCh38
rs377098998 Chr.1: 212619351-212619351 on Build GRCh38
rs374539070 Chr.1: 212619146-212619146 on Build GRCh38
rs183560666 Chr.1: 212619123-212619123 on Build GRCh38
rs771078775 Chr.1: 212619362-212619362 on Build GRCh38
rs983875561 Chr.1: 212619211-212619211 on Build GRCh38
rs781101366 Chr.1: 212619349-212619349 on Build GRCh38
rs999797158 Chr.1: 212618957-212618957 on Build GRCh38
rs780448578 Chr.1: 212619145-212619145 on Build GRCh38
rs1026099972 Chr.1: 212618982-212618982 on Build GRCh38
rs747106349 Chr.1: 212619024-212619024 on Build GRCh38
rs777957966 Chr.1: 212619022-212619022 on Build GRCh38
rs769462789 Chr.1: 212619062-212619062 on Build GRCh38
rs772814987 Chr.1: 212619307-212619307 on Build GRCh38
rs370907615 Chr.1: 212619140-212619140 on Build GRCh38
rs761808787 Chr.1: 212619152-212619152 on Build GRCh38
rs199908865 Chr.1: 212619089-212619089 on Build GRCh38
rs922537105 Chr.1: 212619286-212619286 on Build GRCh38
rs113044456 Chr.1: 212619401-212619401 on Build GRCh38
rs758836964 Chr.1: 212619323-212619323 on Build GRCh38
rs11571554 Chr.1: 212619384-212619384 on Build GRCh38
rs555908176 Chr.1: 212619148-212619148 on Build GRCh38
rs935273434 Chr.1: 212619289-212619289 on Build GRCh38
rs899407377 Chr.1: 212619237-212619237 on Build GRCh38
rs750043276 Chr.1: 212619103-212619103 on Build GRCh38
rs779111172 Chr.1: 212619119-212619119 on Build GRCh38
rs942379643 Chr.1: 212619245-212619245 on Build GRCh38
rs757831153 Chr.1: 212619345-212619345 on Build GRCh38
rs749018719 Chr.1: 212619366-212619366 on Build GRCh38
rs553312800 Chr.1: 212619317-212619317 on Build GRCh38
rs761328272 Chr.1: 212619083-212619083 on Build GRCh38
rs910690005 Chr.1: 212619182-212619182 on Build GRCh38
rs778435451 Chr.1: 212619133-212619133 on Build GRCh38
rs751023084 Chr.1: 212619082-212619082 on Build GRCh38
rs1010687118 Chr.1: 212619376-212619376 on Build GRCh38
rs781389716 Chr.1: 212619139-212619139 on Build GRCh38
rs1052290936 Chr.1: 212619009-212619009 on Build GRCh38
rs952958800 Chr.1: 212619210-212619210 on Build GRCh38
rs762846559 Chr.1: 212619076-212619076 on Build GRCh38
rs748577629 Chr.1: 212618995-212618995 on Build GRCh38
rs755858234 Chr.1: 212619107-212619107 on Build GRCh38

View Full Product Details