Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220059505-220060053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPNT1
Entrez Gene ID
10380
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Gene Aliases
HEL20, PIP
UniGene
Hs.406134
Gene Chromosome Location
Chr.1: 220057482-220089916 on Build GRCh38
Transcripts
NM_001286149.1 NM_001286150.1 NM_001286151.1 NM_006085.5 XM_005272998.2 XM_005272999.4 XM_005273000.4 XM_005273004.2 XM_005273005.4 XM_006711114.3 XM_011509063.2 XM_011509064.2 XM_017000043.1 XM_017000044.1 XM_017000046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1959068 c.760A>G p.I254V Chr.1: 220059704-220059704 on Build GRCh38
400965 c.757G>T p.V253F Chr.1: 220059707-220059707 on Build GRCh38
1639727 c.738G>T p.W246C Chr.1: 220059726-220059726 on Build GRCh38
6244596 c.752C>A p.P251Q Chr.1: 220059712-220059712 on Build GRCh38
6612402 c.718A>G p.S240G Chr.1: 220059746-220059746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548632419 Chr.1: 220059634-220059634 on Build GRCh38
rs905093340 Chr.1: 220059733-220059733 on Build GRCh38
rs571117694 Chr.1: 220059776-220059776 on Build GRCh38
rs201616529 Chr.1: 220059753-220059753 on Build GRCh38
rs141045914 Chr.1: 220059748-220059748 on Build GRCh38
rs772637275 Chr.1: 220059723-220059723 on Build GRCh38
rs527450380 Chr.1: 220059588-220059588 on Build GRCh38
rs549734102 Chr.1: 220059619-220059619 on Build GRCh38
rs747113136 Chr.1: 220059740-220059740 on Build GRCh38
rs897686239 Chr.1: 220059540-220059540 on Build GRCh38
rs932908613 Chr.1: 220059641-220059641 on Build GRCh38
rs753119276 Chr.1: 220059823-220059823 on Build GRCh38
rs561887745 Chr.1: 220059549-220059549 on Build GRCh38
rs762592157 Chr.1: 220059766-220059766 on Build GRCh38
rs749879876 Chr.1: 220059812-220059812 on Build GRCh38
rs185585606 Chr.1: 220059622-220059622 on Build GRCh38
rs529003591 Chr.1: 220059627-220059627 on Build GRCh38
rs778194697 Chr.1: 220059734-220059734 on Build GRCh38
rs41274790 Chr.1: 220059877-220059877 on Build GRCh38
rs757737307 Chr.1: 220059834-220059834 on Build GRCh38
rs1013500374 Chr.1: 220059552-220059552 on Build GRCh38
rs564359240 Chr.1: 220059593-220059593 on Build GRCh38
rs779569648 Chr.1: 220059821-220059821 on Build GRCh38
rs561371436 Chr.1: 220059589-220059589 on Build GRCh38
rs761442614 Chr.1: 220059782-220059782 on Build GRCh38
rs750058247 Chr.1: 220059654-220059654 on Build GRCh38
rs944166995 Chr.1: 220059626-220059626 on Build GRCh38
rs763140465 Chr.1: 220059798-220059798 on Build GRCh38
rs1025271239 Chr.1: 220059587-220059587 on Build GRCh38
rs764769898 Chr.1: 220059822-220059822 on Build GRCh38
rs193243304 Chr.1: 220059602-220059602 on Build GRCh38
rs146770973 Chr.1: 220059972-220059972 on Build GRCh38
rs79763289 Chr.1: 220059606-220059606 on Build GRCh38
rs1024813297 Chr.1: 220059890-220059890 on Build GRCh38
rs866898306 Chr.1: 220059772-220059772 on Build GRCh38
rs554372287 Chr.1: 220059669-220059669 on Build GRCh38
rs746866178 Chr.1: 220059537-220059537 on Build GRCh38
rs990499260 Chr.1: 220059996-220059996 on Build GRCh38
rs779384535 Chr.1: 220059694-220059694 on Build GRCh38
rs372277833 Chr.1: 220059825-220059825 on Build GRCh38
rs773416198 Chr.1: 220059753-220059754 on Build GRCh38
rs988220858 Chr.1: 220059609-220059609 on Build GRCh38
rs745927318 Chr.1: 220059836-220059836 on Build GRCh38
rs187691249 Chr.1: 220059722-220059722 on Build GRCh38
rs751123383 Chr.1: 220059635-220059635 on Build GRCh38
rs144861510 Chr.1: 220059916-220059916 on Build GRCh38
rs375078434 Chr.1: 220059828-220059828 on Build GRCh38
rs75338368 Chr.1: 220059605-220059605 on Build GRCh38
rs781304192 Chr.1: 220059835-220059835 on Build GRCh38
rs374532533 Chr.1: 220059749-220059749 on Build GRCh38
rs958104222 Chr.1: 220059596-220059596 on Build GRCh38
rs572274304 Chr.1: 220059978-220059978 on Build GRCh38
rs755692625 Chr.1: 220059686-220059686 on Build GRCh38
rs60390808 Chr.1: 220059777-220059777 on Build GRCh38
rs984405599 Chr.1: 220059940-220059940 on Build GRCh38
rs756773504 Chr.1: 220059646-220059646 on Build GRCh38
rs767178141 Chr.1: 220059797-220059797 on Build GRCh38
rs34674672 Chr.1: 220059832-220059833 on Build GRCh38
rs1013800679 Chr.1: 220059718-220059718 on Build GRCh38
rs778282560 Chr.1: 220059827-220059827 on Build GRCh38
rs530289003 Chr.1: 220059828-220059831 on Build GRCh38
rs943201247 Chr.1: 220059636-220059636 on Build GRCh38
rs367704081 Chr.1: 220059652-220059652 on Build GRCh38
rs770493894 Chr.1: 220059699-220059700 on Build GRCh38
rs897576181 Chr.1: 220059707-220059707 on Build GRCh38
rs1024819533 Chr.1: 220059731-220059731 on Build GRCh38
rs910438971 Chr.1: 220060003-220060003 on Build GRCh38
rs776341378 Chr.1: 220059760-220059760 on Build GRCh38
rs941750462 Chr.1: 220059693-220059693 on Build GRCh38
rs977300828 Chr.1: 220059862-220059862 on Build GRCh38
rs755854373 Chr.1: 220059818-220059818 on Build GRCh38

View Full Product Details