Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220073695-220074229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPNT1
Entrez Gene ID
10380
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Gene Aliases
HEL20, PIP
UniGene
Hs.406134
Gene Chromosome Location
Chr.1: 220057482-220089916 on Build GRCh38
Transcripts
NM_001286149.1 NM_001286150.1 NM_001286151.1 NM_006085.5 XM_005272998.2 XM_005272999.4 XM_005273000.4 XM_005273004.2 XM_005273005.4 XM_006711114.3 XM_011509063.2 XM_011509064.2 XM_011509065.2 XM_017000043.1 XM_017000044.1 XM_017000045.1 XM_017000046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3671630 c.218G>T p.G73V Chr.1: 220073974-220073974 on Build GRCh38
4622537 c.174A>G p.I58M Chr.1: 220074018-220074018 on Build GRCh38
1196267 c.190C>T p.R64W Chr.1: 220074002-220074002 on Build GRCh38
3483833 c.192G>A p.R64R Chr.1: 220074000-220074000 on Build GRCh38
3385766 c.165G>A p.Q55Q Chr.1: 220074027-220074027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763742768 Chr.1: 220074085-220074085 on Build GRCh38
rs1002386297 Chr.1: 220074134-220074134 on Build GRCh38
rs941434858 Chr.1: 220074145-220074145 on Build GRCh38
rs776487679 Chr.1: 220074037-220074037 on Build GRCh38
rs2053250 Chr.1: 220073754-220073754 on Build GRCh38
rs564017946 Chr.1: 220073961-220073961 on Build GRCh38
rs1050273618 Chr.1: 220073947-220073947 on Build GRCh38
rs777594605 Chr.1: 220073956-220073956 on Build GRCh38
rs779232004 Chr.1: 220074003-220074003 on Build GRCh38
rs1024570021 Chr.1: 220073816-220073816 on Build GRCh38
rs765238928 Chr.1: 220074055-220074055 on Build GRCh38
rs749112003 Chr.1: 220073999-220073999 on Build GRCh38
rs751293308 Chr.1: 220073960-220073960 on Build GRCh38
rs975663071 Chr.1: 220074038-220074038 on Build GRCh38
rs753561097 Chr.1: 220073923-220073923 on Build GRCh38
rs755249048 Chr.1: 220074106-220074106 on Build GRCh38
rs905328039 Chr.1: 220074119-220074119 on Build GRCh38
rs773245779 Chr.1: 220074020-220074020 on Build GRCh38
rs748695809 Chr.1: 220074114-220074114 on Build GRCh38
rs759194137 Chr.1: 220074049-220074049 on Build GRCh38
rs770771610 Chr.1: 220074009-220074009 on Build GRCh38
rs956998553 Chr.1: 220073729-220073729 on Build GRCh38
rs759583480 Chr.1: 220073924-220073924 on Build GRCh38
rs975938875 Chr.1: 220073804-220073804 on Build GRCh38
rs373872580 Chr.1: 220074187-220074187 on Build GRCh38
rs989833279 Chr.1: 220073741-220073741 on Build GRCh38
rs779704931 Chr.1: 220074098-220074098 on Build GRCh38
rs371742684 Chr.1: 220074070-220074070 on Build GRCh38
rs372564231 Chr.1: 220073931-220073931 on Build GRCh38
rs772099307 Chr.1: 220074006-220074006 on Build GRCh38
rs189485725 Chr.1: 220074158-220074158 on Build GRCh38
rs766244615 Chr.1: 220073922-220073922 on Build GRCh38
rs1005999422 Chr.1: 220073750-220073750 on Build GRCh38
rs751103337 Chr.1: 220074087-220074087 on Build GRCh38
rs575413940 Chr.1: 220073884-220073884 on Build GRCh38
rs765015718 Chr.1: 220073941-220073941 on Build GRCh38
rs538156332 Chr.1: 220074105-220074105 on Build GRCh38
rs1014129353 Chr.1: 220074051-220074051 on Build GRCh38
rs199645307 Chr.1: 220074001-220074001 on Build GRCh38
rs757931822 Chr.1: 220073932-220073933 on Build GRCh38
rs779505984 Chr.1: 220074112-220074112 on Build GRCh38
rs958300535 Chr.1: 220074154-220074154 on Build GRCh38
rs775524287 Chr.1: 220074063-220074063 on Build GRCh38
rs752262821 Chr.1: 220073948-220073948 on Build GRCh38
rs369375797 Chr.1: 220074002-220074002 on Build GRCh38
rs73096557 Chr.1: 220073769-220073769 on Build GRCh38
rs201519563 Chr.1: 220073982-220073982 on Build GRCh38
rs757003991 Chr.1: 220074091-220074091 on Build GRCh38
rs748252014 Chr.1: 220074004-220074004 on Build GRCh38
rs1012820749 Chr.1: 220073788-220073788 on Build GRCh38
rs909150243 Chr.1: 220073978-220073978 on Build GRCh38
rs375210389 Chr.1: 220073977-220073977 on Build GRCh38
rs1032573550 Chr.1: 220074144-220074144 on Build GRCh38
rs773964883 Chr.1: 220074025-220074025 on Build GRCh38
rs569073511 Chr.1: 220074101-220074101 on Build GRCh38
rs923057169 Chr.1: 220073888-220073888 on Build GRCh38
rs764265903 Chr.1: 220074184-220074184 on Build GRCh38

View Full Product Details