Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220690504-220691002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf115
Entrez Gene ID
79762
Gene Name
chromosome 1 open reading frame 115
Gene Aliases
-
UniGene
Hs.519839
Gene Chromosome Location
Chr.1: 220690286-220699157 on Build GRCh38
Transcripts
NM_024709.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363296 c.160A>G p.S54G Chr.1: 220690562-220690562 on Build GRCh38
4650297 c.274A>G p.K92E Chr.1: 220690676-220690676 on Build GRCh38
5992499 c.102C>A p.A34A Chr.1: 220690504-220690504 on Build GRCh38
4651136 c.114C>T p.H38H Chr.1: 220690516-220690516 on Build GRCh38
5764323 c.188C>A p.T63N Chr.1: 220690590-220690590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766570284 Chr.1: 220690664-220690664 on Build GRCh38
rs981083621 Chr.1: 220690581-220690581 on Build GRCh38
rs573058187 Chr.1: 220690651-220690651 on Build GRCh38
rs910143111 Chr.1: 220690851-220690851 on Build GRCh38
rs755224463 Chr.1: 220690737-220690738 on Build GRCh38
rs755801510 Chr.1: 220690726-220690726 on Build GRCh38
rs895956262 Chr.1: 220690872-220690872 on Build GRCh38
rs761203884 Chr.1: 220690663-220690663 on Build GRCh38
rs750453432 Chr.1: 220690570-220690570 on Build GRCh38
rs970506921 Chr.1: 220690969-220690969 on Build GRCh38
rs760647085 Chr.1: 220690627-220690627 on Build GRCh38
rs759881686 Chr.1: 220690606-220690606 on Build GRCh38
rs754142767 Chr.1: 220690630-220690630 on Build GRCh38
rs762436308 Chr.1: 220690615-220690615 on Build GRCh38
rs765182139 Chr.1: 220690636-220690636 on Build GRCh38
rs1002395625 Chr.1: 220690977-220690977 on Build GRCh38
rs538020143 Chr.1: 220690724-220690724 on Build GRCh38
rs980481223 Chr.1: 220690798-220690798 on Build GRCh38
rs778809627 Chr.1: 220690588-220690588 on Build GRCh38
rs753734015 Chr.1: 220690578-220690578 on Build GRCh38
rs565010107 Chr.1: 220690714-220690714 on Build GRCh38
rs574239920 Chr.1: 220690613-220690613 on Build GRCh38
rs752536191 Chr.1: 220690642-220690642 on Build GRCh38
rs745538171 Chr.1: 220690657-220690657 on Build GRCh38
rs904558593 Chr.1: 220690870-220690870 on Build GRCh38
rs965865901 Chr.1: 220690809-220690809 on Build GRCh38
rs767644565 Chr.1: 220690558-220690558 on Build GRCh38
rs1024708956 Chr.1: 220690765-220690770 on Build GRCh38
rs770667491 Chr.1: 220690601-220690601 on Build GRCh38
rs529943392 Chr.1: 220690843-220690843 on Build GRCh38
rs755184006 Chr.1: 220690632-220690632 on Build GRCh38
rs767903191 Chr.1: 220690618-220690618 on Build GRCh38
rs765526130 Chr.1: 220690676-220690676 on Build GRCh38
rs544263065 Chr.1: 220690753-220690753 on Build GRCh38
rs1049760912 Chr.1: 220690867-220690867 on Build GRCh38
rs1041448851 Chr.1: 220690582-220690582 on Build GRCh38
rs2960571 Chr.1: 220690720-220690720 on Build GRCh38
rs758255547 Chr.1: 220690595-220690595 on Build GRCh38
rs752590937 Chr.1: 220690679-220690679 on Build GRCh38
rs368368973 Chr.1: 220690901-220690901 on Build GRCh38
rs1013779339 Chr.1: 220690892-220690892 on Build GRCh38
rs891952007 Chr.1: 220690928-220690928 on Build GRCh38
rs957897789 Chr.1: 220690713-220690713 on Build GRCh38
rs756751823 Chr.1: 220690653-220690653 on Build GRCh38
rs371317519 Chr.1: 220690665-220690665 on Build GRCh38
rs754841450 Chr.1: 220690584-220690584 on Build GRCh38
rs751460169 Chr.1: 220690705-220690705 on Build GRCh38
rs746918853 Chr.1: 220690652-220690652 on Build GRCh38
rs546870514 Chr.1: 220690854-220690854 on Build GRCh38
rs1053326013 Chr.1: 220690918-220690918 on Build GRCh38
rs1000404198 Chr.1: 220690879-220690879 on Build GRCh38
rs748428279 Chr.1: 220690660-220690660 on Build GRCh38
rs926494579 Chr.1: 220690802-220690802 on Build GRCh38
rs773613083 Chr.1: 220690662-220690662 on Build GRCh38
rs976837450 Chr.1: 220690810-220690810 on Build GRCh38
rs778132335 Chr.1: 220690760-220690760 on Build GRCh38
rs766476940 Chr.1: 220690574-220690574 on Build GRCh38
rs532315248 Chr.1: 220690925-220690925 on Build GRCh38
rs984076565 Chr.1: 220690811-220690811 on Build GRCh38
rs1046127570 Chr.1: 220690898-220690898 on Build GRCh38
rs762681653 Chr.1: 220690689-220690689 on Build GRCh38
rs899810311 Chr.1: 220690590-220690590 on Build GRCh38
rs748100113 Chr.1: 220690591-220690591 on Build GRCh38
rs545212089 Chr.1: 220690656-220690656 on Build GRCh38
rs745484837 Chr.1: 220690607-220690607 on Build GRCh38
rs1037220493 Chr.1: 220690856-220690856 on Build GRCh38
rs970771734 Chr.1: 220690789-220690789 on Build GRCh38
rs748920051 Chr.1: 220690749-220690749 on Build GRCh38
rs553252064 Chr.1: 220690646-220690646 on Build GRCh38
rs779656582 Chr.1: 220690743-220690743 on Build GRCh38
rs780154748 Chr.1: 220690557-220690569 on Build GRCh38
rs769418928 Chr.1: 220690659-220690659 on Build GRCh38
rs376728254 Chr.1: 220690600-220690600 on Build GRCh38
rs373915621 Chr.1: 220690718-220690718 on Build GRCh38
rs536997214 Chr.1: 220690628-220690628 on Build GRCh38
rs956249624 Chr.1: 220690738-220690738 on Build GRCh38
rs904894571 Chr.1: 220690900-220690900 on Build GRCh38
rs773378717 Chr.1: 220690622-220690622 on Build GRCh38
rs370998488 Chr.1: 220690807-220690807 on Build GRCh38
rs1031935417 Chr.1: 220690933-220690933 on Build GRCh38
rs937274317 Chr.1: 220690604-220690604 on Build GRCh38
rs957953403 Chr.1: 220690961-220690961 on Build GRCh38
rs561375522 Chr.1: 220690839-220690839 on Build GRCh38
rs777409359 Chr.1: 220690599-220690599 on Build GRCh38
rs1001816083 Chr.1: 220690909-220690909 on Build GRCh38
rs772352994 Chr.1: 220690759-220690759 on Build GRCh38
rs1055608198 Chr.1: 220690608-220690608 on Build GRCh38
rs751772788 Chr.1: 220690581-220690582 on Build GRCh38
rs763912380 Chr.1: 220690701-220690701 on Build GRCh38
rs554579162 Chr.1: 220690572-220690572 on Build GRCh38
rs921293693 Chr.1: 220690822-220690822 on Build GRCh38
rs747484133 Chr.1: 220690763-220690763 on Build GRCh38
rs988222523 Chr.1: 220690571-220690571 on Build GRCh38

View Full Product Details