Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 221739243-221739750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP10
Entrez Gene ID
11221
Gene Name
dual specificity phosphatase 10
Gene Aliases
MKP-5, MKP5
UniGene
Hs.497822
Gene Chromosome Location
Chr.1: 221701420-221742176 on Build GRCh38
Transcripts
NM_007207.5 NR_111940.1 XM_017000147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137803 c.55C>T p.R19* Chr.1: 221739690-221739690 on Build GRCh38
4679698 c.319G>A p.G107R Chr.1: 221739426-221739426 on Build GRCh38
1501504 c.471G>C p.K157N Chr.1: 221739274-221739274 on Build GRCh38
4679700 c.250G>A p.A84T Chr.1: 221739495-221739495 on Build GRCh38
7112974 c.67C>G p.L23V Chr.1: 221739678-221739678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759033498 Chr.1: 221739662-221739662 on Build GRCh38
rs910289170 Chr.1: 221739514-221739514 on Build GRCh38
rs752517170 Chr.1: 221739381-221739381 on Build GRCh38
rs371182562 Chr.1: 221739337-221739337 on Build GRCh38
rs199985800 Chr.1: 221739471-221739471 on Build GRCh38
rs778015564 Chr.1: 221739491-221739491 on Build GRCh38
rs760706335 Chr.1: 221739645-221739645 on Build GRCh38
rs1047917515 Chr.1: 221739689-221739689 on Build GRCh38
rs779896271 Chr.1: 221739481-221739481 on Build GRCh38
rs150066182 Chr.1: 221739356-221739356 on Build GRCh38
rs762845278 Chr.1: 221739456-221739456 on Build GRCh38
rs780936253 Chr.1: 221739290-221739290 on Build GRCh38
rs764845200 Chr.1: 221739672-221739672 on Build GRCh38
rs749886489 Chr.1: 221739407-221739407 on Build GRCh38
rs752361079 Chr.1: 221739673-221739673 on Build GRCh38
rs780562110 Chr.1: 221739686-221739686 on Build GRCh38
rs370026750 Chr.1: 221739690-221739690 on Build GRCh38
rs148146409 Chr.1: 221739617-221739617 on Build GRCh38
rs546322180 Chr.1: 221739724-221739724 on Build GRCh38
rs775753885 Chr.1: 221739539-221739539 on Build GRCh38
rs749378098 Chr.1: 221739687-221739687 on Build GRCh38
rs147900867 Chr.1: 221739426-221739426 on Build GRCh38
rs767852820 Chr.1: 221739678-221739678 on Build GRCh38
rs751570837 Chr.1: 221739469-221739469 on Build GRCh38
rs139403142 Chr.1: 221739333-221739333 on Build GRCh38
rs762604027 Chr.1: 221739382-221739382 on Build GRCh38
rs757592372 Chr.1: 221739578-221739578 on Build GRCh38
rs767521771 Chr.1: 221739476-221739476 on Build GRCh38
rs557962750 Chr.1: 221739591-221739591 on Build GRCh38
rs763073068 Chr.1: 221739540-221739540 on Build GRCh38
rs779074182 Chr.1: 221739693-221739693 on Build GRCh38
rs773122250 Chr.1: 221739450-221739450 on Build GRCh38
rs774613067 Chr.1: 221739618-221739618 on Build GRCh38
rs970955138 Chr.1: 221739413-221739413 on Build GRCh38
rs771992135 Chr.1: 221739623-221739623 on Build GRCh38
rs1136448 Chr.1: 221739466-221739466 on Build GRCh38
rs201057695 Chr.1: 221739379-221739379 on Build GRCh38
rs749348862 Chr.1: 221739607-221739607 on Build GRCh38
rs771011005 Chr.1: 221739723-221739723 on Build GRCh38
rs760199414 Chr.1: 221739553-221739553 on Build GRCh38
rs754587624 Chr.1: 221739568-221739568 on Build GRCh38
rs35391196 Chr.1: 221739296-221739297 on Build GRCh38
rs776693283 Chr.1: 221739439-221739439 on Build GRCh38
rs144387908 Chr.1: 221739706-221739706 on Build GRCh38
rs373605299 Chr.1: 221739606-221739606 on Build GRCh38
rs367746036 Chr.1: 221739535-221739535 on Build GRCh38
rs773343058 Chr.1: 221739637-221739637 on Build GRCh38
rs201906952 Chr.1: 221739537-221739537 on Build GRCh38
rs755864933 Chr.1: 221739409-221739409 on Build GRCh38
rs182236491 Chr.1: 221739664-221739664 on Build GRCh38
rs759539849 Chr.1: 221739619-221739619 on Build GRCh38
rs773256376 Chr.1: 221739559-221739559 on Build GRCh38
rs768857289 Chr.1: 221739541-221739541 on Build GRCh38
rs142143528 Chr.1: 221739276-221739276 on Build GRCh38
rs765978282 Chr.1: 221739561-221739561 on Build GRCh38
rs775642653 Chr.1: 221739399-221739399 on Build GRCh38
rs747328324 Chr.1: 221739496-221739496 on Build GRCh38
rs770093063 Chr.1: 221739394-221739394 on Build GRCh38
rs748390873 Chr.1: 221739694-221739694 on Build GRCh38
rs745993241 Chr.1: 221739536-221739536 on Build GRCh38
rs774046325 Chr.1: 221739547-221739547 on Build GRCh38
rs775254813 Chr.1: 221739486-221739486 on Build GRCh38
rs746551024 Chr.1: 221739286-221739286 on Build GRCh38
rs369421974 Chr.1: 221739569-221739569 on Build GRCh38
rs775665938 Chr.1: 221739455-221739455 on Build GRCh38
rs756858765 Chr.1: 221739399-221739399 on Build GRCh38
rs756243800 Chr.1: 221739604-221739604 on Build GRCh38
rs1033573921 Chr.1: 221739432-221739432 on Build GRCh38
rs745510990 Chr.1: 221739319-221739319 on Build GRCh38
rs374050420 Chr.1: 221739283-221739283 on Build GRCh38
rs943040472 Chr.1: 221739598-221739598 on Build GRCh38
rs148718278 Chr.1: 221739417-221739417 on Build GRCh38
rs200957671 Chr.1: 221739389-221739389 on Build GRCh38
rs771407392 Chr.1: 221739438-221739438 on Build GRCh38
rs375651383 Chr.1: 221739622-221739622 on Build GRCh38
rs748576044 Chr.1: 221739424-221739424 on Build GRCh38
rs151204291 Chr.1: 221739427-221739427 on Build GRCh38
rs566662778 Chr.1: 221739266-221739266 on Build GRCh38
rs572257648 Chr.1: 221739477-221739477 on Build GRCh38
rs776900311 Chr.1: 221739358-221739358 on Build GRCh38
rs376468005 Chr.1: 221739494-221739494 on Build GRCh38
rs1051979488 Chr.1: 221739573-221739573 on Build GRCh38
rs368788438 Chr.1: 221739400-221739400 on Build GRCh38
rs371906199 Chr.1: 221739550-221739550 on Build GRCh38
rs756698798 Chr.1: 221739287-221739287 on Build GRCh38
rs757684298 Chr.1: 221739674-221739674 on Build GRCh38
rs368228739 Chr.1: 221739336-221739336 on Build GRCh38
rs924825965 Chr.1: 221739556-221739556 on Build GRCh38
rs753334297 Chr.1: 221739566-221739566 on Build GRCh38
rs144631817 Chr.1: 221739549-221739549 on Build GRCh38
rs750728887 Chr.1: 221739681-221739681 on Build GRCh38
rs373505357 Chr.1: 221739717-221739717 on Build GRCh38
rs773572520 Chr.1: 221739347-221739347 on Build GRCh38
rs201887557 Chr.1: 221739657-221739657 on Build GRCh38
rs747461757 Chr.1: 221739434-221739434 on Build GRCh38
rs755664400 Chr.1: 221739480-221739480 on Build GRCh38
rs781692640 Chr.1: 221739595-221739595 on Build GRCh38
rs189535716 Chr.1: 221739369-221739369 on Build GRCh38
rs376842994 Chr.1: 221739560-221739560 on Build GRCh38
rs138574623 Chr.1: 221739597-221739597 on Build GRCh38
rs770023650 Chr.1: 221739451-221739451 on Build GRCh38
rs766356356 Chr.1: 221739653-221739653 on Build GRCh38
rs772366448 Chr.1: 221739703-221739703 on Build GRCh38
rs771368036 Chr.1: 221739502-221739502 on Build GRCh38

View Full Product Details