Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222687530-222688006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIDA
Entrez Gene ID
64853
Gene Name
axin interactor, dorsalization associated
Gene Aliases
C1orf80
UniGene
Hs.156625
Gene Chromosome Location
Chr.1: 222668013-222712522 on Build GRCh38
Transcripts
NM_022831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3483999 c.325C>T p.P109S Chr.1: 222687623-222687623 on Build GRCh38
4899124 c.253C>T p.P85S Chr.1: 222687623-222687623 on Build GRCh38
4905639 c.263T>C p.V88A Chr.1: 222687613-222687613 on Build GRCh38
2149848 c.291C>G p.I97M Chr.1: 222687657-222687657 on Build GRCh38
4782098 c.219C>G p.I73M Chr.1: 222687657-222687657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967521785 Chr.1: 222687982-222687982 on Build GRCh38
rs770118991 Chr.1: 222687697-222687697 on Build GRCh38
rs746430235 Chr.1: 222687554-222687558 on Build GRCh38
rs1046398457 Chr.1: 222687723-222687723 on Build GRCh38
rs775767459 Chr.1: 222687674-222687677 on Build GRCh38
rs983790251 Chr.1: 222687872-222687872 on Build GRCh38
rs1039652877 Chr.1: 222687905-222687905 on Build GRCh38
rs746246307 Chr.1: 222687685-222687685 on Build GRCh38
rs768776956 Chr.1: 222687570-222687570 on Build GRCh38
rs549554950 Chr.1: 222687754-222687754 on Build GRCh38
rs186621210 Chr.1: 222687741-222687741 on Build GRCh38
rs148997306 Chr.1: 222687579-222687579 on Build GRCh38
rs761676200 Chr.1: 222687808-222687808 on Build GRCh38
rs767171063 Chr.1: 222687600-222687600 on Build GRCh38
rs772251749 Chr.1: 222687580-222687580 on Build GRCh38
rs1020799188 Chr.1: 222687822-222687822 on Build GRCh38
rs1028003224 Chr.1: 222687842-222687842 on Build GRCh38
rs905162724 Chr.1: 222687728-222687728 on Build GRCh38
rs565083327 Chr.1: 222687823-222687823 on Build GRCh38
rs1037842924 Chr.1: 222687773-222687773 on Build GRCh38
rs1054305809 Chr.1: 222687752-222687752 on Build GRCh38
rs778607004 Chr.1: 222687672-222687672 on Build GRCh38
rs919547931 Chr.1: 222687638-222687638 on Build GRCh38
rs780237814 Chr.1: 222687706-222687706 on Build GRCh38
rs972206439 Chr.1: 222687966-222687966 on Build GRCh38
rs760180954 Chr.1: 222687609-222687609 on Build GRCh38
rs566467810 Chr.1: 222687737-222687737 on Build GRCh38
rs991112164 Chr.1: 222687731-222687731 on Build GRCh38
rs961109099 Chr.1: 222687941-222687941 on Build GRCh38
rs375432512 Chr.1: 222687765-222687766 on Build GRCh38
rs754513610 Chr.1: 222687648-222687648 on Build GRCh38
rs774245319 Chr.1: 222687573-222687573 on Build GRCh38
rs200779206 Chr.1: 222687758-222687759 on Build GRCh38
rs143030001 Chr.1: 222687902-222687902 on Build GRCh38
rs941760600 Chr.1: 222687750-222687750 on Build GRCh38
rs1022123670 Chr.1: 222687969-222687969 on Build GRCh38
rs776654377 Chr.1: 222687788-222687788 on Build GRCh38
rs761773618 Chr.1: 222687598-222687598 on Build GRCh38
rs891339216 Chr.1: 222687845-222687845 on Build GRCh38
rs560803025 Chr.1: 222687714-222687714 on Build GRCh38
rs376961021 Chr.1: 222687662-222687662 on Build GRCh38
rs764902652 Chr.1: 222687825-222687825 on Build GRCh38
rs202187548 Chr.1: 222687576-222687576 on Build GRCh38
rs752071735 Chr.1: 222687673-222687673 on Build GRCh38
rs371007133 Chr.1: 222687559-222687559 on Build GRCh38
rs766051663 Chr.1: 222687634-222687634 on Build GRCh38
rs781349921 Chr.1: 222687683-222687683 on Build GRCh38
rs893883894 Chr.1: 222687782-222687782 on Build GRCh38
rs909668590 Chr.1: 222687896-222687896 on Build GRCh38
rs1009440500 Chr.1: 222687801-222687801 on Build GRCh38
rs998476598 Chr.1: 222687831-222687831 on Build GRCh38
rs1036480858 Chr.1: 222687589-222687589 on Build GRCh38
rs967599235 Chr.1: 222687614-222687614 on Build GRCh38
rs757804200 Chr.1: 222687678-222687678 on Build GRCh38
rs953821102 Chr.1: 222687861-222687861 on Build GRCh38
rs753604271 Chr.1: 222687640-222687640 on Build GRCh38
rs376475343 Chr.1: 222687679-222687679 on Build GRCh38

View Full Product Details