Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222694104-222694604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIDA
Entrez Gene ID
64853
Gene Name
axin interactor, dorsalization associated
Gene Aliases
C1orf80
UniGene
Hs.156625
Gene Chromosome Location
Chr.1: 222668013-222712522 on Build GRCh38
Transcripts
NM_022831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
904323 c.195A>C p.K65N Chr.1: 222694249-222694249 on Build GRCh38
7262453 c.119G>A p.G40D Chr.1: 222694253-222694253 on Build GRCh38
268626 c.222T>C p.S74S Chr.1: 222694222-222694222 on Build GRCh38
4859803 c.162+1G>T p.? Chr.1: 222694209-222694209 on Build GRCh38
5911193 c.229T>A p.L77I Chr.1: 222694215-222694215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534018813 Chr.1: 222694325-222694325 on Build GRCh38
rs1050400552 Chr.1: 222694460-222694460 on Build GRCh38
rs777744809 Chr.1: 222694311-222694311 on Build GRCh38
rs932202869 Chr.1: 222694374-222694374 on Build GRCh38
rs913637330 Chr.1: 222694308-222694308 on Build GRCh38
rs539959033 Chr.1: 222694574-222694574 on Build GRCh38
rs776588516 Chr.1: 222694186-222694186 on Build GRCh38
rs946240090 Chr.1: 222694330-222694330 on Build GRCh38
rs752570855 Chr.1: 222694387-222694387 on Build GRCh38
rs199819182 Chr.1: 222694304-222694304 on Build GRCh38
rs201779576 Chr.1: 222694272-222694272 on Build GRCh38
rs977164214 Chr.1: 222694559-222694559 on Build GRCh38
rs924910243 Chr.1: 222694290-222694290 on Build GRCh38
rs746958892 Chr.1: 222694182-222694182 on Build GRCh38
rs964011871 Chr.1: 222694446-222694446 on Build GRCh38
rs779096134 Chr.1: 222694302-222694302 on Build GRCh38
rs74330139 Chr.1: 222694345-222694345 on Build GRCh38
rs372731389 Chr.1: 222694165-222694165 on Build GRCh38
rs1016911265 Chr.1: 222694275-222694276 on Build GRCh38
rs969254232 Chr.1: 222694266-222694266 on Build GRCh38
rs911090567 Chr.1: 222694430-222694430 on Build GRCh38
rs750905527 Chr.1: 222694270-222694270 on Build GRCh38
rs770718554 Chr.1: 222694185-222694185 on Build GRCh38
rs762507637 Chr.1: 222694224-222694224 on Build GRCh38
rs962211695 Chr.1: 222694180-222694180 on Build GRCh38
rs567327136 Chr.1: 222694372-222694372 on Build GRCh38
rs772840394 Chr.1: 222694274-222694275 on Build GRCh38
rs994199795 Chr.1: 222694317-222694317 on Build GRCh38
rs1013408910 Chr.1: 222694205-222694205 on Build GRCh38
rs759205110 Chr.1: 222694187-222694187 on Build GRCh38
rs80287576 Chr.1: 222694577-222694577 on Build GRCh38
rs755403788 Chr.1: 222694291-222694291 on Build GRCh38
rs377168741 Chr.1: 222694209-222694209 on Build GRCh38
rs148556207 Chr.1: 222694520-222694520 on Build GRCh38
rs1006692016 Chr.1: 222694156-222694156 on Build GRCh38
rs764982104 Chr.1: 222694207-222694207 on Build GRCh38
rs887757601 Chr.1: 222694488-222694488 on Build GRCh38
rs754210272 Chr.1: 222694277-222694277 on Build GRCh38
rs986685561 Chr.1: 222694403-222694403 on Build GRCh38
rs367904731 Chr.1: 222694169-222694169 on Build GRCh38
rs897905178 Chr.1: 222694530-222694530 on Build GRCh38
rs763558255 Chr.1: 222694269-222694269 on Build GRCh38
rs758475109 Chr.1: 222694305-222694305 on Build GRCh38
rs190312239 Chr.1: 222694523-222694523 on Build GRCh38

View Full Product Details