Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222715380-222715880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BROX
Entrez Gene ID
148362
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
C1orf58
UniGene
Hs.534965
Gene Chromosome Location
Chr.1: 222712553-222735196 on Build GRCh38
Transcripts
NM_001288579.1 NM_001288580.1 NM_001288581.1 NM_144695.3 XM_005273065.2 XM_005273069.4 XM_006711173.3 XM_011509212.2 XM_011509213.2 XM_011509214.2 XM_017000374.1 XM_017000375.1 XM_017000376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4028532 c.77G>A p.G26D Chr.1: 222715776-222715776 on Build GRCh38
1959986 c.20G>T p.R7M Chr.1: 222715719-222715719 on Build GRCh38
4028531 c.23A>G p.N8S Chr.1: 222715722-222715722 on Build GRCh38
3484000 c.26C>A p.P9Q Chr.1: 222715725-222715725 on Build GRCh38
159546 c.3G>A p.M1I Chr.1: 222715702-222715702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956655908 Chr.1: 222715575-222715575 on Build GRCh38
rs776787088 Chr.1: 222715835-222715835 on Build GRCh38
rs759535708 Chr.1: 222715661-222715661 on Build GRCh38
rs766520579 Chr.1: 222715756-222715756 on Build GRCh38
rs926878548 Chr.1: 222715462-222715462 on Build GRCh38
rs922668938 Chr.1: 222715755-222715755 on Build GRCh38
rs756686374 Chr.1: 222715800-222715800 on Build GRCh38
rs560542421 Chr.1: 222715782-222715782 on Build GRCh38
rs908001907 Chr.1: 222715720-222715720 on Build GRCh38
rs149650233 Chr.1: 222715429-222715429 on Build GRCh38
rs778432196 Chr.1: 222715445-222715445 on Build GRCh38
rs527675627 Chr.1: 222715826-222715826 on Build GRCh38
rs746709099 Chr.1: 222715795-222715795 on Build GRCh38
rs779722702 Chr.1: 222715522-222715522 on Build GRCh38
rs967897202 Chr.1: 222715533-222715533 on Build GRCh38
rs1000718400 Chr.1: 222715536-222715536 on Build GRCh38
rs752216787 Chr.1: 222715670-222715673 on Build GRCh38
rs774871995 Chr.1: 222715723-222715723 on Build GRCh38
rs756439103 Chr.1: 222715436-222715436 on Build GRCh38
rs749838169 Chr.1: 222715714-222715714 on Build GRCh38
rs753864297 Chr.1: 222715668-222715668 on Build GRCh38
rs749705248 Chr.1: 222715806-222715806 on Build GRCh38
rs1057116518 Chr.1: 222715771-222715771 on Build GRCh38
rs981407159 Chr.1: 222715456-222715456 on Build GRCh38
rs374196579 Chr.1: 222715834-222715834 on Build GRCh38
rs773630926 Chr.1: 222715742-222715742 on Build GRCh38
rs764952738 Chr.1: 222715773-222715773 on Build GRCh38
rs1024079877 Chr.1: 222715598-222715598 on Build GRCh38
rs765153855 Chr.1: 222715838-222715838 on Build GRCh38
rs919661073 Chr.1: 222715645-222715645 on Build GRCh38
rs752490722 Chr.1: 222715778-222715778 on Build GRCh38
rs750156826 Chr.1: 222715478-222715478 on Build GRCh38
rs748514779 Chr.1: 222715822-222715822 on Build GRCh38
rs1049677900 Chr.1: 222715423-222715423 on Build GRCh38
rs1012377844 Chr.1: 222715512-222715512 on Build GRCh38
rs754698088 Chr.1: 222715674-222715674 on Build GRCh38
rs539504411 Chr.1: 222715446-222715446 on Build GRCh38
rs762063391 Chr.1: 222715726-222715726 on Build GRCh38
rs774313401 Chr.1: 222715657-222715657 on Build GRCh38
rs545143655 Chr.1: 222715688-222715688 on Build GRCh38
rs952990625 Chr.1: 222715662-222715662 on Build GRCh38
rs555367823 Chr.1: 222715537-222715537 on Build GRCh38
rs367964617 Chr.1: 222715717-222715717 on Build GRCh38
rs368572779 Chr.1: 222715706-222715706 on Build GRCh38
rs111256107 Chr.1: 222715680-222715680 on Build GRCh38
rs912267326 Chr.1: 222715591-222715591 on Build GRCh38
rs543983010 Chr.1: 222715595-222715595 on Build GRCh38
rs780782954 Chr.1: 222715805-222715805 on Build GRCh38
rs963675232 Chr.1: 222715633-222715633 on Build GRCh38
rs760989297 Chr.1: 222715832-222715832 on Build GRCh38
rs775067918 Chr.1: 222715818-222715818 on Build GRCh38
rs755699412 Chr.1: 222715799-222715802 on Build GRCh38
rs932516549 Chr.1: 222715546-222715546 on Build GRCh38
rs370523578 Chr.1: 222715761-222715761 on Build GRCh38
rs930801860 Chr.1: 222715469-222715469 on Build GRCh38
rs374313797 Chr.1: 222715415-222715415 on Build GRCh38
rs975163619 Chr.1: 222715643-222715643 on Build GRCh38
rs940956711 Chr.1: 222715746-222715746 on Build GRCh38
rs750538251 Chr.1: 222715651-222715651 on Build GRCh38
rs904424327 Chr.1: 222715554-222715554 on Build GRCh38
rs1049326683 Chr.1: 222715474-222715474 on Build GRCh38
rs534105904 Chr.1: 222715527-222715527 on Build GRCh38
rs759322783 Chr.1: 222715836-222715836 on Build GRCh38
rs778801048 Chr.1: 222715676-222715676 on Build GRCh38
rs770453035 Chr.1: 222715764-222715764 on Build GRCh38
rs756332371 Chr.1: 222715655-222715655 on Build GRCh38
rs557754008 Chr.1: 222715450-222715450 on Build GRCh38
rs183027373 Chr.1: 222715579-222715579 on Build GRCh38
rs570393869 Chr.1: 222715617-222715617 on Build GRCh38
rs752688870 Chr.1: 222715842-222715842 on Build GRCh38
rs1014052764 Chr.1: 222715438-222715438 on Build GRCh38
rs753680507 Chr.1: 222715760-222715760 on Build GRCh38
rs757125276 Chr.1: 222715433-222715433 on Build GRCh38
rs556275061 Chr.1: 222715657-222715657 on Build GRCh38
rs763840640 Chr.1: 222715840-222715840 on Build GRCh38
rs767851376 Chr.1: 222715737-222715737 on Build GRCh38
rs896513333 Chr.1: 222715810-222715810 on Build GRCh38
rs777196603 Chr.1: 222715683-222715683 on Build GRCh38
rs190839788 Chr.1: 222715495-222715495 on Build GRCh38
rs1005516822 Chr.1: 222715480-222715480 on Build GRCh38
rs767001904 Chr.1: 222715661-222715662 on Build GRCh38
rs746450676 Chr.1: 222715649-222715649 on Build GRCh38
rs760649298 Chr.1: 222715748-222715748 on Build GRCh38
rs186130053 Chr.1: 222715435-222715435 on Build GRCh38
rs896439781 Chr.1: 222715496-222715496 on Build GRCh38
rs1050047037 Chr.1: 222715546-222715547 on Build GRCh38
rs550497917 Chr.1: 222715461-222715461 on Build GRCh38
rs935949165 Chr.1: 222715570-222715570 on Build GRCh38
rs999844036 Chr.1: 222715432-222715432 on Build GRCh38
rs984132595 Chr.1: 222715490-222715490 on Build GRCh38
rs772618131 Chr.1: 222715823-222715823 on Build GRCh38
rs769213199 Chr.1: 222715813-222715813 on Build GRCh38
rs777389198 Chr.1: 222715788-222715788 on Build GRCh38
rs778372248 Chr.1: 222715821-222715821 on Build GRCh38

View Full Product Details