Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222718846-222719349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BROX
Entrez Gene ID
148362
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
C1orf58
UniGene
Hs.534965
Gene Chromosome Location
Chr.1: 222712553-222735196 on Build GRCh38
Transcripts
NM_001288579.1 NM_001288580.1 NM_001288581.1 NM_144695.3 XM_005273065.2 XM_005273069.4 XM_006711173.3 XM_011509212.2 XM_011509213.2 XM_011509214.2 XM_017000374.1 XM_017000375.1 XM_017000376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687179 c.145G>C p.D49H Chr.1: 222718968-222718968 on Build GRCh38
904324 c.221C>A p.S74Y Chr.1: 222719275-222719275 on Build GRCh38
6541803 c.180A>T p.A60A Chr.1: 222719003-222719003 on Build GRCh38
6612896 c.241G>T p.E81* Chr.1: 222719295-222719295 on Build GRCh38
4531018 c.174G>C p.K58N Chr.1: 222718997-222718997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051619119 Chr.1: 222719209-222719209 on Build GRCh38
rs368370531 Chr.1: 222718883-222718883 on Build GRCh38
rs1019430845 Chr.1: 222719149-222719149 on Build GRCh38
rs557135868 Chr.1: 222719068-222719068 on Build GRCh38
rs779189209 Chr.1: 222719032-222719035 on Build GRCh38
rs767260851 Chr.1: 222719071-222719071 on Build GRCh38
rs760338078 Chr.1: 222719294-222719294 on Build GRCh38
rs759093572 Chr.1: 222719311-222719311 on Build GRCh38
rs756954881 Chr.1: 222718887-222718887 on Build GRCh38
rs1013486623 Chr.1: 222718984-222718984 on Build GRCh38
rs903013176 Chr.1: 222719134-222719134 on Build GRCh38
rs749162851 Chr.1: 222719063-222719063 on Build GRCh38
rs758989209 Chr.1: 222719004-222719004 on Build GRCh38
rs765931382 Chr.1: 222719220-222719220 on Build GRCh38
rs201755100 Chr.1: 222718914-222718914 on Build GRCh38
rs950792559 Chr.1: 222719099-222719099 on Build GRCh38
rs113851318 Chr.1: 222719206-222719206 on Build GRCh38
rs541007138 Chr.1: 222719252-222719252 on Build GRCh38
rs771188237 Chr.1: 222718933-222718933 on Build GRCh38
rs779742969 Chr.1: 222719001-222719001 on Build GRCh38
rs373129795 Chr.1: 222719243-222719243 on Build GRCh38
rs369104334 Chr.1: 222719281-222719281 on Build GRCh38
rs963468609 Chr.1: 222719249-222719249 on Build GRCh38
rs764647410 Chr.1: 222719232-222719232 on Build GRCh38
rs542099510 Chr.1: 222719313-222719313 on Build GRCh38
rs192920101 Chr.1: 222718878-222718878 on Build GRCh38
rs772880582 Chr.1: 222719074-222719074 on Build GRCh38
rs372097102 Chr.1: 222718913-222718913 on Build GRCh38
rs964922635 Chr.1: 222718912-222718912 on Build GRCh38
rs370537093 Chr.1: 222719186-222719186 on Build GRCh38
rs569689181 Chr.1: 222718936-222718936 on Build GRCh38
rs1044575249 Chr.1: 222718997-222718997 on Build GRCh38
rs747393706 Chr.1: 222719035-222719035 on Build GRCh38
rs903106770 Chr.1: 222718868-222718868 on Build GRCh38
rs746115476 Chr.1: 222719054-222719054 on Build GRCh38
rs143269651 Chr.1: 222718872-222718872 on Build GRCh38
rs189010024 Chr.1: 222719127-222719127 on Build GRCh38
rs553424529 Chr.1: 222718989-222718989 on Build GRCh38
rs148199221 Chr.1: 222719274-222719274 on Build GRCh38
rs774215997 Chr.1: 222719069-222719069 on Build GRCh38
rs776481981 Chr.1: 222719308-222719308 on Build GRCh38
rs568430174 Chr.1: 222718993-222718993 on Build GRCh38
rs964854241 Chr.1: 222719150-222719150 on Build GRCh38
rs781437594 Chr.1: 222719245-222719245 on Build GRCh38
rs767188717 Chr.1: 222718991-222718991 on Build GRCh38
rs951510653 Chr.1: 222719287-222719287 on Build GRCh38
rs758120296 Chr.1: 222719070-222719070 on Build GRCh38
rs138642708 Chr.1: 222719106-222719106 on Build GRCh38
rs373708090 Chr.1: 222719307-222719307 on Build GRCh38
rs779769628 Chr.1: 222719310-222719310 on Build GRCh38
rs149121376 Chr.1: 222719002-222719002 on Build GRCh38
rs928646385 Chr.1: 222718987-222718987 on Build GRCh38
rs768669947 Chr.1: 222719258-222719258 on Build GRCh38
rs754600462 Chr.1: 222719228-222719228 on Build GRCh38
rs371765877 Chr.1: 222718922-222718922 on Build GRCh38
rs368833998 Chr.1: 222718917-222718917 on Build GRCh38
rs752054288 Chr.1: 222719239-222719239 on Build GRCh38
rs375053073 Chr.1: 222718947-222718947 on Build GRCh38
rs757624267 Chr.1: 222718970-222718970 on Build GRCh38
rs974566953 Chr.1: 222719200-222719200 on Build GRCh38
rs200062613 Chr.1: 222719259-222719259 on Build GRCh38
rs754105375 Chr.1: 222718919-222718920 on Build GRCh38
rs1041944261 Chr.1: 222719132-222719132 on Build GRCh38
rs778306632 Chr.1: 222718923-222718923 on Build GRCh38
rs145225286 Chr.1: 222719248-222719248 on Build GRCh38
rs73124895 Chr.1: 222718940-222718940 on Build GRCh38
rs748070974 Chr.1: 222719273-222719273 on Build GRCh38
rs947458557 Chr.1: 222719130-222719130 on Build GRCh38
rs570798531 Chr.1: 222718965-222718965 on Build GRCh38
rs777935159 Chr.1: 222719032-222719032 on Build GRCh38
rs73124896 Chr.1: 222718983-222718983 on Build GRCh38
rs773069171 Chr.1: 222719135-222719135 on Build GRCh38
rs113449964 Chr.1: 222719318-222719318 on Build GRCh38
rs747143013 Chr.1: 222718928-222718928 on Build GRCh38
rs939085791 Chr.1: 222718948-222718948 on Build GRCh38
rs763913738 Chr.1: 222718882-222718882 on Build GRCh38
rs781452943 Chr.1: 222719051-222719051 on Build GRCh38
rs200655590 Chr.1: 222718920-222718920 on Build GRCh38
rs1056634679 Chr.1: 222718959-222718959 on Build GRCh38
rs769820232 Chr.1: 222719056-222719056 on Build GRCh38
rs146216028 Chr.1: 222719312-222719312 on Build GRCh38
rs762904375 Chr.1: 222718971-222718971 on Build GRCh38
rs377402054 Chr.1: 222719247-222719247 on Build GRCh38
rs775371560 Chr.1: 222718963-222718963 on Build GRCh38
rs749358803 Chr.1: 222719253-222719253 on Build GRCh38
rs953041609 Chr.1: 222718901-222718901 on Build GRCh38
rs779544274 Chr.1: 222718921-222718921 on Build GRCh38
rs753405978 Chr.1: 222719225-222719225 on Build GRCh38
rs760427631 Chr.1: 222719076-222719076 on Build GRCh38

View Full Product Details