Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222746428-222746975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177B
Entrez Gene ID
400823
Gene Name
family with sequence similarity 177 member B
Gene Aliases
-
UniGene
Hs.697608
Gene Chromosome Location
Chr.1: 222737216-222772193 on Build GRCh38
Transcripts
NM_001324080.1 NM_207468.2 NR_136691.1 XM_017001279.1 XM_017001280.1 XM_017001281.1 XM_017001282.1 XM_017001283.1 XM_017001284.1 XM_017001285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4557247 c.71G>A p.R24K Chr.1: 222746616-222746616 on Build GRCh38
6192169 c.12C>G p.D4E Chr.1: 222746557-222746557 on Build GRCh38
4455165 c.68A>T p.K23I Chr.1: 222746613-222746613 on Build GRCh38
3400357 c.89A>G p.D30G Chr.1: 222746634-222746634 on Build GRCh38
3484003 c.14G>A p.G5D Chr.1: 222746559-222746559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747074182 Chr.1: 222746578-222746578 on Build GRCh38
rs887023456 Chr.1: 222746453-222746453 on Build GRCh38
rs760292499 Chr.1: 222746766-222746766 on Build GRCh38
rs1024160448 Chr.1: 222746736-222746738 on Build GRCh38
rs141453662 Chr.1: 222746557-222746557 on Build GRCh38
rs143780662 Chr.1: 222746687-222746687 on Build GRCh38
rs749013228 Chr.1: 222746738-222746738 on Build GRCh38
rs528737246 Chr.1: 222746810-222746810 on Build GRCh38
rs111682421 Chr.1: 222746513-222746514 on Build GRCh38
rs752994188 Chr.1: 222746660-222746660 on Build GRCh38
rs756885886 Chr.1: 222746537-222746537 on Build GRCh38
rs920017946 Chr.1: 222746818-222746818 on Build GRCh38
rs569763698 Chr.1: 222746912-222746912 on Build GRCh38
rs190168272 Chr.1: 222746926-222746926 on Build GRCh38
rs1001038635 Chr.1: 222746606-222746606 on Build GRCh38
rs755321427 Chr.1: 222746548-222746548 on Build GRCh38
rs775156920 Chr.1: 222746509-222746509 on Build GRCh38
rs754340760 Chr.1: 222746651-222746651 on Build GRCh38
rs750765402 Chr.1: 222746607-222746609 on Build GRCh38
rs771642809 Chr.1: 222746763-222746763 on Build GRCh38
rs371611917 Chr.1: 222746558-222746558 on Build GRCh38
rs982597849 Chr.1: 222746854-222746854 on Build GRCh38
rs1056828380 Chr.1: 222746481-222746481 on Build GRCh38
rs2378607 Chr.1: 222746553-222746553 on Build GRCh38
rs1002566309 Chr.1: 222746468-222746468 on Build GRCh38
rs761368397 Chr.1: 222746647-222746647 on Build GRCh38
rs1031142586 Chr.1: 222746928-222746928 on Build GRCh38
rs955891843 Chr.1: 222746700-222746700 on Build GRCh38
rs372627601 Chr.1: 222746743-222746743 on Build GRCh38
rs202070793 Chr.1: 222746710-222746710 on Build GRCh38
rs763385193 Chr.1: 222746663-222746665 on Build GRCh38
rs754428668 Chr.1: 222746542-222746542 on Build GRCh38
rs190923475 Chr.1: 222746511-222746511 on Build GRCh38
rs569893766 Chr.1: 222746904-222746904 on Build GRCh38
rs780096121 Chr.1: 222746730-222746730 on Build GRCh38
rs186709458 Chr.1: 222746883-222746883 on Build GRCh38
rs956070500 Chr.1: 222746472-222746472 on Build GRCh38
rs774098514 Chr.1: 222746758-222746758 on Build GRCh38
rs752101641 Chr.1: 222746709-222746710 on Build GRCh38
rs1000242408 Chr.1: 222746684-222746684 on Build GRCh38
rs530297618 Chr.1: 222746872-222746872 on Build GRCh38
rs938596729 Chr.1: 222746870-222746870 on Build GRCh38
rs756141837 Chr.1: 222746882-222746882 on Build GRCh38
rs761571195 Chr.1: 222746762-222746762 on Build GRCh38
rs768179396 Chr.1: 222746636-222746636 on Build GRCh38
rs200760779 Chr.1: 222746698-222746698 on Build GRCh38
rs547724622 Chr.1: 222746799-222746799 on Build GRCh38
rs367930283 Chr.1: 222746648-222746648 on Build GRCh38
rs777944958 Chr.1: 222746685-222746685 on Build GRCh38
rs771042451 Chr.1: 222746588-222746588 on Build GRCh38
rs555768490 Chr.1: 222746508-222746508 on Build GRCh38
rs763592282 Chr.1: 222746515-222746515 on Build GRCh38
rs992756977 Chr.1: 222746476-222746476 on Build GRCh38
rs140872706 Chr.1: 222746623-222746623 on Build GRCh38
rs138554889 Chr.1: 222746656-222746656 on Build GRCh38
rs781063729 Chr.1: 222746596-222746596 on Build GRCh38
rs368688839 Chr.1: 222746844-222746844 on Build GRCh38
rs368658830 Chr.1: 222746538-222746538 on Build GRCh38
rs577562452 Chr.1: 222746510-222746510 on Build GRCh38
rs904253589 Chr.1: 222746921-222746921 on Build GRCh38
rs920220615 Chr.1: 222746783-222746783 on Build GRCh38
rs745824339 Chr.1: 222746597-222746597 on Build GRCh38
rs371775222 Chr.1: 222746661-222746661 on Build GRCh38
rs374982213 Chr.1: 222746640-222746640 on Build GRCh38
rs908343792 Chr.1: 222746865-222746865 on Build GRCh38
rs772846977 Chr.1: 222746765-222746765 on Build GRCh38
rs1004820509 Chr.1: 222746576-222746576 on Build GRCh38
rs769572528 Chr.1: 222746608-222746608 on Build GRCh38
rs762871916 Chr.1: 222746635-222746635 on Build GRCh38
rs751109004 Chr.1: 222746520-222746520 on Build GRCh38
rs766889802 Chr.1: 222746671-222746672 on Build GRCh38
rs199959781 Chr.1: 222746699-222746699 on Build GRCh38
rs778118066 Chr.1: 222746567-222746567 on Build GRCh38

View Full Product Details