Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223550568-223551067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN8
Entrez Gene ID
388743
Gene Name
calpain 8
Gene Aliases
nCL-2
UniGene
Hs.291487
Gene Chromosome Location
Chr.1: 223541609-223665734 on Build GRCh38
Transcripts
NM_001143962.1 XM_017001265.1 XM_017001266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4439027 c.58+1G>A p.? Chr.1: 223550959-223550959 on Build GRCh38
3943489 c.34C>A p.L12I Chr.1: 223550984-223550984 on Build GRCh38
904415 c.48G>A p.A16A Chr.1: 223550970-223550970 on Build GRCh38
6880466 c.46G>A p.A16T Chr.1: 223550972-223550972 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368722561 Chr.1: 223550779-223550779 on Build GRCh38
rs768376231 Chr.1: 223550892-223550892 on Build GRCh38
rs993469669 Chr.1: 223550775-223550775 on Build GRCh38
rs73130104 Chr.1: 223550629-223550629 on Build GRCh38
rs888539365 Chr.1: 223550991-223550991 on Build GRCh38
rs569437573 Chr.1: 223550706-223550706 on Build GRCh38
rs534192162 Chr.1: 223550771-223550771 on Build GRCh38
rs541695102 Chr.1: 223550885-223550885 on Build GRCh38
rs201599470 Chr.1: 223550980-223550980 on Build GRCh38
rs997259309 Chr.1: 223550867-223550867 on Build GRCh38
rs74145912 Chr.1: 223550683-223550683 on Build GRCh38
rs189736783 Chr.1: 223550729-223550729 on Build GRCh38
rs911058128 Chr.1: 223550901-223550901 on Build GRCh38
rs1034446964 Chr.1: 223550751-223550751 on Build GRCh38
rs982643368 Chr.1: 223550753-223550753 on Build GRCh38
rs1034504016 Chr.1: 223551000-223551000 on Build GRCh38
rs1035979567 Chr.1: 223550856-223550856 on Build GRCh38
rs546418011 Chr.1: 223550612-223550612 on Build GRCh38
rs1026163104 Chr.1: 223550735-223550735 on Build GRCh38
rs756239797 Chr.1: 223551039-223551039 on Build GRCh38
rs958985723 Chr.1: 223550759-223550759 on Build GRCh38
rs1035989471 Chr.1: 223550621-223550621 on Build GRCh38
rs575069110 Chr.1: 223550998-223550998 on Build GRCh38
rs751681462 Chr.1: 223550975-223550975 on Build GRCh38
rs577911219 Chr.1: 223550695-223550695 on Build GRCh38
rs544307376 Chr.1: 223550780-223550780 on Build GRCh38
rs929987367 Chr.1: 223550952-223550952 on Build GRCh38
rs1042517695 Chr.1: 223550607-223550607 on Build GRCh38
rs182999659 Chr.1: 223550748-223550748 on Build GRCh38
rs750494639 Chr.1: 223551020-223551020 on Build GRCh38
rs865782707 Chr.1: 223550653-223550653 on Build GRCh38
rs574060323 Chr.1: 223550882-223550882 on Build GRCh38
rs926793447 Chr.1: 223550754-223550754 on Build GRCh38
rs977761029 Chr.1: 223550849-223550849 on Build GRCh38
rs188090739 Chr.1: 223550927-223550927 on Build GRCh38
rs555629831 Chr.1: 223550778-223550778 on Build GRCh38
rs540156600 Chr.1: 223550726-223550726 on Build GRCh38
rs139868535 Chr.1: 223550731-223550742 on Build GRCh38
rs776390909 Chr.1: 223550628-223550628 on Build GRCh38
rs940286401 Chr.1: 223550823-223550823 on Build GRCh38
rs942618392 Chr.1: 223550906-223550906 on Build GRCh38
rs923685752 Chr.1: 223550864-223550864 on Build GRCh38
rs952775862 Chr.1: 223550736-223550736 on Build GRCh38
rs373430013 Chr.1: 223550972-223550972 on Build GRCh38
rs75802707 Chr.1: 223550970-223550970 on Build GRCh38
rs763344061 Chr.1: 223550910-223550910 on Build GRCh38
rs781240926 Chr.1: 223550980-223550980 on Build GRCh38
rs762312391 Chr.1: 223550979-223550979 on Build GRCh38
rs888886413 Chr.1: 223551010-223551010 on Build GRCh38
rs71644399 Chr.1: 223550750-223550750 on Build GRCh38
rs74145911 Chr.1: 223550680-223550680 on Build GRCh38
rs1030163506 Chr.1: 223550661-223550661 on Build GRCh38
rs200488592 Chr.1: 223550772-223550772 on Build GRCh38
rs1041349039 Chr.1: 223550926-223550926 on Build GRCh38

View Full Product Details