Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225487518-225488028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENAH
Entrez Gene ID
55740
Gene Name
enabled homolog (Drosophila)
Gene Aliases
ENA, MENA, NDPP1
UniGene
Hs.497893
Gene Chromosome Location
Chr.1: 225486832-225660045 on Build GRCh38
Transcripts
NM_001008493.1 NM_018212.4 XM_005273187.2 XM_017001746.1 XM_017001751.1 XM_017001753.1 XM_017001754.1 XM_017001755.1 XM_017001756.1 XM_017001757.1

SNP Detail

SNP ID Chromosome Location
rs1036852100 Chr.1: 225487939-225487939 on Build GRCh38
rs926282905 Chr.1: 225487843-225487843 on Build GRCh38
rs922534658 Chr.1: 225487942-225487943 on Build GRCh38
rs181109591 Chr.1: 225487830-225487830 on Build GRCh38
rs4653643 Chr.1: 225487757-225487757 on Build GRCh38
rs765147511 Chr.1: 225487655-225487655 on Build GRCh38
rs1051746898 Chr.1: 225487717-225487717 on Build GRCh38
rs952952775 Chr.1: 225487690-225487690 on Build GRCh38
rs145346102 Chr.1: 225487897-225487897 on Build GRCh38
rs958452974 Chr.1: 225487804-225487804 on Build GRCh38
rs571677445 Chr.1: 225487802-225487802 on Build GRCh38
rs1041505846 Chr.1: 225487826-225487826 on Build GRCh38
rs756067421 Chr.1: 225487801-225487801 on Build GRCh38
rs945691552 Chr.1: 225487786-225487786 on Build GRCh38
rs553500521 Chr.1: 225487760-225487760 on Build GRCh38
rs1005800621 Chr.1: 225487918-225487918 on Build GRCh38
rs959035274 Chr.1: 225487885-225487886 on Build GRCh38
rs56181999 Chr.1: 225487593-225487593 on Build GRCh38
rs901284618 Chr.1: 225487827-225487827 on Build GRCh38
rs1009719361 Chr.1: 225487995-225487995 on Build GRCh38
rs763274351 Chr.1: 225487734-225487734 on Build GRCh38
rs762974685 Chr.1: 225487705-225487705 on Build GRCh38
rs925850131 Chr.1: 225487797-225487797 on Build GRCh38
rs992988120 Chr.1: 225487759-225487759 on Build GRCh38
rs879708496 Chr.1: 225487987-225487987 on Build GRCh38
rs113500329 Chr.1: 225487944-225487944 on Build GRCh38
rs889247991 Chr.1: 225487726-225487726 on Build GRCh38
rs901060180 Chr.1: 225487707-225487707 on Build GRCh38
rs997544831 Chr.1: 225487715-225487715 on Build GRCh38
rs1054300848 Chr.1: 225487976-225487981 on Build GRCh38
rs1050182235 Chr.1: 225487803-225487803 on Build GRCh38
rs757679262 Chr.1: 225487975-225487975 on Build GRCh38
rs578189477 Chr.1: 225487694-225487694 on Build GRCh38
rs888939064 Chr.1: 225487819-225487819 on Build GRCh38
rs1036967183 Chr.1: 225487656-225487656 on Build GRCh38
rs901167179 Chr.1: 225487545-225487545 on Build GRCh38
rs909713767 Chr.1: 225487606-225487606 on Build GRCh38
rs944595975 Chr.1: 225487822-225487822 on Build GRCh38
rs531444445 Chr.1: 225487993-225487993 on Build GRCh38
rs1034527307 Chr.1: 225487713-225487713 on Build GRCh38
rs1034642090 Chr.1: 225487922-225487922 on Build GRCh38
rs933891741 Chr.1: 225487943-225487943 on Build GRCh38
rs888668022 Chr.1: 225487911-225487911 on Build GRCh38
rs1051801910 Chr.1: 225487943-225487943 on Build GRCh38
rs977071074 Chr.1: 225487789-225487789 on Build GRCh38

View Full Product Details