Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225489640-225490184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENAH
Entrez Gene ID
55740
Gene Name
enabled homolog (Drosophila)
Gene Aliases
ENA, MENA, NDPP1
UniGene
Hs.497893
Gene Chromosome Location
Chr.1: 225486832-225660045 on Build GRCh38
Transcripts
NM_001008493.1 NM_018212.4 XM_005273187.2 XM_017001746.1 XM_017001751.1 XM_017001753.1 XM_017001754.1 XM_017001755.1 XM_017001756.1 XM_017001757.1

SNP Detail

SNP ID Chromosome Location
rs532643561 Chr.1: 225489908-225489908 on Build GRCh38
rs887223820 Chr.1: 225490018-225490019 on Build GRCh38
rs868102949 Chr.1: 225489674-225489674 on Build GRCh38
rs775405957 Chr.1: 225489676-225489676 on Build GRCh38
rs964777261 Chr.1: 225490106-225490106 on Build GRCh38
rs1020191873 Chr.1: 225489785-225489785 on Build GRCh38
rs947973220 Chr.1: 225489966-225489966 on Build GRCh38
rs954845129 Chr.1: 225490147-225490147 on Build GRCh38
rs1002904420 Chr.1: 225489824-225489824 on Build GRCh38
rs971472183 Chr.1: 225489804-225489804 on Build GRCh38
rs550040306 Chr.1: 225489758-225489758 on Build GRCh38
rs768363103 Chr.1: 225489805-225489805 on Build GRCh38
rs550308449 Chr.1: 225489680-225489680 on Build GRCh38
rs968925645 Chr.1: 225490074-225490075 on Build GRCh38
rs1011493381 Chr.1: 225490073-225490073 on Build GRCh38
rs577231409 Chr.1: 225490074-225490077 on Build GRCh38
rs1056998520 Chr.1: 225489949-225489954 on Build GRCh38
rs965336482 Chr.1: 225489879-225489879 on Build GRCh38
rs116114741 Chr.1: 225489959-225489959 on Build GRCh38
rs767517063 Chr.1: 225489972-225489972 on Build GRCh38
rs773372170 Chr.1: 225489675-225489675 on Build GRCh38
rs373249030 Chr.1: 225489767-225489767 on Build GRCh38
rs907652378 Chr.1: 225489927-225489927 on Build GRCh38
rs1031292377 Chr.1: 225490114-225490115 on Build GRCh38
rs1047547976 Chr.1: 225489711-225489711 on Build GRCh38
rs1008321589 Chr.1: 225490040-225490040 on Build GRCh38
rs766920090 Chr.1: 225490132-225490132 on Build GRCh38
rs915641276 Chr.1: 225490152-225490152 on Build GRCh38
rs956027762 Chr.1: 225489863-225489863 on Build GRCh38
rs974906723 Chr.1: 225489884-225489884 on Build GRCh38
rs975399320 Chr.1: 225490126-225490126 on Build GRCh38
rs924389901 Chr.1: 225489946-225489946 on Build GRCh38
rs1031755222 Chr.1: 225489831-225489831 on Build GRCh38
rs976071209 Chr.1: 225489868-225489868 on Build GRCh38
rs1025231072 Chr.1: 225489778-225489778 on Build GRCh38
rs900015954 Chr.1: 225490052-225490052 on Build GRCh38
rs914855665 Chr.1: 225489672-225489672 on Build GRCh38
rs35767573 Chr.1: 225489962-225489962 on Build GRCh38
rs796421632 Chr.1: 225489761-225489761 on Build GRCh38
rs933074756 Chr.1: 225489702-225489702 on Build GRCh38
rs763178053 Chr.1: 225489706-225489706 on Build GRCh38
rs767443612 Chr.1: 225490143-225490143 on Build GRCh38
rs565368819 Chr.1: 225489875-225489875 on Build GRCh38
rs868374262 Chr.1: 225489766-225489766 on Build GRCh38
rs977045157 Chr.1: 225489936-225489936 on Build GRCh38
rs531707902 Chr.1: 225489747-225489747 on Build GRCh38
rs992991349 Chr.1: 225490150-225490150 on Build GRCh38
rs1022325009 Chr.1: 225490071-225490071 on Build GRCh38
rs964208615 Chr.1: 225489842-225489842 on Build GRCh38
rs553218035 Chr.1: 225490114-225490114 on Build GRCh38
rs146691918 Chr.1: 225490079-225490079 on Build GRCh38
rs923427902 Chr.1: 225489896-225489896 on Build GRCh38
rs898493493 Chr.1: 225489687-225489687 on Build GRCh38
rs192177970 Chr.1: 225490094-225490094 on Build GRCh38
rs561289273 Chr.1: 225489700-225489700 on Build GRCh38
rs899068252 Chr.1: 225489974-225489974 on Build GRCh38
rs761156692 Chr.1: 225490110-225490110 on Build GRCh38
rs886105737 Chr.1: 225489746-225489746 on Build GRCh38
rs537037297 Chr.1: 225490017-225490017 on Build GRCh38
rs773346215 Chr.1: 225490032-225490032 on Build GRCh38
rs996995165 Chr.1: 225490070-225490070 on Build GRCh38
rs999344933 Chr.1: 225489978-225489978 on Build GRCh38
rs941646326 Chr.1: 225489935-225489935 on Build GRCh38
rs145844582 Chr.1: 225489921-225489921 on Build GRCh38
rs187326341 Chr.1: 225489975-225489975 on Build GRCh38
rs990122558 Chr.1: 225489864-225489864 on Build GRCh38
rs182677340 Chr.1: 225489947-225489947 on Build GRCh38
rs558411889 Chr.1: 225490043-225490043 on Build GRCh38

View Full Product Details