Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225999115-225999610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDE2
Entrez Gene ID
163859
Gene Name
SDE2 telomere maintenance homolog
Gene Aliases
C1orf55, dJ671D7.1
UniGene
Hs.520192
Gene Chromosome Location
Chr.1: 225982702-225999365 on Build GRCh38
Transcripts
NM_152608.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4888442 c.80G>T p.C27F Chr.1: 225999233-225999233 on Build GRCh38
4646899 c.36C>T p.G12G Chr.1: 225999277-225999277 on Build GRCh38
3741055 c.103C>A p.R35R Chr.1: 225999210-225999210 on Build GRCh38
5701094 c.35_36insG p.G14fs*44 Chr.1: 225999277-225999278 on Build GRCh38
6267070 c.42C>A p.G14G Chr.1: 225999271-225999271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183044649 Chr.1: 225999136-225999136 on Build GRCh38
rs1037318551 Chr.1: 225999230-225999230 on Build GRCh38
rs953775801 Chr.1: 225999408-225999431 on Build GRCh38
rs746558864 Chr.1: 225999321-225999321 on Build GRCh38
rs376616547 Chr.1: 225999142-225999142 on Build GRCh38
rs913176329 Chr.1: 225999284-225999284 on Build GRCh38
rs1035497909 Chr.1: 225999276-225999276 on Build GRCh38
rs776562741 Chr.1: 225999354-225999354 on Build GRCh38
rs754823765 Chr.1: 225999313-225999313 on Build GRCh38
rs918852436 Chr.1: 225999518-225999518 on Build GRCh38
rs1006653867 Chr.1: 225999456-225999456 on Build GRCh38
rs967094519 Chr.1: 225999351-225999351 on Build GRCh38
rs767777681 Chr.1: 225999304-225999304 on Build GRCh38
rs776057731 Chr.1: 225999234-225999234 on Build GRCh38
rs942774618 Chr.1: 225999572-225999572 on Build GRCh38
rs937362613 Chr.1: 225999538-225999538 on Build GRCh38
rs367813006 Chr.1: 225999312-225999312 on Build GRCh38
rs764737370 Chr.1: 225999340-225999340 on Build GRCh38
rs941622654 Chr.1: 225999397-225999398 on Build GRCh38
rs758182714 Chr.1: 225999360-225999360 on Build GRCh38
rs770085902 Chr.1: 225999189-225999189 on Build GRCh38
rs150202613 Chr.1: 225999275-225999275 on Build GRCh38
rs777350842 Chr.1: 225999281-225999281 on Build GRCh38
rs781225166 Chr.1: 225999187-225999187 on Build GRCh38
rs531284199 Chr.1: 225999319-225999319 on Build GRCh38
rs370941727 Chr.1: 225999242-225999242 on Build GRCh38
rs367925084 Chr.1: 225999286-225999286 on Build GRCh38
rs377317026 Chr.1: 225999336-225999336 on Build GRCh38
rs905861095 Chr.1: 225999534-225999534 on Build GRCh38
rs750557774 Chr.1: 225999307-225999307 on Build GRCh38
rs373033801 Chr.1: 225999359-225999359 on Build GRCh38
rs1001620974 Chr.1: 225999419-225999419 on Build GRCh38
rs897210734 Chr.1: 225999303-225999303 on Build GRCh38
rs201347746 Chr.1: 225999277-225999277 on Build GRCh38
rs370780477 Chr.1: 225999149-225999149 on Build GRCh38
rs751711575 Chr.1: 225999185-225999185 on Build GRCh38
rs990959842 Chr.1: 225999478-225999478 on Build GRCh38
rs776972535 Chr.1: 225999160-225999160 on Build GRCh38
rs944224411 Chr.1: 225999344-225999344 on Build GRCh38
rs771443315 Chr.1: 225999212-225999212 on Build GRCh38
rs754693992 Chr.1: 225999272-225999272 on Build GRCh38
rs911651345 Chr.1: 225999468-225999468 on Build GRCh38
rs916724797 Chr.1: 225999479-225999479 on Build GRCh38
rs374605550 Chr.1: 225999207-225999207 on Build GRCh38
rs754709441 Chr.1: 225999235-225999235 on Build GRCh38
rs1010053969 Chr.1: 225999441-225999441 on Build GRCh38
rs548805085 Chr.1: 225999225-225999225 on Build GRCh38
rs758051236 Chr.1: 225999320-225999320 on Build GRCh38
rs1020088932 Chr.1: 225999446-225999446 on Build GRCh38
rs763871990 Chr.1: 225999363-225999363 on Build GRCh38
rs778514010 Chr.1: 225999318-225999318 on Build GRCh38
rs1010294542 Chr.1: 225999558-225999558 on Build GRCh38
rs771623946 Chr.1: 225999279-225999279 on Build GRCh38
rs891180409 Chr.1: 225999566-225999566 on Build GRCh38
rs776110951 Chr.1: 225999291-225999291 on Build GRCh38
rs747729982 Chr.1: 225999210-225999210 on Build GRCh38
rs895876603 Chr.1: 225999552-225999552 on Build GRCh38
rs558819303 Chr.1: 225999451-225999451 on Build GRCh38
rs970145308 Chr.1: 225999270-225999270 on Build GRCh38
rs968581632 Chr.1: 225999447-225999447 on Build GRCh38
rs767440989 Chr.1: 225999255-225999255 on Build GRCh38
rs761877141 Chr.1: 225999299-225999299 on Build GRCh38
rs563576866 Chr.1: 225999266-225999266 on Build GRCh38
rs541090958 Chr.1: 225999460-225999460 on Build GRCh38
rs931521353 Chr.1: 225999517-225999517 on Build GRCh38
rs559169366 Chr.1: 225999458-225999458 on Build GRCh38
rs766957978 Chr.1: 225999154-225999154 on Build GRCh38
rs761767521 Chr.1: 225999435-225999435 on Build GRCh38
rs769073833 Chr.1: 225999343-225999343 on Build GRCh38
rs749704959 Chr.1: 225999338-225999338 on Build GRCh38
rs905868905 Chr.1: 225999414-225999414 on Build GRCh38
rs542012426 Chr.1: 225999265-225999265 on Build GRCh38
rs959847653 Chr.1: 225999438-225999438 on Build GRCh38
rs530425290 Chr.1: 225999475-225999475 on Build GRCh38
rs751209317 Chr.1: 225999364-225999364 on Build GRCh38
rs759324455 Chr.1: 225999355-225999355 on Build GRCh38
rs568033343 Chr.1: 225999349-225999349 on Build GRCh38
rs972570867 Chr.1: 225999153-225999153 on Build GRCh38
rs768409794 Chr.1: 225999208-225999208 on Build GRCh38
rs916817411 Chr.1: 225999390-225999390 on Build GRCh38
rs1057205699 Chr.1: 225999549-225999549 on Build GRCh38
rs931711304 Chr.1: 225999311-225999311 on Build GRCh38
rs556829616 Chr.1: 225999387-225999387 on Build GRCh38
rs773389325 Chr.1: 225999350-225999350 on Build GRCh38
rs542583963 Chr.1: 225999585-225999585 on Build GRCh38
rs372672932 Chr.1: 225999327-225999327 on Build GRCh38
rs904372732 Chr.1: 225999579-225999579 on Build GRCh38
rs1051093077 Chr.1: 225999433-225999433 on Build GRCh38
rs1049158532 Chr.1: 225999527-225999527 on Build GRCh38
rs1006684706 Chr.1: 225999326-225999326 on Build GRCh38
rs918762275 Chr.1: 225999483-225999483 on Build GRCh38
rs774650120 Chr.1: 225999294-225999294 on Build GRCh38
rs777216716 Chr.1: 225999221-225999221 on Build GRCh38
rs760010637 Chr.1: 225999164-225999164 on Build GRCh38
rs187014150 Chr.1: 225999461-225999461 on Build GRCh38
rs73100512 Chr.1: 225999428-225999428 on Build GRCh38
rs372463604 Chr.1: 225999180-225999180 on Build GRCh38
rs967027826 Chr.1: 225999469-225999469 on Build GRCh38
rs1049897246 Chr.1: 225999195-225999195 on Build GRCh38
rs376444841 Chr.1: 225999308-225999308 on Build GRCh38
rs201120938 Chr.1: 225999241-225999241 on Build GRCh38
rs972874164 Chr.1: 225999472-225999472 on Build GRCh38
rs369402843 Chr.1: 225999203-225999203 on Build GRCh38
rs375745106 Chr.1: 225999181-225999181 on Build GRCh38
rs750368230 Chr.1: 225999256-225999256 on Build GRCh38
rs1020034980 Chr.1: 225999567-225999567 on Build GRCh38
rs200698120 Chr.1: 225999246-225999246 on Build GRCh38
rs941575033 Chr.1: 225999506-225999506 on Build GRCh38
rs779760865 Chr.1: 225999205-225999205 on Build GRCh38
rs202232659 Chr.1: 225999352-225999352 on Build GRCh38
rs974720962 Chr.1: 225999513-225999513 on Build GRCh38
rs957025326 Chr.1: 225999201-225999201 on Build GRCh38
rs968343439 Chr.1: 225999287-225999287 on Build GRCh38
rs944172576 Chr.1: 225999470-225999470 on Build GRCh38
rs575145395 Chr.1: 225999395-225999395 on Build GRCh38
rs199904241 Chr.1: 225999292-225999292 on Build GRCh38
rs763389130 Chr.1: 225999237-225999237 on Build GRCh38
rs774648532 Chr.1: 225999345-225999345 on Build GRCh38
rs757254289 Chr.1: 225999186-225999186 on Build GRCh38
rs768095541 Chr.1: 225999389-225999389 on Build GRCh38
rs746511692 Chr.1: 225999282-225999282 on Build GRCh38
rs921611345 Chr.1: 225999296-225999296 on Build GRCh38
rs760534981 Chr.1: 225999535-225999535 on Build GRCh38

View Full Product Details