Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226992915-226993418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPA
Entrez Gene ID
8476
Gene Name
CDC42 binding protein kinase alpha
Gene Aliases
MRCK, MRCKA, PK428
UniGene
Hs.35433
Gene Chromosome Location
Chr.1: 226989865-227318492 on Build GRCh38
Transcripts
NM_003607.3 NM_014826.4 XM_005273317.3 XM_005273318.3 XM_005273320.3 XM_005273321.3 XM_005273322.3 XM_005273324.3 XM_006711834.3 XM_006711835.3 XM_011544306.2 XM_011544307.2 XM_011544308.2 XM_011544309.2 XM_017002578.1 XM_017002579.1 XM_017002580.1 XM_017002581.1

SNP Detail

SNP ID Chromosome Location
rs939919343 Chr.1: 226993028-226993028 on Build GRCh38
rs761777992 Chr.1: 226993097-226993097 on Build GRCh38
rs546719806 Chr.1: 226993270-226993270 on Build GRCh38
rs1011864384 Chr.1: 226993169-226993169 on Build GRCh38
rs35600383 Chr.1: 226993240-226993241 on Build GRCh38
rs559376238 Chr.1: 226993023-226993023 on Build GRCh38
rs772768240 Chr.1: 226993120-226993120 on Build GRCh38
rs1045921402 Chr.1: 226993271-226993271 on Build GRCh38
rs1021858688 Chr.1: 226993170-226993170 on Build GRCh38
rs563460045 Chr.1: 226993139-226993139 on Build GRCh38
rs1011046316 Chr.1: 226993364-226993364 on Build GRCh38
rs3795445 Chr.1: 226993246-226993246 on Build GRCh38
rs955368876 Chr.1: 226993084-226993084 on Build GRCh38
rs968157088 Chr.1: 226993174-226993174 on Build GRCh38
rs759969612 Chr.1: 226993363-226993363 on Build GRCh38
rs149839514 Chr.1: 226993346-226993346 on Build GRCh38
rs960405821 Chr.1: 226993295-226993295 on Build GRCh38
rs34133198 Chr.1: 226992950-226992951 on Build GRCh38
rs749561934 Chr.1: 226993252-226993252 on Build GRCh38
rs375271423 Chr.1: 226993199-226993199 on Build GRCh38
rs530866371 Chr.1: 226993153-226993153 on Build GRCh38
rs141866072 Chr.1: 226993009-226993009 on Build GRCh38
rs576809928 Chr.1: 226992941-226992941 on Build GRCh38
rs916050224 Chr.1: 226993177-226993177 on Build GRCh38
rs1038349665 Chr.1: 226993038-226993038 on Build GRCh38
rs941549297 Chr.1: 226993126-226993126 on Build GRCh38
rs139998015 Chr.1: 226993281-226993281 on Build GRCh38
rs991972587 Chr.1: 226993296-226993296 on Build GRCh38
rs926391138 Chr.1: 226992950-226992950 on Build GRCh38
rs1003201047 Chr.1: 226993282-226993282 on Build GRCh38
rs939222528 Chr.1: 226992956-226992956 on Build GRCh38
rs555115801 Chr.1: 226993304-226993304 on Build GRCh38
rs1801783 Chr.1: 226993157-226993157 on Build GRCh38
rs541568970 Chr.1: 226993050-226993050 on Build GRCh38
rs908470056 Chr.1: 226993003-226993003 on Build GRCh38
rs3795446 Chr.1: 226993280-226993280 on Build GRCh38
rs753957885 Chr.1: 226993310-226993311 on Build GRCh38
rs980952576 Chr.1: 226992945-226992945 on Build GRCh38
rs1038717375 Chr.1: 226993378-226993378 on Build GRCh38
rs570505729 Chr.1: 226993240-226993240 on Build GRCh38
rs113604509 Chr.1: 226993104-226993104 on Build GRCh38
rs368066339 Chr.1: 226993127-226993127 on Build GRCh38
rs532859401 Chr.1: 226993089-226993089 on Build GRCh38
rs975307005 Chr.1: 226993027-226993027 on Build GRCh38
rs919854009 Chr.1: 226993384-226993384 on Build GRCh38
rs1056192499 Chr.1: 226992976-226992976 on Build GRCh38

View Full Product Details