Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228101923-228102419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
UniGene
Hs.445952
Gene Chromosome Location
Chr.1: 228100727-228103462 on Build GRCh38
Transcripts
NM_024319.3 NR_130975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4837361 c.491C>A p.S164* Chr.1: 228102122-228102122 on Build GRCh38
3750935 c.471G>A p.G157G Chr.1: 228102142-228102142 on Build GRCh38
7358299 c.506C>T p.P169L Chr.1: 228102107-228102107 on Build GRCh38
5832498 c.478delA p.T160fs*>104 Chr.1: 228102135-228102135 on Build GRCh38
5443019 c.482C>T p.S161L Chr.1: 228102131-228102131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369194534 Chr.1: 228102324-228102324 on Build GRCh38
rs768806967 Chr.1: 228102058-228102058 on Build GRCh38
rs886251826 Chr.1: 228101981-228101981 on Build GRCh38
rs371322063 Chr.1: 228102069-228102069 on Build GRCh38
rs773858150 Chr.1: 228102281-228102281 on Build GRCh38
rs535500617 Chr.1: 228102070-228102070 on Build GRCh38
rs768292808 Chr.1: 228102370-228102370 on Build GRCh38
rs1037634609 Chr.1: 228102288-228102288 on Build GRCh38
rs1030007746 Chr.1: 228102305-228102305 on Build GRCh38
rs758352158 Chr.1: 228102145-228102145 on Build GRCh38
rs778552072 Chr.1: 228102375-228102375 on Build GRCh38
rs773046979 Chr.1: 228102067-228102067 on Build GRCh38
rs777713372 Chr.1: 228102261-228102261 on Build GRCh38
rs1128456 Chr.1: 228102142-228102142 on Build GRCh38
rs777130616 Chr.1: 228102290-228102290 on Build GRCh38
rs750895668 Chr.1: 228101983-228101983 on Build GRCh38
rs766729361 Chr.1: 228102120-228102120 on Build GRCh38
rs781000351 Chr.1: 228102339-228102339 on Build GRCh38
rs780318186 Chr.1: 228102051-228102051 on Build GRCh38
rs763126994 Chr.1: 228102321-228102321 on Build GRCh38
rs1037499470 Chr.1: 228102133-228102133 on Build GRCh38
rs933203759 Chr.1: 228101969-228101969 on Build GRCh38
rs768239498 Chr.1: 228102279-228102279 on Build GRCh38
rs1011133799 Chr.1: 228102198-228102198 on Build GRCh38
rs373634959 Chr.1: 228102171-228102171 on Build GRCh38
rs1012005115 Chr.1: 228102022-228102022 on Build GRCh38
rs757053740 Chr.1: 228102265-228102265 on Build GRCh38
rs553048285 Chr.1: 228101993-228101993 on Build GRCh38
rs942727658 Chr.1: 228102354-228102354 on Build GRCh38
rs765301504 Chr.1: 228102204-228102204 on Build GRCh38
rs61825155 Chr.1: 228102332-228102332 on Build GRCh38
rs558352129 Chr.1: 228102192-228102192 on Build GRCh38
rs754071074 Chr.1: 228102124-228102124 on Build GRCh38
rs750207588 Chr.1: 228102344-228102344 on Build GRCh38
rs754125957 Chr.1: 228102199-228102199 on Build GRCh38
rs760263336 Chr.1: 228102029-228102029 on Build GRCh38
rs753286442 Chr.1: 228102199-228102200 on Build GRCh38
rs764026267 Chr.1: 228102182-228102182 on Build GRCh38
rs764768698 Chr.1: 228102087-228102087 on Build GRCh38
rs769205183 Chr.1: 228102167-228102167 on Build GRCh38
rs780744622 Chr.1: 228102047-228102047 on Build GRCh38
rs955546434 Chr.1: 228102258-228102258 on Build GRCh38
rs756450716 Chr.1: 228102117-228102117 on Build GRCh38
rs202176716 Chr.1: 228102160-228102160 on Build GRCh38
rs769453961 Chr.1: 228102269-228102269 on Build GRCh38
rs901636058 Chr.1: 228102002-228102002 on Build GRCh38
rs776298667 Chr.1: 228102161-228102161 on Build GRCh38
rs767831891 Chr.1: 228102107-228102107 on Build GRCh38
rs539733055 Chr.1: 228101973-228101973 on Build GRCh38
rs540925037 Chr.1: 228102232-228102232 on Build GRCh38
rs778747113 Chr.1: 228102059-228102059 on Build GRCh38
rs755813107 Chr.1: 228102349-228102349 on Build GRCh38
rs773802973 Chr.1: 228102172-228102172 on Build GRCh38
rs975448763 Chr.1: 228102233-228102233 on Build GRCh38
rs922580740 Chr.1: 228102112-228102112 on Build GRCh38
rs757597518 Chr.1: 228102044-228102044 on Build GRCh38
rs777586746 Chr.1: 228102148-228102148 on Build GRCh38
rs746786530 Chr.1: 228102153-228102153 on Build GRCh38
rs759071624 Chr.1: 228102085-228102085 on Build GRCh38
rs956940702 Chr.1: 228102191-228102191 on Build GRCh38
rs753343443 Chr.1: 228102036-228102036 on Build GRCh38
rs1016013739 Chr.1: 228101994-228101994 on Build GRCh38
rs947909071 Chr.1: 228102301-228102301 on Build GRCh38
rs578158660 Chr.1: 228102223-228102223 on Build GRCh38
rs758317136 Chr.1: 228102215-228102215 on Build GRCh38
rs752058700 Chr.1: 228102039-228102039 on Build GRCh38
rs982769714 Chr.1: 228102262-228102262 on Build GRCh38
rs1039400723 Chr.1: 228102218-228102218 on Build GRCh38
rs192414159 Chr.1: 228101996-228101996 on Build GRCh38
rs771534451 Chr.1: 228102398-228102398 on Build GRCh38
rs779475958 Chr.1: 228102271-228102271 on Build GRCh38
rs187987054 Chr.1: 228101949-228101949 on Build GRCh38
rs528084635 Chr.1: 228102295-228102295 on Build GRCh38
rs746911412 Chr.1: 228102264-228102264 on Build GRCh38
rs145743393 Chr.1: 228102350-228102350 on Build GRCh38
rs746733325 Chr.1: 228102068-228102068 on Build GRCh38
rs367573467 Chr.1: 228102055-228102055 on Build GRCh38
rs886390091 Chr.1: 228102121-228102121 on Build GRCh38
rs752792503 Chr.1: 228102207-228102207 on Build GRCh38
rs758155115 Chr.1: 228102285-228102292 on Build GRCh38
rs1039642715 Chr.1: 228101958-228101958 on Build GRCh38
rs756850969 Chr.1: 228102204-228102205 on Build GRCh38
rs750728575 Chr.1: 228102046-228102046 on Build GRCh38
rs934029212 Chr.1: 228102115-228102115 on Build GRCh38
rs755215152 Chr.1: 228102203-228102203 on Build GRCh38
rs372383894 Chr.1: 228102181-228102181 on Build GRCh38
rs778992920 Chr.1: 228102131-228102131 on Build GRCh38
rs901749116 Chr.1: 228102219-228102219 on Build GRCh38
rs762065505 Chr.1: 228102270-228102271 on Build GRCh38
rs755017803 Chr.1: 228102125-228102125 on Build GRCh38
rs1048856897 Chr.1: 228101961-228101961 on Build GRCh38
rs764490059 Chr.1: 228102037-228102037 on Build GRCh38
rs745461545 Chr.1: 228102164-228102164 on Build GRCh38
rs979113669 Chr.1: 228102043-228102043 on Build GRCh38
rs1007858444 Chr.1: 228101985-228101985 on Build GRCh38
rs750783640 Chr.1: 228102116-228102116 on Build GRCh38
rs747586817 Chr.1: 228102393-228102393 on Build GRCh38
rs780516497 Chr.1: 228102317-228102320 on Build GRCh38
rs920409852 Chr.1: 228102292-228102292 on Build GRCh38
rs1011907889 Chr.1: 228102272-228102272 on Build GRCh38
rs762364530 Chr.1: 228102092-228102092 on Build GRCh38
rs902280027 Chr.1: 228102248-228102248 on Build GRCh38
rs774728235 Chr.1: 228102088-228102088 on Build GRCh38
rs368709965 Chr.1: 228102169-228102169 on Build GRCh38
rs771778489 Chr.1: 228102066-228102066 on Build GRCh38
rs906248662 Chr.1: 228102328-228102328 on Build GRCh38
rs922747896 Chr.1: 228102255-228102255 on Build GRCh38
rs2295995 Chr.1: 228102275-228102275 on Build GRCh38
rs901202011 Chr.1: 228101960-228101960 on Build GRCh38
rs762263318 Chr.1: 228102170-228102170 on Build GRCh38
rs752814685 Chr.1: 228102304-228102304 on Build GRCh38
rs997346458 Chr.1: 228102021-228102021 on Build GRCh38
rs570628641 Chr.1: 228101972-228101972 on Build GRCh38
rs970449522 Chr.1: 228102038-228102038 on Build GRCh38
rs915415252 Chr.1: 228102064-228102064 on Build GRCh38
rs765931801 Chr.1: 228102030-228102030 on Build GRCh38
rs748051165 Chr.1: 228102060-228102060 on Build GRCh38
rs780910425 Chr.1: 228102266-228102266 on Build GRCh38
rs765476215 Chr.1: 228102298-228102298 on Build GRCh38
rs761151810 Chr.1: 228102176-228102176 on Build GRCh38
rs761205135 Chr.1: 228102282-228102282 on Build GRCh38
rs771479442 Chr.1: 228102286-228102286 on Build GRCh38
rs899673663 Chr.1: 228102213-228102213 on Build GRCh38
rs373950653 Chr.1: 228102353-228102353 on Build GRCh38
rs764775944 Chr.1: 228102260-228102261 on Build GRCh38
rs373337967 Chr.1: 228102105-228102105 on Build GRCh38

View Full Product Details