Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235167541-235168069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID4B
Entrez Gene ID
51742
Gene Name
AT-rich interaction domain 4B
Gene Aliases
BCAA, BRCAA1, RBBP1L1, RBP1L1, SAP180
UniGene
Hs.575782
Gene Chromosome Location
Chr.1: 235166895-235328240 on Build GRCh38
Transcripts
NM_001206794.1 NM_016374.5 NM_031371.3 NR_135074.1 XM_006711781.2 XM_011544212.2 XM_017001468.1 XM_017001469.1 XM_017001470.1 XM_017001472.1

SNP Detail

SNP ID Chromosome Location
rs1004283667 Chr.1: 235167721-235167724 on Build GRCh38
rs1021103927 Chr.1: 235167892-235167892 on Build GRCh38
rs79459839 Chr.1: 235167891-235167891 on Build GRCh38
rs920544203 Chr.1: 235167972-235167972 on Build GRCh38
rs529694375 Chr.1: 235167765-235167765 on Build GRCh38
rs991457205 Chr.1: 235167753-235167753 on Build GRCh38
rs942996358 Chr.1: 235167954-235167954 on Build GRCh38
rs987209381 Chr.1: 235167912-235167912 on Build GRCh38
rs933511315 Chr.1: 235167935-235167935 on Build GRCh38
rs990090432 Chr.1: 235167874-235167874 on Build GRCh38
rs753913267 Chr.1: 235167764-235167767 on Build GRCh38
rs899004771 Chr.1: 235167568-235167568 on Build GRCh38
rs1002464795 Chr.1: 235167826-235167827 on Build GRCh38
rs748009838 Chr.1: 235167672-235167672 on Build GRCh38
rs1037056906 Chr.1: 235167907-235167907 on Build GRCh38
rs942256096 Chr.1: 235167827-235167827 on Build GRCh38
rs974375206 Chr.1: 235167964-235167964 on Build GRCh38
rs887220926 Chr.1: 235167716-235167716 on Build GRCh38
rs543747173 Chr.1: 235167659-235167659 on Build GRCh38
rs769735310 Chr.1: 235167930-235167930 on Build GRCh38
rs776787285 Chr.1: 235167615-235167615 on Build GRCh38
rs1047547569 Chr.1: 235167682-235167682 on Build GRCh38
rs187938326 Chr.1: 235167974-235167974 on Build GRCh38
rs928728010 Chr.1: 235167628-235167628 on Build GRCh38
rs1007272072 Chr.1: 235168005-235168005 on Build GRCh38
rs566578366 Chr.1: 235167793-235167793 on Build GRCh38
rs562041179 Chr.1: 235167700-235167700 on Build GRCh38
rs913732077 Chr.1: 235167592-235167592 on Build GRCh38
rs572260519 Chr.1: 235167827-235167827 on Build GRCh38
rs1025416609 Chr.1: 235167816-235167816 on Build GRCh38
rs909219146 Chr.1: 235167820-235167820 on Build GRCh38
rs762472902 Chr.1: 235168021-235168031 on Build GRCh38
rs1048047306 Chr.1: 235168032-235168032 on Build GRCh38
rs755532474 Chr.1: 235167938-235167938 on Build GRCh38
rs981511918 Chr.1: 235167624-235167624 on Build GRCh38
rs751052632 Chr.1: 235167903-235167904 on Build GRCh38
rs966879746 Chr.1: 235167896-235167896 on Build GRCh38
rs1051902920 Chr.1: 235167973-235167973 on Build GRCh38
rs867263426 Chr.1: 235167872-235167872 on Build GRCh38
rs527370167 Chr.1: 235167817-235167817 on Build GRCh38
rs1031930313 Chr.1: 235168010-235168010 on Build GRCh38
rs547833359 Chr.1: 235167781-235167781 on Build GRCh38
rs937424104 Chr.1: 235167667-235167667 on Build GRCh38

View Full Product Details