Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235343655-235344151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGPS1
Entrez Gene ID
9453
Gene Name
geranylgeranyl diphosphate synthase 1
Gene Aliases
GGPPS, GGPPS1
UniGene
Hs.730768
Gene Chromosome Location
Chr.1: 235328438-235344532 on Build GRCh38
Transcripts
NM_001037277.1 NR_036605.1

SNP Detail

SNP ID Chromosome Location
rs147721869 Chr.1: 235343775-235343775 on Build GRCh38
rs894521326 Chr.1: 235343928-235343928 on Build GRCh38
rs957461658 Chr.1: 235343704-235343704 on Build GRCh38
rs766495567 Chr.1: 235343976-235343976 on Build GRCh38
rs1013614052 Chr.1: 235343765-235343765 on Build GRCh38
rs955184255 Chr.1: 235343910-235343910 on Build GRCh38
rs191550055 Chr.1: 235343816-235343816 on Build GRCh38
rs954422969 Chr.1: 235343805-235343805 on Build GRCh38
rs1054150815 Chr.1: 235343834-235343834 on Build GRCh38
rs987012904 Chr.1: 235344069-235344069 on Build GRCh38
rs1032974084 Chr.1: 235344030-235344030 on Build GRCh38
rs915121118 Chr.1: 235343849-235343849 on Build GRCh38
rs550550968 Chr.1: 235343785-235343785 on Build GRCh38
rs182829142 Chr.1: 235343713-235343713 on Build GRCh38
rs555522692 Chr.1: 235343861-235343861 on Build GRCh38
rs1009399278 Chr.1: 235343696-235343696 on Build GRCh38
rs1016515789 Chr.1: 235343773-235343773 on Build GRCh38
rs76362749 Chr.1: 235343728-235343728 on Build GRCh38
rs896270725 Chr.1: 235343931-235343931 on Build GRCh38
rs1057129359 Chr.1: 235343927-235343927 on Build GRCh38
rs910249781 Chr.1: 235343818-235343818 on Build GRCh38
rs987255562 Chr.1: 235344047-235344047 on Build GRCh38
rs943040663 Chr.1: 235343820-235343820 on Build GRCh38
rs186747001 Chr.1: 235343732-235343732 on Build GRCh38
rs750781673 Chr.1: 235343900-235343900 on Build GRCh38
rs988898042 Chr.1: 235343860-235343860 on Build GRCh38
rs1010275842 Chr.1: 235344032-235344032 on Build GRCh38
rs906081481 Chr.1: 235343997-235343997 on Build GRCh38
rs993889117 Chr.1: 235343769-235343769 on Build GRCh38
rs184808976 Chr.1: 235343912-235343912 on Build GRCh38
rs1014461445 Chr.1: 235343776-235343776 on Build GRCh38
rs533260065 Chr.1: 235343701-235343701 on Build GRCh38
rs919598907 Chr.1: 235343929-235343929 on Build GRCh38
rs565164591 Chr.1: 235343774-235343774 on Build GRCh38
rs748742720 Chr.1: 235343818-235343820 on Build GRCh38
rs942650095 Chr.1: 235343926-235343926 on Build GRCh38
rs750007958 Chr.1: 235343749-235343749 on Build GRCh38
rs754014821 Chr.1: 235344093-235344093 on Build GRCh38
rs577395024 Chr.1: 235344046-235344046 on Build GRCh38
rs1018119098 Chr.1: 235344079-235344079 on Build GRCh38
rs945208916 Chr.1: 235343892-235343892 on Build GRCh38
rs78846778 Chr.1: 235343979-235343979 on Build GRCh38
rs1042194196 Chr.1: 235343899-235343899 on Build GRCh38
rs774939059 Chr.1: 235343706-235343706 on Build GRCh38
rs942347438 Chr.1: 235343740-235343740 on Build GRCh38
rs761005903 Chr.1: 235343930-235343930 on Build GRCh38
rs986678992 Chr.1: 235343911-235343911 on Build GRCh38
rs16832568 Chr.1: 235343798-235343798 on Build GRCh38
rs994093235 Chr.1: 235344012-235344012 on Build GRCh38
rs913078652 Chr.1: 235343724-235343724 on Build GRCh38
rs113744670 Chr.1: 235343913-235343913 on Build GRCh38
rs534372014 Chr.1: 235343926-235343926 on Build GRCh38
rs1033028441 Chr.1: 235343825-235343825 on Build GRCh38
rs964267927 Chr.1: 235344127-235344127 on Build GRCh38
rs148006750 Chr.1: 235344069-235344069 on Build GRCh38
rs1012881676 Chr.1: 235343943-235343943 on Build GRCh38
rs1024966038 Chr.1: 235343786-235343786 on Build GRCh38
rs899062644 Chr.1: 235343914-235343925 on Build GRCh38
rs1045918870 Chr.1: 235343981-235343981 on Build GRCh38
rs59003657 Chr.1: 235343688-235343688 on Build GRCh38
rs371654078 Chr.1: 235344031-235344031 on Build GRCh38
rs767949430 Chr.1: 235343847-235343847 on Build GRCh38

View Full Product Details