Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235344014-235344559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGPS1
Entrez Gene ID
9453
Gene Name
geranylgeranyl diphosphate synthase 1
Gene Aliases
GGPPS, GGPPS1
UniGene
Hs.730768
Gene Chromosome Location
Chr.1: 235328438-235344532 on Build GRCh38
Transcripts
NM_001037277.1 NR_036605.1

SNP Detail

SNP ID Chromosome Location
rs565556878 Chr.1: 235344150-235344151 on Build GRCh38
rs890665351 Chr.1: 235344420-235344420 on Build GRCh38
rs568833751 Chr.1: 235344310-235344310 on Build GRCh38
rs532916278 Chr.1: 235344502-235344506 on Build GRCh38
rs190706591 Chr.1: 235344365-235344365 on Build GRCh38
rs893420663 Chr.1: 235344417-235344417 on Build GRCh38
rs989569367 Chr.1: 235344475-235344475 on Build GRCh38
rs1057148603 Chr.1: 235344311-235344311 on Build GRCh38
rs987012904 Chr.1: 235344069-235344069 on Build GRCh38
rs141430544 Chr.1: 235344503-235344503 on Build GRCh38
rs1026562509 Chr.1: 235344385-235344385 on Build GRCh38
rs754014821 Chr.1: 235344093-235344093 on Build GRCh38
rs1037686901 Chr.1: 235344337-235344337 on Build GRCh38
rs879253406 Chr.1: 235344152-235344152 on Build GRCh38
rs577395024 Chr.1: 235344046-235344046 on Build GRCh38
rs1020314720 Chr.1: 235344441-235344441 on Build GRCh38
rs1018119098 Chr.1: 235344079-235344079 on Build GRCh38
rs977960675 Chr.1: 235344241-235344241 on Build GRCh38
rs774773442 Chr.1: 235344339-235344339 on Build GRCh38
rs1009028239 Chr.1: 235344425-235344425 on Build GRCh38
rs77179024 Chr.1: 235344168-235344170 on Build GRCh38
rs57621540 Chr.1: 235344152-235344152 on Build GRCh38
rs939179699 Chr.1: 235344307-235344307 on Build GRCh38
rs894531358 Chr.1: 235344315-235344315 on Build GRCh38
rs1054706561 Chr.1: 235344416-235344416 on Build GRCh38
rs1018482865 Chr.1: 235344494-235344494 on Build GRCh38
rs974017700 Chr.1: 235344170-235344170 on Build GRCh38
rs535982364 Chr.1: 235344342-235344342 on Build GRCh38
rs868863485 Chr.1: 235344181-235344181 on Build GRCh38
rs1017372085 Chr.1: 235344456-235344456 on Build GRCh38
rs915154318 Chr.1: 235344175-235344175 on Build GRCh38
rs139019369 Chr.1: 235344458-235344458 on Build GRCh38
rs987255562 Chr.1: 235344047-235344047 on Build GRCh38
rs1026843843 Chr.1: 235344524-235344524 on Build GRCh38
rs557001525 Chr.1: 235344298-235344298 on Build GRCh38
rs964267927 Chr.1: 235344127-235344127 on Build GRCh38
rs148006750 Chr.1: 235344069-235344069 on Build GRCh38
rs989124660 Chr.1: 235344171-235344171 on Build GRCh38
rs779624909 Chr.1: 235344277-235344277 on Build GRCh38
rs781330540 Chr.1: 235344151-235344152 on Build GRCh38
rs945289664 Chr.1: 235344178-235344178 on Build GRCh38
rs1008319154 Chr.1: 235344475-235344476 on Build GRCh38
rs899145452 Chr.1: 235344353-235344353 on Build GRCh38
rs572981753 Chr.1: 235344446-235344446 on Build GRCh38
rs948744529 Chr.1: 235344332-235344332 on Build GRCh38

View Full Product Details