Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235548405-235548914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG4
Entrez Gene ID
2786
Gene Name
G protein subunit gamma 4
Gene Aliases
-
UniGene
Hs.159711
Gene Chromosome Location
Chr.1: 235547685-235650754 on Build GRCh38
Transcripts
NM_001098721.1 NM_001098722.1 NM_004485.3 XM_006711761.2 XM_011544167.2

SNP Detail

SNP ID Chromosome Location
rs142486030 Chr.1: 235548547-235548547 on Build GRCh38
rs190268895 Chr.1: 235548823-235548823 on Build GRCh38
rs377543389 Chr.1: 235548599-235548599 on Build GRCh38
rs972240988 Chr.1: 235548433-235548433 on Build GRCh38
rs1016678042 Chr.1: 235548673-235548673 on Build GRCh38
rs919505544 Chr.1: 235548438-235548438 on Build GRCh38
rs185463345 Chr.1: 235548540-235548540 on Build GRCh38
rs911617914 Chr.1: 235548603-235548603 on Build GRCh38
rs910564586 Chr.1: 235548479-235548479 on Build GRCh38
rs1046444680 Chr.1: 235548444-235548444 on Build GRCh38
rs975571251 Chr.1: 235548624-235548624 on Build GRCh38
rs1055000882 Chr.1: 235548783-235548783 on Build GRCh38
rs965546270 Chr.1: 235548615-235548615 on Build GRCh38
rs966381462 Chr.1: 235548840-235548840 on Build GRCh38
rs1005660724 Chr.1: 235548666-235548666 on Build GRCh38
rs574160566 Chr.1: 235548694-235548694 on Build GRCh38
rs769226238 Chr.1: 235548526-235548526 on Build GRCh38
rs7524940 Chr.1: 235548480-235548480 on Build GRCh38
rs576149683 Chr.1: 235548569-235548569 on Build GRCh38
rs945183592 Chr.1: 235548563-235548563 on Build GRCh38
rs764015280 Chr.1: 235548525-235548525 on Build GRCh38
rs1008535956 Chr.1: 235548842-235548842 on Build GRCh38
rs398089969 Chr.1: 235548635-235548636 on Build GRCh38
rs1046477991 Chr.1: 235548700-235548700 on Build GRCh38
rs370440027 Chr.1: 235548635-235548635 on Build GRCh38
rs182770704 Chr.1: 235548830-235548830 on Build GRCh38
rs908571236 Chr.1: 235548667-235548667 on Build GRCh38
rs181043113 Chr.1: 235548443-235548443 on Build GRCh38
rs1011232886 Chr.1: 235548536-235548536 on Build GRCh38
rs964728247 Chr.1: 235548850-235548850 on Build GRCh38
rs545634706 Chr.1: 235548851-235548851 on Build GRCh38
rs555869552 Chr.1: 235548651-235548651 on Build GRCh38
rs1002449515 Chr.1: 235548736-235548736 on Build GRCh38
rs1026446188 Chr.1: 235548819-235548819 on Build GRCh38
rs987396943 Chr.1: 235548472-235548472 on Build GRCh38
rs78095380 Chr.1: 235548721-235548721 on Build GRCh38
rs906392826 Chr.1: 235548475-235548475 on Build GRCh38
rs966349377 Chr.1: 235548579-235548579 on Build GRCh38
rs368097413 Chr.1: 235548632-235548632 on Build GRCh38
rs1018710917 Chr.1: 235548845-235548845 on Build GRCh38
rs933574238 Chr.1: 235548442-235548442 on Build GRCh38
rs541573786 Chr.1: 235548789-235548789 on Build GRCh38
rs993584911 Chr.1: 235548796-235548796 on Build GRCh38
rs906592332 Chr.1: 235548722-235548722 on Build GRCh38
rs931613378 Chr.1: 235548642-235548642 on Build GRCh38
rs906415058 Chr.1: 235548588-235548588 on Build GRCh38
rs910817335 Chr.1: 235548838-235548838 on Build GRCh38
rs10545643 Chr.1: 235548631-235548632 on Build GRCh38
rs1054773102 Chr.1: 235548601-235548601 on Build GRCh38

View Full Product Details