Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235548844-235549386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG4
Entrez Gene ID
2786
Gene Name
G protein subunit gamma 4
Gene Aliases
-
UniGene
Hs.159711
Gene Chromosome Location
Chr.1: 235547685-235650754 on Build GRCh38
Transcripts
NM_001098721.1 NM_001098722.1 NM_004485.3 XM_006711761.2 XM_011544167.2

SNP Detail

SNP ID Chromosome Location
rs748518813 Chr.1: 235548918-235548918 on Build GRCh38
rs10802805 Chr.1: 235549326-235549326 on Build GRCh38
rs766508375 Chr.1: 235548994-235548994 on Build GRCh38
rs964498835 Chr.1: 235548932-235548932 on Build GRCh38
rs536115259 Chr.1: 235549011-235549011 on Build GRCh38
rs577928217 Chr.1: 235549149-235549149 on Build GRCh38
rs759579371 Chr.1: 235549066-235549066 on Build GRCh38
rs192183671 Chr.1: 235549082-235549082 on Build GRCh38
rs539073741 Chr.1: 235549039-235549039 on Build GRCh38
rs553505919 Chr.1: 235549107-235549107 on Build GRCh38
rs141554657 Chr.1: 235549050-235549050 on Build GRCh38
rs561135050 Chr.1: 235549264-235549264 on Build GRCh38
rs938138238 Chr.1: 235549048-235549048 on Build GRCh38
rs567509303 Chr.1: 235549090-235549090 on Build GRCh38
rs763828058 Chr.1: 235549172-235549172 on Build GRCh38
rs370465786 Chr.1: 235549261-235549261 on Build GRCh38
rs946515482 Chr.1: 235549086-235549086 on Build GRCh38
rs565669015 Chr.1: 235549191-235549192 on Build GRCh38
rs993889290 Chr.1: 235549043-235549043 on Build GRCh38
rs1038444249 Chr.1: 235548999-235548999 on Build GRCh38
rs139415519 Chr.1: 235549171-235549171 on Build GRCh38
rs868456245 Chr.1: 235549000-235549000 on Build GRCh38
rs150891868 Chr.1: 235549067-235549067 on Build GRCh38
rs563836439 Chr.1: 235548903-235548903 on Build GRCh38
rs575824153 Chr.1: 235549227-235549227 on Build GRCh38
rs927800783 Chr.1: 235548940-235548940 on Build GRCh38
rs528666722 Chr.1: 235549313-235549313 on Build GRCh38
rs528938202 Chr.1: 235548976-235548976 on Build GRCh38
rs559040282 Chr.1: 235549340-235549340 on Build GRCh38
rs187702976 Chr.1: 235549061-235549061 on Build GRCh38
rs368224317 Chr.1: 235549116-235549116 on Build GRCh38
rs746940960 Chr.1: 235549031-235549031 on Build GRCh38
rs1046342643 Chr.1: 235548984-235548984 on Build GRCh38
rs939161335 Chr.1: 235549239-235549239 on Build GRCh38
rs940031835 Chr.1: 235548975-235548975 on Build GRCh38
rs1055039433 Chr.1: 235548964-235548964 on Build GRCh38
rs367635037 Chr.1: 235549215-235549216 on Build GRCh38
rs7525328 Chr.1: 235548935-235548935 on Build GRCh38
rs765158221 Chr.1: 235549085-235549085 on Build GRCh38
rs1009048782 Chr.1: 235549071-235549071 on Build GRCh38
rs1027795113 Chr.1: 235549216-235549218 on Build GRCh38
rs115099233 Chr.1: 235549260-235549260 on Build GRCh38
rs747668674 Chr.1: 235549362-235549364 on Build GRCh38
rs915325849 Chr.1: 235548920-235548920 on Build GRCh38
rs1015802502 Chr.1: 235549301-235549301 on Build GRCh38
rs752533180 Chr.1: 235549155-235549155 on Build GRCh38
rs10802804 Chr.1: 235548968-235548968 on Build GRCh38
rs965370712 Chr.1: 235549091-235549091 on Build GRCh38
rs895091895 Chr.1: 235548967-235548967 on Build GRCh38
rs557601304 Chr.1: 235549197-235549197 on Build GRCh38
rs929666471 Chr.1: 235549337-235549337 on Build GRCh38
rs565292608 Chr.1: 235549037-235549037 on Build GRCh38
rs776505517 Chr.1: 235549002-235549002 on Build GRCh38
rs896873811 Chr.1: 235549023-235549023 on Build GRCh38
rs751257390 Chr.1: 235549246-235549246 on Build GRCh38
rs1008819001 Chr.1: 235549109-235549109 on Build GRCh38
rs995965059 Chr.1: 235549175-235549175 on Build GRCh38
rs945451116 Chr.1: 235548921-235548921 on Build GRCh38
rs549331053 Chr.1: 235548963-235548963 on Build GRCh38
rs535118046 Chr.1: 235549096-235549096 on Build GRCh38
rs890909152 Chr.1: 235549343-235549343 on Build GRCh38
rs547128785 Chr.1: 235549021-235549021 on Build GRCh38

View Full Product Details