Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235551184-235551711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG4
Entrez Gene ID
2786
Gene Name
G protein subunit gamma 4
Gene Aliases
-
UniGene
Hs.159711
Gene Chromosome Location
Chr.1: 235547685-235650754 on Build GRCh38
Transcripts
NM_001098721.1 NM_001098722.1 NM_004485.3 XM_006711761.2 XM_011544167.2

SNP Detail

SNP ID Chromosome Location
rs549955337 Chr.1: 235551262-235551262 on Build GRCh38
rs921321840 Chr.1: 235551413-235551413 on Build GRCh38
rs934505960 Chr.1: 235551481-235551481 on Build GRCh38
rs925795198 Chr.1: 235551471-235551471 on Build GRCh38
rs973416685 Chr.1: 235551495-235551495 on Build GRCh38
rs1044322256 Chr.1: 235551544-235551544 on Build GRCh38
rs978823570 Chr.1: 235551612-235551612 on Build GRCh38
rs764543739 Chr.1: 235551281-235551281 on Build GRCh38
rs895465874 Chr.1: 235551310-235551310 on Build GRCh38
rs924322218 Chr.1: 235551222-235551222 on Build GRCh38
rs1002341000 Chr.1: 235551362-235551362 on Build GRCh38
rs777143806 Chr.1: 235551284-235551284 on Build GRCh38
rs994671410 Chr.1: 235551608-235551628 on Build GRCh38
rs950020780 Chr.1: 235551680-235551680 on Build GRCh38
rs942040884 Chr.1: 235551491-235551491 on Build GRCh38
rs966018083 Chr.1: 235551547-235551547 on Build GRCh38
rs568808419 Chr.1: 235551458-235551458 on Build GRCh38
rs746420415 Chr.1: 235551499-235551499 on Build GRCh38
rs540246541 Chr.1: 235551661-235551661 on Build GRCh38
rs926055583 Chr.1: 235551638-235551638 on Build GRCh38
rs965402309 Chr.1: 235551411-235551411 on Build GRCh38
rs560854880 Chr.1: 235551236-235551236 on Build GRCh38
rs10925979 Chr.1: 235551269-235551269 on Build GRCh38
rs895627666 Chr.1: 235551325-235551325 on Build GRCh38
rs759042678 Chr.1: 235551370-235551370 on Build GRCh38
rs917138271 Chr.1: 235551669-235551669 on Build GRCh38
rs1043285623 Chr.1: 235551289-235551289 on Build GRCh38
rs933615370 Chr.1: 235551237-235551237 on Build GRCh38
rs976747661 Chr.1: 235551559-235551559 on Build GRCh38
rs1027382935 Chr.1: 235551529-235551529 on Build GRCh38
rs1017772253 Chr.1: 235551585-235551585 on Build GRCh38
rs368607213 Chr.1: 235551570-235551570 on Build GRCh38
rs750528605 Chr.1: 235551475-235551475 on Build GRCh38
rs568255617 Chr.1: 235551265-235551265 on Build GRCh38
rs189513363 Chr.1: 235551537-235551537 on Build GRCh38
rs993542337 Chr.1: 235551291-235551291 on Build GRCh38
rs1057458446 Chr.1: 235551512-235551512 on Build GRCh38
rs982976152 Chr.1: 235551375-235551375 on Build GRCh38
rs978526153 Chr.1: 235551221-235551221 on Build GRCh38
rs904407616 Chr.1: 235551296-235551296 on Build GRCh38
rs115691204 Chr.1: 235551684-235551684 on Build GRCh38
rs895760026 Chr.1: 235551485-235551485 on Build GRCh38
rs977199118 Chr.1: 235551412-235551412 on Build GRCh38
rs550731407 Chr.1: 235551315-235551315 on Build GRCh38
rs373276264 Chr.1: 235551371-235551371 on Build GRCh38
rs538447812 Chr.1: 235551205-235551205 on Build GRCh38
rs185562540 Chr.1: 235551387-235551387 on Build GRCh38
rs891448528 Chr.1: 235551565-235551565 on Build GRCh38
rs556753140 Chr.1: 235551217-235551217 on Build GRCh38
rs11549298 Chr.1: 235551226-235551226 on Build GRCh38
rs997951856 Chr.1: 235551570-235551570 on Build GRCh38
rs1000105103 Chr.1: 235551557-235551562 on Build GRCh38
rs1044461409 Chr.1: 235551500-235551500 on Build GRCh38
rs556922122 Chr.1: 235551229-235551229 on Build GRCh38
rs999823082 Chr.1: 235551359-235551359 on Build GRCh38
rs923424829 Chr.1: 235551232-235551232 on Build GRCh38
rs12064698 Chr.1: 235551271-235551271 on Build GRCh38
rs527689338 Chr.1: 235551414-235551414 on Build GRCh38
rs910763198 Chr.1: 235551423-235551423 on Build GRCh38
rs968428011 Chr.1: 235551357-235551357 on Build GRCh38
rs973614687 Chr.1: 235551616-235551616 on Build GRCh38
rs752005878 Chr.1: 235551521-235551521 on Build GRCh38
rs386640603 Chr.1: 235551269-235551271 on Build GRCh38
rs1031369976 Chr.1: 235551557-235551557 on Build GRCh38
rs147039227 Chr.1: 235551394-235551394 on Build GRCh38
rs954906377 Chr.1: 235551571-235551573 on Build GRCh38
rs186002750 Chr.1: 235551626-235551626 on Build GRCh38
rs538184431 Chr.1: 235551504-235551504 on Build GRCh38
rs1052748069 Chr.1: 235551309-235551309 on Build GRCh38
rs919283778 Chr.1: 235551636-235551636 on Build GRCh38
rs189124403 Chr.1: 235551270-235551270 on Build GRCh38
rs562380450 Chr.1: 235551340-235551340 on Build GRCh38
rs988645848 Chr.1: 235551657-235551657 on Build GRCh38
rs370257117 Chr.1: 235551297-235551297 on Build GRCh38
rs556701436 Chr.1: 235551530-235551530 on Build GRCh38
rs952971335 Chr.1: 235551216-235551216 on Build GRCh38
rs986666818 Chr.1: 235551575-235551575 on Build GRCh38
rs111566975 Chr.1: 235551313-235551313 on Build GRCh38
rs60121178 Chr.1: 235551560-235551562 on Build GRCh38
rs934478242 Chr.1: 235551655-235551655 on Build GRCh38
rs1030521616 Chr.1: 235551380-235551380 on Build GRCh38
rs67745431 Chr.1: 235551271-235551271 on Build GRCh38
rs796614314 Chr.1: 235551567-235551569 on Build GRCh38
rs571144948 Chr.1: 235551482-235551482 on Build GRCh38
rs531036209 Chr.1: 235551227-235551227 on Build GRCh38
rs1015797916 Chr.1: 235551538-235551538 on Build GRCh38
rs497263 Chr.1: 235551562-235551562 on Build GRCh38
rs1012860452 Chr.1: 235551334-235551334 on Build GRCh38
rs181536228 Chr.1: 235551600-235551600 on Build GRCh38
rs970213253 Chr.1: 235551351-235551351 on Build GRCh38
rs920137274 Chr.1: 235551206-235551206 on Build GRCh38
rs374636331 Chr.1: 235551567-235551567 on Build GRCh38

View Full Product Details