Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 239632131-239632676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRM3
Entrez Gene ID
1131
Gene Name
cholinergic receptor muscarinic 3
Gene Aliases
EGBRS, HM3, PBS
UniGene
Hs.155736
Gene Chromosome Location
Chr.1: 239386565-239915450 on Build GRCh38
Transcripts
NM_000740.2 XM_005273032.3 XM_011544041.2 XM_011544043.2 XM_011544044.2 XM_011544045.2 XM_011544046.2 XM_011544047.2 XM_017000152.1 XM_017000153.1 XM_017000154.1 XM_017000155.1 XM_017000156.1 XM_017000157.1 XM_017000158.1 XM_017000159.1 XM_017000161.1 XM_017000162.1

SNP Detail

SNP ID Chromosome Location
rs771317867 Chr.1: 239632389-239632389 on Build GRCh38
rs911136223 Chr.1: 239632163-239632163 on Build GRCh38
rs201669945 Chr.1: 239632324-239632324 on Build GRCh38
rs972369912 Chr.1: 239632381-239632381 on Build GRCh38
rs913122347 Chr.1: 239632489-239632489 on Build GRCh38
rs968885112 Chr.1: 239632610-239632610 on Build GRCh38
rs531926732 Chr.1: 239632377-239632377 on Build GRCh38
rs201882207 Chr.1: 239632325-239632325 on Build GRCh38
rs41269367 Chr.1: 239632253-239632253 on Build GRCh38
rs895972662 Chr.1: 239632615-239632615 on Build GRCh38
rs1043133993 Chr.1: 239632285-239632285 on Build GRCh38
rs1010305957 Chr.1: 239632206-239632206 on Build GRCh38
rs866821645 Chr.1: 239632538-239632538 on Build GRCh38
rs200408139 Chr.1: 239632320-239632320 on Build GRCh38
rs1014404876 Chr.1: 239632619-239632619 on Build GRCh38
rs1024675520 Chr.1: 239632371-239632371 on Build GRCh38
rs755322072 Chr.1: 239632193-239632193 on Build GRCh38
rs891777376 Chr.1: 239632202-239632202 on Build GRCh38
rs143710557 Chr.1: 239632541-239632541 on Build GRCh38
rs912956708 Chr.1: 239632605-239632605 on Build GRCh38
rs145195938 Chr.1: 239632190-239632190 on Build GRCh38
rs536097291 Chr.1: 239632469-239632469 on Build GRCh38
rs1005530872 Chr.1: 239632442-239632442 on Build GRCh38
rs985729729 Chr.1: 239632395-239632395 on Build GRCh38
rs36078512 Chr.1: 239632363-239632364 on Build GRCh38
rs988569678 Chr.1: 239632204-239632204 on Build GRCh38
rs200102920 Chr.1: 239632212-239632212 on Build GRCh38
rs956860382 Chr.1: 239632568-239632569 on Build GRCh38
rs184291667 Chr.1: 239632558-239632558 on Build GRCh38
rs749301818 Chr.1: 239632292-239632292 on Build GRCh38
rs538040324 Chr.1: 239632352-239632352 on Build GRCh38
rs149157312 Chr.1: 239632353-239632353 on Build GRCh38
rs756177823 Chr.1: 239632342-239632342 on Build GRCh38
rs911053571 Chr.1: 239632401-239632401 on Build GRCh38
rs554051315 Chr.1: 239632470-239632470 on Build GRCh38
rs568580786 Chr.1: 239632369-239632369 on Build GRCh38
rs774706446 Chr.1: 239632419-239632419 on Build GRCh38
rs572100368 Chr.1: 239632498-239632498 on Build GRCh38
rs10925933 Chr.1: 239632349-239632349 on Build GRCh38
rs200918714 Chr.1: 239632245-239632245 on Build GRCh38
rs937440558 Chr.1: 239632600-239632600 on Build GRCh38
rs1034516610 Chr.1: 239632338-239632338 on Build GRCh38
rs768438156 Chr.1: 239632625-239632625 on Build GRCh38
rs1002111027 Chr.1: 239632333-239632333 on Build GRCh38
rs1056390986 Chr.1: 239632606-239632606 on Build GRCh38
rs952460184 Chr.1: 239632392-239632392 on Build GRCh38
rs969398531 Chr.1: 239632485-239632485 on Build GRCh38
rs904640682 Chr.1: 239632591-239632591 on Build GRCh38
rs911109560 Chr.1: 239632293-239632293 on Build GRCh38
rs187651581 Chr.1: 239632197-239632197 on Build GRCh38
rs202098800 Chr.1: 239632208-239632208 on Build GRCh38
rs532997435 Chr.1: 239632236-239632236 on Build GRCh38
rs988716676 Chr.1: 239632584-239632584 on Build GRCh38
rs200973680 Chr.1: 239632198-239632198 on Build GRCh38
rs976624738 Chr.1: 239632452-239632452 on Build GRCh38

View Full Product Details