Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 239826887-239827395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRM3
Entrez Gene ID
1131
Gene Name
cholinergic receptor muscarinic 3
Gene Aliases
EGBRS, HM3, PBS
UniGene
Hs.155736
Gene Chromosome Location
Chr.1: 239386565-239915450 on Build GRCh38
Transcripts
NM_000740.2 XM_005273032.3 XM_011544041.2 XM_011544043.2 XM_011544044.2 XM_011544045.2 XM_011544046.2 XM_011544047.2 XM_017000152.1 XM_017000153.1 XM_017000154.1 XM_017000155.1 XM_017000156.1 XM_017000157.1 XM_017000158.1 XM_017000159.1 XM_017000160.1 XM_017000161.1 XM_017000162.1 XM_017000163.1

SNP Detail

SNP ID Chromosome Location
rs1001445936 Chr.1: 239827356-239827356 on Build GRCh38
rs992925176 Chr.1: 239827069-239827069 on Build GRCh38
rs959882793 Chr.1: 239827360-239827360 on Build GRCh38
rs201663470 Chr.1: 239827346-239827346 on Build GRCh38
rs915232459 Chr.1: 239827165-239827165 on Build GRCh38
rs935365056 Chr.1: 239827147-239827148 on Build GRCh38
rs928011117 Chr.1: 239827142-239827142 on Build GRCh38
rs201700807 Chr.1: 239827322-239827322 on Build GRCh38
rs528014196 Chr.1: 239827261-239827261 on Build GRCh38
rs946985607 Chr.1: 239827306-239827306 on Build GRCh38
rs564760565 Chr.1: 239827308-239827308 on Build GRCh38
rs1010339061 Chr.1: 239826990-239826990 on Build GRCh38
rs1021305536 Chr.1: 239827001-239827001 on Build GRCh38
rs907975655 Chr.1: 239827072-239827072 on Build GRCh38
rs1021263472 Chr.1: 239827329-239827329 on Build GRCh38
rs201315393 Chr.1: 239827368-239827368 on Build GRCh38
rs993911276 Chr.1: 239827349-239827349 on Build GRCh38
rs1038077921 Chr.1: 239827115-239827115 on Build GRCh38
rs927078905 Chr.1: 239827043-239827043 on Build GRCh38
rs552862786 Chr.1: 239827269-239827269 on Build GRCh38
rs944821380 Chr.1: 239827325-239827325 on Build GRCh38
rs779384890 Chr.1: 239827305-239827305 on Build GRCh38
rs188998882 Chr.1: 239827355-239827355 on Build GRCh38
rs201022871 Chr.1: 239827307-239827307 on Build GRCh38
rs200262973 Chr.1: 239827359-239827359 on Build GRCh38
rs184667301 Chr.1: 239827089-239827089 on Build GRCh38
rs764386181 Chr.1: 239826986-239826986 on Build GRCh38
rs201055260 Chr.1: 239827217-239827217 on Build GRCh38
rs982170562 Chr.1: 239827130-239827130 on Build GRCh38
rs900892975 Chr.1: 239827344-239827344 on Build GRCh38
rs757343185 Chr.1: 239826972-239826972 on Build GRCh38
rs959896320 Chr.1: 239827067-239827067 on Build GRCh38
rs1040779346 Chr.1: 239827295-239827295 on Build GRCh38
rs151295192 Chr.1: 239827123-239827123 on Build GRCh38
rs890614603 Chr.1: 239827274-239827274 on Build GRCh38
rs932196386 Chr.1: 239827168-239827168 on Build GRCh38
rs199671600 Chr.1: 239827369-239827369 on Build GRCh38
rs1019333674 Chr.1: 239826935-239826935 on Build GRCh38
rs988571056 Chr.1: 239827155-239827155 on Build GRCh38
rs200353904 Chr.1: 239827273-239827273 on Build GRCh38
rs200401521 Chr.1: 239827340-239827340 on Build GRCh38
rs201800713 Chr.1: 239827264-239827264 on Build GRCh38
rs980019668 Chr.1: 239827028-239827028 on Build GRCh38
rs750555853 Chr.1: 239827314-239827314 on Build GRCh38
rs1019816185 Chr.1: 239826953-239826953 on Build GRCh38
rs1005428964 Chr.1: 239827358-239827358 on Build GRCh38
rs1009516742 Chr.1: 239827281-239827281 on Build GRCh38
rs201892041 Chr.1: 239827210-239827210 on Build GRCh38
rs1025576580 Chr.1: 239827037-239827037 on Build GRCh38
rs1007942424 Chr.1: 239826939-239826939 on Build GRCh38
rs968601967 Chr.1: 239827022-239827022 on Build GRCh38
rs199897692 Chr.1: 239827216-239827216 on Build GRCh38

View Full Product Details