Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 240092683-240093178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
-
UniGene
Hs.24889
Gene Chromosome Location
Chr.1: 240013776-240475189 on Build GRCh38
Transcripts
NM_001305424.1 NM_020066.4 XM_011544237.2 XM_017001837.1 XM_017001838.1 XM_017001839.1 XM_017001842.1 XM_017001843.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5542942 c.1423G>A p.E475K Chr.1: 240093103-240093103 on Build GRCh38
3804541 c.1090C>T p.Q364* Chr.1: 240092770-240092770 on Build GRCh38
5823423 c.1341G>T p.A447A Chr.1: 240093021-240093021 on Build GRCh38
5547590 c.1063_1068delCAGCTC p.L360_Q361delLQ Chr.1: 240092743-240092748 on Build GRCh38
4767787 c.1158C>T p.P386P Chr.1: 240092838-240092838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759662315 Chr.1: 240093124-240093124 on Build GRCh38
rs532298569 Chr.1: 240092766-240092767 on Build GRCh38
rs559793960 Chr.1: 240093139-240093139 on Build GRCh38
rs760728296 Chr.1: 240092744-240092744 on Build GRCh38
rs565189382 Chr.1: 240092723-240092724 on Build GRCh38
rs745427484 Chr.1: 240092726-240092726 on Build GRCh38
rs149103365 Chr.1: 240092812-240092812 on Build GRCh38
rs1038601475 Chr.1: 240093122-240093122 on Build GRCh38
rs776930344 Chr.1: 240092724-240092726 on Build GRCh38
rs200364388 Chr.1: 240092727-240092727 on Build GRCh38
rs967255135 Chr.1: 240093008-240093008 on Build GRCh38
rs968213008 Chr.1: 240093131-240093131 on Build GRCh38
rs753172887 Chr.1: 240092803-240092803 on Build GRCh38
rs761505012 Chr.1: 240092766-240092766 on Build GRCh38
rs772895522 Chr.1: 240093080-240093080 on Build GRCh38
rs1048931607 Chr.1: 240093010-240093010 on Build GRCh38
rs776623314 Chr.1: 240092765-240092765 on Build GRCh38
rs750588031 Chr.1: 240092786-240092786 on Build GRCh38
rs1032655654 Chr.1: 240093016-240093016 on Build GRCh38
rs556876873 Chr.1: 240093002-240093002 on Build GRCh38
rs766230062 Chr.1: 240092724-240092735 on Build GRCh38
rs575261046 Chr.1: 240093018-240093018 on Build GRCh38
rs534629981 Chr.1: 240092834-240092834 on Build GRCh38
rs769703635 Chr.1: 240093062-240093062 on Build GRCh38
rs768654005 Chr.1: 240092764-240092764 on Build GRCh38
rs773018609 Chr.1: 240092848-240092848 on Build GRCh38
rs867969810 Chr.1: 240092831-240092831 on Build GRCh38
rs778426244 Chr.1: 240092715-240092715 on Build GRCh38
rs781656437 Chr.1: 240092799-240092799 on Build GRCh38
rs1033849931 Chr.1: 240093143-240093143 on Build GRCh38
rs755777720 Chr.1: 240092767-240092769 on Build GRCh38
rs940251183 Chr.1: 240093098-240093098 on Build GRCh38
rs908717359 Chr.1: 240093095-240093095 on Build GRCh38
rs975549424 Chr.1: 240093024-240093024 on Build GRCh38
rs752321286 Chr.1: 240092746-240092748 on Build GRCh38
rs755491434 Chr.1: 240092793-240092793 on Build GRCh38
rs768778411 Chr.1: 240092829-240092829 on Build GRCh38
rs979681813 Chr.1: 240092849-240092849 on Build GRCh38
rs767816504 Chr.1: 240093023-240093023 on Build GRCh38
rs940088527 Chr.1: 240092947-240092947 on Build GRCh38
rs930480044 Chr.1: 240093006-240093006 on Build GRCh38
rs868406353 Chr.1: 240092952-240092952 on Build GRCh38
rs965778842 Chr.1: 240092819-240092819 on Build GRCh38
rs142232913 Chr.1: 240092871-240092871 on Build GRCh38
rs562296393 Chr.1: 240093037-240093037 on Build GRCh38
rs745532786 Chr.1: 240092815-240092815 on Build GRCh38
rs541790538 Chr.1: 240093123-240093123 on Build GRCh38
rs763166094 Chr.1: 240092869-240092869 on Build GRCh38
rs1033800034 Chr.1: 240092965-240092965 on Build GRCh38
rs751570120 Chr.1: 240092791-240092791 on Build GRCh38
rs968640066 Chr.1: 240092842-240092842 on Build GRCh38
rs771814202 Chr.1: 240092817-240092817 on Build GRCh38
rs537510607 Chr.1: 240092787-240092787 on Build GRCh38
rs369650259 Chr.1: 240092910-240092910 on Build GRCh38
rs533165347 Chr.1: 240092713-240092713 on Build GRCh38
rs779845544 Chr.1: 240092993-240092993 on Build GRCh38
rs754511896 Chr.1: 240093012-240093012 on Build GRCh38
rs1024222163 Chr.1: 240093004-240093004 on Build GRCh38
rs542716680 Chr.1: 240093036-240093036 on Build GRCh38
rs775094543 Chr.1: 240092735-240092735 on Build GRCh38
rs764400388 Chr.1: 240092959-240092959 on Build GRCh38
rs551900026 Chr.1: 240092714-240092714 on Build GRCh38
rs759609306 Chr.1: 240092908-240092908 on Build GRCh38
rs761713019 Chr.1: 240092838-240092838 on Build GRCh38
rs754168045 Chr.1: 240092976-240092976 on Build GRCh38
rs765531374 Chr.1: 240092784-240092784 on Build GRCh38
rs934144264 Chr.1: 240093045-240093045 on Build GRCh38
rs867121123 Chr.1: 240093120-240093120 on Build GRCh38
rs147858483 Chr.1: 240092887-240092887 on Build GRCh38
rs886043509 Chr.1: 240092955-240092955 on Build GRCh38
rs959329073 Chr.1: 240092875-240092875 on Build GRCh38
rs767102734 Chr.1: 240092742-240092743 on Build GRCh38
rs766474706 Chr.1: 240092790-240092790 on Build GRCh38
rs752855450 Chr.1: 240093096-240093096 on Build GRCh38
rs1009295852 Chr.1: 240093038-240093038 on Build GRCh38
rs769499457 Chr.1: 240092843-240092843 on Build GRCh38
rs901534417 Chr.1: 240093033-240093033 on Build GRCh38
rs1007515366 Chr.1: 240093019-240093019 on Build GRCh38
rs1040050645 Chr.1: 240093020-240093020 on Build GRCh38
rs746501698 Chr.1: 240092828-240092828 on Build GRCh38
rs748405938 Chr.1: 240092711-240092711 on Build GRCh38
rs11583501 Chr.1: 240092805-240092805 on Build GRCh38
rs574272337 Chr.1: 240093070-240093070 on Build GRCh38
rs1021104317 Chr.1: 240092840-240092840 on Build GRCh38
rs898517609 Chr.1: 240092889-240092889 on Build GRCh38
rs759794039 Chr.1: 240092915-240092915 on Build GRCh38
rs1037539031 Chr.1: 240092990-240092990 on Build GRCh38
rs977120019 Chr.1: 240092748-240092748 on Build GRCh38
rs756598447 Chr.1: 240092923-240092923 on Build GRCh38
rs201538863 Chr.1: 240092992-240092992 on Build GRCh38

View Full Product Details