Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241502161-241502691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FH
Entrez Gene ID
2271
Gene Name
fumarate hydratase
Gene Aliases
FMRD, HLRCC, LRCC, MCL, MCUL1
UniGene
Hs.592490
Gene Chromosome Location
Chr.1: 241497557-241519785 on Build GRCh38
Transcripts
NM_000143.3 XM_011544132.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6966851 c.1126C>G p.Q376E Chr.1: 241502553-241502553 on Build GRCh38
5607398 c.1113G>T p.K371N Chr.1: 241502566-241502566 on Build GRCh38
290796 c.1155C>T p.A385A Chr.1: 241502524-241502524 on Build GRCh38
6923302 c.1236G>A p.M412I Chr.1: 241502443-241502443 on Build GRCh38
5846004 c.1196G>A p.S399N Chr.1: 241502483-241502483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053612119 Chr.1: 241502353-241502353 on Build GRCh38
rs542511255 Chr.1: 241502394-241502394 on Build GRCh38
rs904463039 Chr.1: 241502528-241502528 on Build GRCh38
rs978096535 Chr.1: 241502270-241502270 on Build GRCh38
rs727503926 Chr.1: 241502525-241502525 on Build GRCh38
rs777797874 Chr.1: 241502581-241502583 on Build GRCh38
rs769517349 Chr.1: 241502406-241502406 on Build GRCh38
rs760433637 Chr.1: 241502338-241502338 on Build GRCh38
rs539939778 Chr.1: 241502259-241502259 on Build GRCh38
rs767865646 Chr.1: 241502595-241502595 on Build GRCh38
rs774493741 Chr.1: 241502452-241502452 on Build GRCh38
rs924025449 Chr.1: 241502285-241502285 on Build GRCh38
rs763667698 Chr.1: 241502609-241502609 on Build GRCh38
rs753996659 Chr.1: 241502493-241502493 on Build GRCh38
rs876660830 Chr.1: 241502516-241502516 on Build GRCh38
rs778969674 Chr.1: 241502512-241502512 on Build GRCh38
rs876659362 Chr.1: 241502463-241502463 on Build GRCh38
rs944191775 Chr.1: 241502325-241502325 on Build GRCh38
rs797044974 Chr.1: 241502469-241502469 on Build GRCh38
rs1009861325 Chr.1: 241502402-241502402 on Build GRCh38
rs373903301 Chr.1: 241502621-241502621 on Build GRCh38
rs863223974 Chr.1: 241502511-241502511 on Build GRCh38
rs199638673 Chr.1: 241502506-241502506 on Build GRCh38
rs568356071 Chr.1: 241502209-241502209 on Build GRCh38
rs753489352 Chr.1: 241502626-241502626 on Build GRCh38
rs931522797 Chr.1: 241502227-241502227 on Build GRCh38
rs758927661 Chr.1: 241502531-241502531 on Build GRCh38
rs147583475 Chr.1: 241502604-241502605 on Build GRCh38
rs200796606 Chr.1: 241502552-241502552 on Build GRCh38
rs764430466 Chr.1: 241502480-241502480 on Build GRCh38
rs755041218 Chr.1: 241502421-241502421 on Build GRCh38
rs750447792 Chr.1: 241502522-241502522 on Build GRCh38
rs587778362 Chr.1: 241502541-241502541 on Build GRCh38
rs878853690 Chr.1: 241502495-241502495 on Build GRCh38
rs752558644 Chr.1: 241502467-241502467 on Build GRCh38
rs199913699 Chr.1: 241502612-241502612 on Build GRCh38
rs548551026 Chr.1: 241502205-241502205 on Build GRCh38
rs770548498 Chr.1: 241502614-241502614 on Build GRCh38
rs972688066 Chr.1: 241502207-241502207 on Build GRCh38
rs367638510 Chr.1: 241502358-241502358 on Build GRCh38
rs863224005 Chr.1: 241502567-241502567 on Build GRCh38
rs759548413 Chr.1: 241502455-241502455 on Build GRCh38
rs886039366 Chr.1: 241502474-241502474 on Build GRCh38
rs554187124 Chr.1: 241502218-241502218 on Build GRCh38
rs138413334 Chr.1: 241502228-241502228 on Build GRCh38
rs2185626 Chr.1: 241502210-241502210 on Build GRCh38
rs886039365 Chr.1: 241502535-241502535 on Build GRCh38
rs149241949 Chr.1: 241502429-241502429 on Build GRCh38
rs398123162 Chr.1: 241502479-241502479 on Build GRCh38
rs398123160 Chr.1: 241502553-241502553 on Build GRCh38
rs863224007 Chr.1: 241502490-241502490 on Build GRCh38
rs760360724 Chr.1: 241502475-241502475 on Build GRCh38
rs959933221 Chr.1: 241502574-241502574 on Build GRCh38
rs779534691 Chr.1: 241502393-241502393 on Build GRCh38
rs949394252 Chr.1: 241502243-241502243 on Build GRCh38
rs374494513 Chr.1: 241502438-241502438 on Build GRCh38
rs780411318 Chr.1: 241502399-241502399 on Build GRCh38
rs398123161 Chr.1: 241502550-241502550 on Build GRCh38
rs781466938 Chr.1: 241502540-241502541 on Build GRCh38
rs911420888 Chr.1: 241502320-241502320 on Build GRCh38
rs863223988 Chr.1: 241502537-241502540 on Build GRCh38
rs1057517735 Chr.1: 241502450-241502450 on Build GRCh38
rs891537769 Chr.1: 241502398-241502398 on Build GRCh38
rs201381362 Chr.1: 241502448-241502448 on Build GRCh38
rs777538608 Chr.1: 241502598-241502598 on Build GRCh38
rs767500963 Chr.1: 241502460-241502460 on Build GRCh38
rs747325945 Chr.1: 241502405-241502405 on Build GRCh38
rs919900269 Chr.1: 241502653-241502653 on Build GRCh38
rs876660446 Chr.1: 241502559-241502559 on Build GRCh38
rs863224006 Chr.1: 241502533-241502533 on Build GRCh38
rs1040836699 Chr.1: 241502346-241502346 on Build GRCh38
rs199782639 Chr.1: 241502407-241502407 on Build GRCh38
rs201600104 Chr.1: 241502604-241502604 on Build GRCh38
rs1027590634 Chr.1: 241502217-241502217 on Build GRCh38
rs972740195 Chr.1: 241502637-241502637 on Build GRCh38
rs542014575 Chr.1: 241502560-241502560 on Build GRCh38

View Full Product Details