Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244352536-244353079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf100
Entrez Gene ID
200159
Gene Name
chromosome 1 open reading frame 100
Gene Aliases
-
UniGene
Hs.442703
Gene Chromosome Location
Chr.1: 244352063-244389896 on Build GRCh38
Transcripts
NM_001012970.2 NM_001276348.1 NM_001276349.1 XM_011544124.2 XM_011544125.2 XM_011544128.2 XM_011544129.2 XM_011544130.2 XM_017000571.1 XM_017000572.1

SNP Detail

SNP ID Chromosome Location
rs563163149 Chr.1: 244352592-244352592 on Build GRCh38
rs908012797 Chr.1: 244352834-244352834 on Build GRCh38
rs913774435 Chr.1: 244352754-244352754 on Build GRCh38
rs895695356 Chr.1: 244352675-244352675 on Build GRCh38
rs886615846 Chr.1: 244352591-244352591 on Build GRCh38
rs559505044 Chr.1: 244352743-244352743 on Build GRCh38
rs576593453 Chr.1: 244352748-244352748 on Build GRCh38
rs563842049 Chr.1: 244352761-244352761 on Build GRCh38
rs746473313 Chr.1: 244352745-244352745 on Build GRCh38
rs568992341 Chr.1: 244352954-244352955 on Build GRCh38
rs898851966 Chr.1: 244352855-244352855 on Build GRCh38
rs1014463736 Chr.1: 244352971-244352971 on Build GRCh38
rs576684134 Chr.1: 244352606-244352606 on Build GRCh38
rs778102731 Chr.1: 244352697-244352697 on Build GRCh38
rs973169562 Chr.1: 244352847-244352847 on Build GRCh38
rs949921966 Chr.1: 244352962-244352962 on Build GRCh38
rs189439881 Chr.1: 244352949-244352949 on Build GRCh38
rs750685301 Chr.1: 244352866-244352866 on Build GRCh38
rs527328681 Chr.1: 244352726-244352726 on Build GRCh38
rs113788700 Chr.1: 244352974-244352974 on Build GRCh38
rs1001638394 Chr.1: 244353028-244353028 on Build GRCh38
rs561333405 Chr.1: 244352723-244352723 on Build GRCh38
rs886685635 Chr.1: 244352950-244352950 on Build GRCh38
rs10927202 Chr.1: 244353032-244353032 on Build GRCh38
rs1032426935 Chr.1: 244353034-244353034 on Build GRCh38
rs745341459 Chr.1: 244352852-244352852 on Build GRCh38
rs182224984 Chr.1: 244352837-244352837 on Build GRCh38
rs903031601 Chr.1: 244352725-244352725 on Build GRCh38
rs4658628 Chr.1: 244352788-244352788 on Build GRCh38
rs7538448 Chr.1: 244353010-244353010 on Build GRCh38
rs995084652 Chr.1: 244352559-244352559 on Build GRCh38
rs546089422 Chr.1: 244352571-244352571 on Build GRCh38
rs900243553 Chr.1: 244352627-244352627 on Build GRCh38
rs542537348 Chr.1: 244352720-244352720 on Build GRCh38
rs911678133 Chr.1: 244352804-244352804 on Build GRCh38
rs549727831 Chr.1: 244352798-244352798 on Build GRCh38
rs974755913 Chr.1: 244352746-244352746 on Build GRCh38
rs768100781 Chr.1: 244352827-244352827 on Build GRCh38
rs773756153 Chr.1: 244352757-244352757 on Build GRCh38
rs1026951616 Chr.1: 244352560-244352560 on Build GRCh38
rs184511226 Chr.1: 244352896-244352896 on Build GRCh38
rs970121201 Chr.1: 244352638-244352638 on Build GRCh38
rs957576788 Chr.1: 244352732-244352732 on Build GRCh38
rs752030723 Chr.1: 244352596-244352596 on Build GRCh38
rs945232673 Chr.1: 244352774-244352774 on Build GRCh38
rs756512732 Chr.1: 244352934-244352934 on Build GRCh38
rs558055004 Chr.1: 244352716-244352716 on Build GRCh38

View Full Product Details