Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244375051-244375562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf100
Entrez Gene ID
200159
Gene Name
chromosome 1 open reading frame 100
Gene Aliases
-
UniGene
Hs.442703
Gene Chromosome Location
Chr.1: 244352063-244389896 on Build GRCh38
Transcripts
NM_001012970.2 NM_001276348.1 NM_001276349.1 XM_011544124.2 XM_011544125.2 XM_011544128.2 XM_011544129.2 XM_011544130.2 XM_017000571.1 XM_017000572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4030329 c.86T>C p.V29A Chr.1: 244375401-244375401 on Build GRCh38
5380304 c.77G>A p.G26E Chr.1: 244375392-244375392 on Build GRCh38
6125545 c.111G>T p.L37L Chr.1: 244375426-244375426 on Build GRCh38
7358194 c.68C>T p.A23V Chr.1: 244375383-244375383 on Build GRCh38
5846042 c.104G>A p.G35E Chr.1: 244375419-244375419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142343898 Chr.1: 244375400-244375400 on Build GRCh38
rs557258239 Chr.1: 244375447-244375447 on Build GRCh38
rs555471362 Chr.1: 244375213-244375213 on Build GRCh38
rs528793930 Chr.1: 244375533-244375533 on Build GRCh38
rs749990466 Chr.1: 244375458-244375458 on Build GRCh38
rs779628965 Chr.1: 244375463-244375463 on Build GRCh38
rs182389723 Chr.1: 244375298-244375298 on Build GRCh38
rs571466263 Chr.1: 244375428-244375428 on Build GRCh38
rs764007366 Chr.1: 244375330-244375330 on Build GRCh38
rs1026661511 Chr.1: 244375281-244375281 on Build GRCh38
rs780431582 Chr.1: 244375362-244375362 on Build GRCh38
rs747396414 Chr.1: 244375379-244375379 on Build GRCh38
rs745533738 Chr.1: 244375346-244375346 on Build GRCh38
rs756049985 Chr.1: 244375324-244375324 on Build GRCh38
rs759826895 Chr.1: 244375139-244375139 on Build GRCh38
rs185427099 Chr.1: 244375347-244375347 on Build GRCh38
rs556280726 Chr.1: 244375148-244375148 on Build GRCh38
rs749438390 Chr.1: 244375516-244375516 on Build GRCh38
rs979367725 Chr.1: 244375394-244375394 on Build GRCh38
rs370180345 Chr.1: 244375296-244375296 on Build GRCh38
rs971732522 Chr.1: 244375275-244375275 on Build GRCh38
rs564714753 Chr.1: 244375308-244375308 on Build GRCh38
rs139431277 Chr.1: 244375383-244375383 on Build GRCh38
rs755301856 Chr.1: 244375477-244375477 on Build GRCh38
rs145778363 Chr.1: 244375399-244375399 on Build GRCh38
rs747338685 Chr.1: 244375467-244375467 on Build GRCh38
rs763747834 Chr.1: 244375532-244375532 on Build GRCh38
rs748677260 Chr.1: 244375485-244375485 on Build GRCh38
rs773374923 Chr.1: 244375385-244375385 on Build GRCh38
rs1028629818 Chr.1: 244375164-244375164 on Build GRCh38
rs541906247 Chr.1: 244375185-244375185 on Build GRCh38
rs190548594 Chr.1: 244375232-244375232 on Build GRCh38
rs1011044067 Chr.1: 244375153-244375153 on Build GRCh38
rs115000493 Chr.1: 244375133-244375133 on Build GRCh38
rs993682834 Chr.1: 244375536-244375536 on Build GRCh38
rs1052198349 Chr.1: 244375421-244375421 on Build GRCh38
rs888867988 Chr.1: 244375163-244375163 on Build GRCh38
rs1052145519 Chr.1: 244375126-244375126 on Build GRCh38
rs901615854 Chr.1: 244375456-244375456 on Build GRCh38
rs536937363 Chr.1: 244375444-244375444 on Build GRCh38
rs140735071 Chr.1: 244375389-244375389 on Build GRCh38
rs777801174 Chr.1: 244375510-244375510 on Build GRCh38
rs370643169 Chr.1: 244375507-244375507 on Build GRCh38
rs759490000 Chr.1: 244375330-244375331 on Build GRCh38
rs769190584 Chr.1: 244375381-244375381 on Build GRCh38
rs966123247 Chr.1: 244375237-244375237 on Build GRCh38
rs150899045 Chr.1: 244375382-244375382 on Build GRCh38
rs199968599 Chr.1: 244375408-244375408 on Build GRCh38
rs764037259 Chr.1: 244375425-244375425 on Build GRCh38
rs369582776 Chr.1: 244375484-244375484 on Build GRCh38
rs1050538846 Chr.1: 244375479-244375479 on Build GRCh38
rs1016524066 Chr.1: 244375518-244375518 on Build GRCh38
rs1035778308 Chr.1: 244375294-244375294 on Build GRCh38
rs561680348 Chr.1: 244375154-244375154 on Build GRCh38
rs180744835 Chr.1: 244375395-244375395 on Build GRCh38
rs373073945 Chr.1: 244375337-244375337 on Build GRCh38
rs533410488 Chr.1: 244375341-244375341 on Build GRCh38
rs1052419984 Chr.1: 244375420-244375420 on Build GRCh38
rs889395550 Chr.1: 244375168-244375168 on Build GRCh38
rs758116124 Chr.1: 244375461-244375461 on Build GRCh38
rs151220322 Chr.1: 244375460-244375460 on Build GRCh38
rs999266580 Chr.1: 244375113-244375113 on Build GRCh38
rs777726055 Chr.1: 244375283-244375283 on Build GRCh38
rs796206325 Chr.1: 244375527-244375527 on Build GRCh38
rs962196640 Chr.1: 244375259-244375259 on Build GRCh38

View Full Product Details