Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244842768-244843304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX20
Entrez Gene ID
116228
Gene Name
COX20, cytochrome c oxidase assembly factor
Gene Aliases
FAM36A
UniGene
Hs.411490
Gene Chromosome Location
Chr.1: 244835306-244845063 on Build GRCh38
Transcripts
NM_001312871.1 NM_001312872.1 NM_001312873.1 NM_001312874.1 NM_198076.5 NR_132419.1 NR_132420.1 NR_132421.1 XM_017001115.1 XM_017001116.1 XM_017001117.1

Gene Symbol
HNRNPU
Entrez Gene ID
3192
Gene Name
heterogeneous nuclear ribonucleoprotein U
Gene Aliases
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U
UniGene
Hs.106212
Gene Chromosome Location
Chr.1: 244842123-244864720 on Build GRCh38
Transcripts
NM_001312871.1 NM_001312872.1 NM_001312873.1 NM_001312874.1 NM_198076.5 NR_132419.1 NR_132420.1 NR_132421.1 XM_017001115.1 XM_017001116.1 XM_017001117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641715 c.258-2A>G p.? Chr.1: 244843039-244843039 on Build GRCh38
3486407 c.307G>A p.G103S Chr.1: 244843126-244843126 on Build GRCh38
7185550 c.288A>G p.K96K Chr.1: 244843107-244843107 on Build GRCh38
4748233 c.332_334delTAT p.L112delL Chr.1: 244843115-244843117 on Build GRCh38
2051454 c.266G>T p.R89I Chr.1: 244843085-244843085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780760696 Chr.1: 244843047-244843047 on Build GRCh38
rs1050572930 Chr.1: 244842807-244842807 on Build GRCh38
rs749626450 Chr.1: 244843077-244843077 on Build GRCh38
rs948087697 Chr.1: 244842937-244842937 on Build GRCh38
rs777326199 Chr.1: 244843076-244843076 on Build GRCh38
rs988125986 Chr.1: 244843114-244843114 on Build GRCh38
rs951626382 Chr.1: 244842833-244842833 on Build GRCh38
rs889035347 Chr.1: 244843118-244843118 on Build GRCh38
rs868648645 Chr.1: 244843033-244843033 on Build GRCh38
rs763515115 Chr.1: 244843223-244843223 on Build GRCh38
rs942254694 Chr.1: 244843151-244843151 on Build GRCh38
rs757549228 Chr.1: 244843154-244843154 on Build GRCh38
rs776974675 Chr.1: 244843157-244843158 on Build GRCh38
rs749004480 Chr.1: 244843146-244843146 on Build GRCh38
rs747702163 Chr.1: 244843053-244843053 on Build GRCh38
rs969459231 Chr.1: 244843052-244843052 on Build GRCh38
rs1025787315 Chr.1: 244842827-244842827 on Build GRCh38
rs59190751 Chr.1: 244843194-244843195 on Build GRCh38
rs1023227356 Chr.1: 244843025-244843025 on Build GRCh38
rs560330031 Chr.1: 244842901-244842901 on Build GRCh38
rs775660475 Chr.1: 244843094-244843094 on Build GRCh38
rs150172035 Chr.1: 244843088-244843088 on Build GRCh38
rs767462980 Chr.1: 244843206-244843206 on Build GRCh38
rs1015895063 Chr.1: 244842898-244842898 on Build GRCh38
rs751606534 Chr.1: 244843046-244843046 on Build GRCh38
rs771340542 Chr.1: 244843081-244843081 on Build GRCh38
rs373502901 Chr.1: 244843138-244843138 on Build GRCh38
rs779262148 Chr.1: 244843155-244843155 on Build GRCh38
rs865889424 Chr.1: 244843196-244843196 on Build GRCh38
rs1037851042 Chr.1: 244843156-244843156 on Build GRCh38
rs863223946 Chr.1: 244843045-244843045 on Build GRCh38
rs993198530 Chr.1: 244842907-244842907 on Build GRCh38
rs766397584 Chr.1: 244843041-244843041 on Build GRCh38
rs141476240 Chr.1: 244843159-244843159 on Build GRCh38
rs759357042 Chr.1: 244843199-244843199 on Build GRCh38
rs371098560 Chr.1: 244843128-244843128 on Build GRCh38
rs895700293 Chr.1: 244843236-244843236 on Build GRCh38
rs913979972 Chr.1: 244843275-244843275 on Build GRCh38
rs78677269 Chr.1: 244843005-244843005 on Build GRCh38
rs935322065 Chr.1: 244843085-244843085 on Build GRCh38
rs141561272 Chr.1: 244843158-244843158 on Build GRCh38
rs755816815 Chr.1: 244843055-244843055 on Build GRCh38
rs745397579 Chr.1: 244843026-244843030 on Build GRCh38
rs1035585706 Chr.1: 244842931-244842931 on Build GRCh38
rs397829223 Chr.1: 244843067-244843067 on Build GRCh38
rs200065889 Chr.1: 244843038-244843038 on Build GRCh38
rs959962433 Chr.1: 244842972-244842972 on Build GRCh38
rs1012840418 Chr.1: 244843278-244843278 on Build GRCh38
rs776991341 Chr.1: 244843179-244843179 on Build GRCh38
rs1053697 Chr.1: 244843137-244843137 on Build GRCh38
rs61749963 Chr.1: 244843172-244843172 on Build GRCh38
rs750263153 Chr.1: 244843022-244843022 on Build GRCh38
rs769116815 Chr.1: 244843138-244843138 on Build GRCh38
rs777465156 Chr.1: 244843142-244843142 on Build GRCh38
rs776767802 Chr.1: 244843120-244843120 on Build GRCh38
rs939457834 Chr.1: 244843261-244843264 on Build GRCh38
rs532347642 Chr.1: 244842946-244842946 on Build GRCh38
rs988209507 Chr.1: 244843265-244843266 on Build GRCh38
rs747091122 Chr.1: 244843165-244843165 on Build GRCh38
rs984397609 Chr.1: 244842849-244842850 on Build GRCh38
rs888843696 Chr.1: 244842832-244842832 on Build GRCh38
rs764346782 Chr.1: 244843113-244843113 on Build GRCh38
rs766346210 Chr.1: 244843131-244843131 on Build GRCh38
rs771647113 Chr.1: 244843090-244843090 on Build GRCh38
rs778454364 Chr.1: 244843015-244843018 on Build GRCh38
rs767647989 Chr.1: 244842997-244842997 on Build GRCh38
rs976924054 Chr.1: 244842826-244842826 on Build GRCh38
rs1006393699 Chr.1: 244842847-244842847 on Build GRCh38
rs923502888 Chr.1: 244843117-244843117 on Build GRCh38
rs759911217 Chr.1: 244843105-244843105 on Build GRCh38
rs771987123 Chr.1: 244843093-244843093 on Build GRCh38
rs766221684 Chr.1: 244843191-244843191 on Build GRCh38
rs11544344 Chr.1: 244843127-244843127 on Build GRCh38
rs752630666 Chr.1: 244843207-244843207 on Build GRCh38
rs764892273 Chr.1: 244843021-244843021 on Build GRCh38
rs760225037 Chr.1: 244843213-244843213 on Build GRCh38
rs778452416 Chr.1: 244843265-244843270 on Build GRCh38
rs984299020 Chr.1: 244843167-244843167 on Build GRCh38
rs925425021 Chr.1: 244843068-244843068 on Build GRCh38
rs779792502 Chr.1: 244843108-244843110 on Build GRCh38
rs762438072 Chr.1: 244843188-244843189 on Build GRCh38
rs145325896 Chr.1: 244843135-244843135 on Build GRCh38
rs775901983 Chr.1: 244843160-244843160 on Build GRCh38
rs774833234 Chr.1: 244843086-244843086 on Build GRCh38
rs774313774 Chr.1: 244843198-244843198 on Build GRCh38
rs892277959 Chr.1: 244843015-244843015 on Build GRCh38
rs747291859 Chr.1: 244843115-244843117 on Build GRCh38
rs762133192 Chr.1: 244843190-244843190 on Build GRCh38
rs760963966 Chr.1: 244843013-244843013 on Build GRCh38
rs751479035 Chr.1: 244843133-244843133 on Build GRCh38

View Full Product Details