Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 246566640-246567135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNST
Entrez Gene ID
163882
Gene Name
consortin, connexin sorting protein
Gene Aliases
C1orf71, PPP1R64
UniGene
Hs.368353
Gene Chromosome Location
Chr.1: 246566337-246668587 on Build GRCh38
Transcripts
NM_001139459.1 NM_152609.2 XM_005273083.4 XM_011544111.1 XM_011544112.1 XM_011544113.1 XM_011544114.2 XM_017000487.1 XM_017000488.1 XM_017000489.1

SNP Detail

SNP ID Chromosome Location
rs988651483 Chr.1: 246567009-246567009 on Build GRCh38
rs3120714 Chr.1: 246566788-246566788 on Build GRCh38
rs1018315324 Chr.1: 246566696-246566696 on Build GRCh38
rs1018278323 Chr.1: 246566813-246566813 on Build GRCh38
rs760320509 Chr.1: 246566814-246566814 on Build GRCh38
rs927051778 Chr.1: 246567034-246567034 on Build GRCh38
rs974649857 Chr.1: 246566723-246566723 on Build GRCh38
rs1000855418 Chr.1: 246567091-246567091 on Build GRCh38
rs976870969 Chr.1: 246566815-246566815 on Build GRCh38
rs925871561 Chr.1: 246566733-246566733 on Build GRCh38
rs1000143870 Chr.1: 246567011-246567011 on Build GRCh38
rs114750587 Chr.1: 246566820-246566820 on Build GRCh38
rs916650489 Chr.1: 246566922-246566922 on Build GRCh38
rs1043720086 Chr.1: 246566973-246566973 on Build GRCh38
rs939392936 Chr.1: 246566742-246566742 on Build GRCh38
rs35278012 Chr.1: 246566810-246566811 on Build GRCh38
rs957220357 Chr.1: 246566734-246566734 on Build GRCh38
rs957542406 Chr.1: 246566936-246566936 on Build GRCh38
rs12123751 Chr.1: 246566749-246566749 on Build GRCh38
rs531247691 Chr.1: 246567041-246567041 on Build GRCh38
rs1055256395 Chr.1: 246566908-246566908 on Build GRCh38
rs542435328 Chr.1: 246566878-246566878 on Build GRCh38
rs1018375582 Chr.1: 246566830-246566830 on Build GRCh38
rs909830522 Chr.1: 246566869-246566869 on Build GRCh38
rs753225500 Chr.1: 246567085-246567085 on Build GRCh38
rs974183243 Chr.1: 246567036-246567036 on Build GRCh38
rs565816997 Chr.1: 246566690-246566690 on Build GRCh38
rs75812177 Chr.1: 246567090-246567090 on Build GRCh38
rs572092503 Chr.1: 246566739-246566739 on Build GRCh38
rs1038582186 Chr.1: 246566776-246566776 on Build GRCh38
rs976461340 Chr.1: 246566676-246566676 on Build GRCh38
rs1019509462 Chr.1: 246566810-246566810 on Build GRCh38
rs1028051829 Chr.1: 246567073-246567073 on Build GRCh38
rs983732277 Chr.1: 246566856-246566856 on Build GRCh38
rs971224799 Chr.1: 246567022-246567022 on Build GRCh38
rs909799351 Chr.1: 246566695-246566695 on Build GRCh38
rs1025717249 Chr.1: 246566683-246566683 on Build GRCh38
rs540985679 Chr.1: 246566923-246566923 on Build GRCh38
rs564161790 Chr.1: 246566943-246566943 on Build GRCh38
rs902752367 Chr.1: 246566784-246566784 on Build GRCh38
rs3806271 Chr.1: 246567062-246567062 on Build GRCh38
rs182907112 Chr.1: 246567059-246567059 on Build GRCh38
rs145240290 Chr.1: 246566872-246566872 on Build GRCh38
rs898774650 Chr.1: 246566783-246566783 on Build GRCh38
rs775825644 Chr.1: 246566691-246566691 on Build GRCh38
rs55972918 Chr.1: 246566903-246566903 on Build GRCh38
rs1055138120 Chr.1: 246566748-246566748 on Build GRCh38
rs555576276 Chr.1: 246566887-246566887 on Build GRCh38
rs902785714 Chr.1: 246566993-246566993 on Build GRCh38
rs964051923 Chr.1: 246566705-246566705 on Build GRCh38
rs3795474 Chr.1: 246566688-246566688 on Build GRCh38
rs1010910849 Chr.1: 246566765-246566765 on Build GRCh38
rs927754177 Chr.1: 246566881-246566881 on Build GRCh38
rs895183323 Chr.1: 246566763-246566763 on Build GRCh38
rs1039804322 Chr.1: 246567086-246567086 on Build GRCh38
rs761388024 Chr.1: 246567088-246567088 on Build GRCh38
rs937012509 Chr.1: 246567035-246567035 on Build GRCh38
rs953750929 Chr.1: 246566855-246566855 on Build GRCh38
rs558119037 Chr.1: 246566865-246566865 on Build GRCh38
rs946729974 Chr.1: 246566962-246566962 on Build GRCh38
rs1037259143 Chr.1: 246566740-246566740 on Build GRCh38
rs1008287110 Chr.1: 246566825-246566825 on Build GRCh38
rs557799802 Chr.1: 246566747-246566747 on Build GRCh38
rs577563358 Chr.1: 246566777-246566777 on Build GRCh38
rs902041143 Chr.1: 246567056-246567056 on Build GRCh38
rs906630500 Chr.1: 246566741-246566741 on Build GRCh38
rs1028019267 Chr.1: 246566828-246566828 on Build GRCh38
rs920171730 Chr.1: 246567040-246567040 on Build GRCh38
rs891275876 Chr.1: 246567081-246567081 on Build GRCh38
rs1040553530 Chr.1: 246567025-246567025 on Build GRCh38

View Full Product Details