Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 309949-310448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2C
Entrez Gene ID
22982
Gene Name
disco interacting protein 2 homolog C
Gene Aliases
KIAA0934
UniGene
Hs.432397
Gene Chromosome Location
Chr.10: 274190-689710 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_005252430.2 XM_011519428.2 XM_011519429.2 XM_011519430.2 XM_011519431.2 XM_011519432.1 XM_011519433.2 XM_017015977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086847 c.3935G>A p.G1312E Chr.10: 310082-310082 on Build GRCh38
917895 c.3981C>T p.H1327H Chr.10: 310036-310036 on Build GRCh38
1956028 c.3965T>C p.M1322T Chr.10: 310052-310052 on Build GRCh38
1956027 c.3986G>A p.R1329K Chr.10: 310031-310031 on Build GRCh38
5573374 c.3986+2T>C p.? Chr.10: 310029-310029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896563366 Chr.10: 310360-310360 on Build GRCh38
rs748877695 Chr.10: 310007-310007 on Build GRCh38
rs776938070 Chr.10: 310045-310045 on Build GRCh38
rs45608133 Chr.10: 310140-310140 on Build GRCh38
rs191755936 Chr.10: 310111-310111 on Build GRCh38
rs528283488 Chr.10: 310400-310400 on Build GRCh38
rs187332353 Chr.10: 309999-309999 on Build GRCh38
rs768616009 Chr.10: 310035-310035 on Build GRCh38
rs975891843 Chr.10: 310423-310423 on Build GRCh38
rs974135568 Chr.10: 310359-310359 on Build GRCh38
rs776165992 Chr.10: 310066-310066 on Build GRCh38
rs150410788 Chr.10: 309977-309977 on Build GRCh38
rs763423347 Chr.10: 310105-310105 on Build GRCh38
rs769224552 Chr.10: 310003-310003 on Build GRCh38
rs761664186 Chr.10: 310038-310038 on Build GRCh38
rs1024192785 Chr.10: 310289-310289 on Build GRCh38
rs912787472 Chr.10: 310379-310379 on Build GRCh38
rs1056591229 Chr.10: 310347-310347 on Build GRCh38
rs141790996 Chr.10: 310217-310218 on Build GRCh38
rs749173027 Chr.10: 310017-310017 on Build GRCh38
rs771116499 Chr.10: 310041-310041 on Build GRCh38
rs756427621 Chr.10: 309989-309990 on Build GRCh38
rs987023676 Chr.10: 310369-310370 on Build GRCh38
rs200666436 Chr.10: 310016-310016 on Build GRCh38
rs1034460603 Chr.10: 310335-310335 on Build GRCh38
rs767159040 Chr.10: 310129-310129 on Build GRCh38
rs112535917 Chr.10: 310024-310024 on Build GRCh38
rs148914383 Chr.10: 310160-310160 on Build GRCh38
rs866034398 Chr.10: 310055-310055 on Build GRCh38
rs550961966 Chr.10: 310269-310269 on Build GRCh38
rs184524196 Chr.10: 310132-310132 on Build GRCh38
rs573499024 Chr.10: 310150-310150 on Build GRCh38
rs756654974 Chr.10: 310228-310228 on Build GRCh38
rs751923146 Chr.10: 310118-310118 on Build GRCh38
rs572122207 Chr.10: 310399-310399 on Build GRCh38
rs561118389 Chr.10: 310306-310306 on Build GRCh38
rs754938192 Chr.10: 310141-310141 on Build GRCh38
rs779681538 Chr.10: 309990-309990 on Build GRCh38
rs374162032 Chr.10: 309990-309990 on Build GRCh38
rs145570975 Chr.10: 310275-310275 on Build GRCh38
rs777955587 Chr.10: 309982-309982 on Build GRCh38
rs1012215260 Chr.10: 310361-310361 on Build GRCh38
rs138147106 Chr.10: 310134-310134 on Build GRCh38
rs759695360 Chr.10: 310059-310059 on Build GRCh38
rs575803933 Chr.10: 310259-310259 on Build GRCh38
rs200561296 Chr.10: 310036-310036 on Build GRCh38
rs900227190 Chr.10: 310158-310158 on Build GRCh38
rs746444115 Chr.10: 310220-310220 on Build GRCh38
rs528126847 Chr.10: 310120-310120 on Build GRCh38
rs757787462 Chr.10: 310060-310060 on Build GRCh38
rs780610364 Chr.10: 310232-310232 on Build GRCh38
rs753609319 Chr.10: 309978-309978 on Build GRCh38
rs956579506 Chr.10: 310403-310403 on Build GRCh38
rs1038529620 Chr.10: 310280-310280 on Build GRCh38
rs540306218 Chr.10: 310362-310362 on Build GRCh38
rs778896889 Chr.10: 310143-310143 on Build GRCh38
rs751934482 Chr.10: 309985-309985 on Build GRCh38
rs750772904 Chr.10: 309982-309986 on Build GRCh38
rs1050522723 Chr.10: 310254-310254 on Build GRCh38
rs757581603 Chr.10: 310122-310122 on Build GRCh38
rs891296602 Chr.10: 310123-310123 on Build GRCh38
rs941613878 Chr.10: 310277-310277 on Build GRCh38
rs994243195 Chr.10: 310325-310325 on Build GRCh38
rs768951251 Chr.10: 310281-310281 on Build GRCh38
rs780880911 Chr.10: 309993-309993 on Build GRCh38
rs545102882 Chr.10: 310154-310154 on Build GRCh38
rs187448472 Chr.10: 310363-310363 on Build GRCh38
rs377632100 Chr.10: 310108-310108 on Build GRCh38
rs574909682 Chr.10: 310316-310316 on Build GRCh38
rs34534378 Chr.10: 309981-309982 on Build GRCh38

View Full Product Details