Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 4963057-4963591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1C1
Entrez Gene ID
1645
Gene Name
aldo-keto reductase family 1 member C1
Gene Aliases
2-ALPHA-HSD, 20-ALPHA-HSD, C9, DD1, DD1/DD2, DDH, DDH1, H-37, HAKRC, HBAB, MBAB
UniGene
Hs.460260
Gene Chromosome Location
Chr.10: 4963262-4979967 on Build GRCh38
Transcripts
NM_001353.5 XM_017015791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5415383 c.1A>G p.M1V Chr.10: 4963445-4963445 on Build GRCh38
918534 c.3G>T p.M1I Chr.10: 4963447-4963447 on Build GRCh38
5817661 c.9G>T p.S3S Chr.10: 4963453-4963453 on Build GRCh38
5368691 c.4G>A p.D2N Chr.10: 4963448-4963448 on Build GRCh38
5771388 c.32A>G p.N11S Chr.10: 4963476-4963476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752938448 Chr.10: 4963466-4963466 on Build GRCh38
rs547565374 Chr.10: 4963395-4963395 on Build GRCh38
rs142261618 Chr.10: 4963285-4963285 on Build GRCh38
rs1031281488 Chr.10: 4963236-4963236 on Build GRCh38
rs142429052 Chr.10: 4963461-4963461 on Build GRCh38
rs71499618 Chr.10: 4963408-4963411 on Build GRCh38
rs777665259 Chr.10: 4963422-4963422 on Build GRCh38
rs546745235 Chr.10: 4963198-4963198 on Build GRCh38
rs878915696 Chr.10: 4963531-4963531 on Build GRCh38
rs71477884 Chr.10: 4963081-4963081 on Build GRCh38
rs1138566 Chr.10: 4963459-4963459 on Build GRCh38
rs780527109 Chr.10: 4963427-4963427 on Build GRCh38
rs766325220 Chr.10: 4963396-4963396 on Build GRCh38
rs771188283 Chr.10: 4963452-4963452 on Build GRCh38
rs71477885 Chr.10: 4963100-4963100 on Build GRCh38
rs757239913 Chr.10: 4963425-4963425 on Build GRCh38
rs570992649 Chr.10: 4963296-4963296 on Build GRCh38
rs71479127 Chr.10: 4963434-4963434 on Build GRCh38
rs575034166 Chr.10: 4963423-4963423 on Build GRCh38
rs200972237 Chr.10: 4963511-4963511 on Build GRCh38
rs529733760 Chr.10: 4963262-4963262 on Build GRCh38
rs763317381 Chr.10: 4963496-4963496 on Build GRCh38
rs200189716 Chr.10: 4963400-4963400 on Build GRCh38
rs759768301 Chr.10: 4963402-4963402 on Build GRCh38
rs879238708 Chr.10: 4963532-4963532 on Build GRCh38
rs558598949 Chr.10: 4963373-4963373 on Build GRCh38
rs748912524 Chr.10: 4963529-4963529 on Build GRCh38
rs930003545 Chr.10: 4963115-4963115 on Build GRCh38
rs765572995 Chr.10: 4963404-4963404 on Build GRCh38
rs569530119 Chr.10: 4963323-4963323 on Build GRCh38
rs71477886 Chr.10: 4963113-4963113 on Build GRCh38
rs2904798 Chr.10: 4963555-4963555 on Build GRCh38
rs531262749 Chr.10: 4963099-4963099 on Build GRCh38
rs546162868 Chr.10: 4963111-4963111 on Build GRCh38
rs749004430 Chr.10: 4963445-4963445 on Build GRCh38
rs199732938 Chr.10: 4963410-4963410 on Build GRCh38
rs770137049 Chr.10: 4963413-4963414 on Build GRCh38
rs771261308 Chr.10: 4963548-4963548 on Build GRCh38
rs1040220938 Chr.10: 4963469-4963469 on Build GRCh38
rs768389441 Chr.10: 4963538-4963538 on Build GRCh38
rs7069696 Chr.10: 4963357-4963357 on Build GRCh38
rs779576403 Chr.10: 4963527-4963527 on Build GRCh38
rs1031229568 Chr.10: 4963517-4963517 on Build GRCh38
rs753985891 Chr.10: 4963399-4963399 on Build GRCh38
rs998425471 Chr.10: 4963203-4963203 on Build GRCh38
rs535931014 Chr.10: 4963453-4963453 on Build GRCh38
rs1018440518 Chr.10: 4963176-4963176 on Build GRCh38
rs552755181 Chr.10: 4963164-4963164 on Build GRCh38
rs777937103 Chr.10: 4963539-4963539 on Build GRCh38
rs565447061 Chr.10: 4963245-4963245 on Build GRCh38
rs750415442 Chr.10: 4963519-4963519 on Build GRCh38
rs145424901 Chr.10: 4963447-4963447 on Build GRCh38
rs781200594 Chr.10: 4963518-4963518 on Build GRCh38
rs200933984 Chr.10: 4963434-4963434 on Build GRCh38
rs372524463 Chr.10: 4963382-4963382 on Build GRCh38
rs769377816 Chr.10: 4963436-4963436 on Build GRCh38
rs368611374 Chr.10: 4963503-4963503 on Build GRCh38
rs534818681 Chr.10: 4963387-4963387 on Build GRCh38
rs753028508 Chr.10: 4963406-4963406 on Build GRCh38
rs546153006 Chr.10: 4963541-4963541 on Build GRCh38
rs570710332 Chr.10: 4963381-4963381 on Build GRCh38
rs138117679 Chr.10: 4963135-4963135 on Build GRCh38
rs779665083 Chr.10: 4963442-4963442 on Build GRCh38
rs755613712 Chr.10: 4963520-4963520 on Build GRCh38
rs955667379 Chr.10: 4963249-4963249 on Build GRCh38
rs1048441785 Chr.10: 4963122-4963122 on Build GRCh38

View Full Product Details