Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 7787690-7788223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5C1
Entrez Gene ID
509
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1
Gene Aliases
ATP5C, ATP5CL1
UniGene
Hs.271135
Gene Chromosome Location
Chr.10: 7788104-7807801 on Build GRCh38
Transcripts
NM_001001973.2 NM_001320886.1 NM_005174.3 XM_017016290.1 NM_012311.3 NR_045609.1 XM_006717434.3 XM_006717435.3 XM_011519427.1 XM_017015946.1

Gene Symbol
KIN
Entrez Gene ID
22944
Gene Name
Kin17 DNA and RNA binding protein
Gene Aliases
BTCD, KIN17, Rts2
UniGene
Hs.397918
Gene Chromosome Location
Chr.10: 7750962-7788027 on Build GRCh38
Transcripts
NM_001001973.2 NM_001320886.1 NM_005174.3 XM_017016290.1 NM_012311.3 NR_045609.1 XM_006717434.3 XM_006717435.3 XM_011519427.1 XM_017015946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1321191 c.10C>T p.R4C Chr.10: 7788217-7788217 on Build GRCh38
1171725 c.3G>T p.M1I Chr.10: 7788210-7788210 on Build GRCh38
1297448 c.87G>C p.Q29H Chr.10: 7787847-7787847 on Build GRCh38
6550684 c.112G>A p.E38K Chr.10: 7787822-7787822 on Build GRCh38
6452309 c.23C>T p.T8I Chr.10: 7787911-7787911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981768347 Chr.10: 7788154-7788154 on Build GRCh38
rs756899527 Chr.10: 7787956-7787956 on Build GRCh38
rs373340703 Chr.10: 7787892-7787892 on Build GRCh38
rs374803284 Chr.10: 7787779-7787779 on Build GRCh38
rs778937701 Chr.10: 7788026-7788026 on Build GRCh38
rs971305478 Chr.10: 7788147-7788147 on Build GRCh38
rs17553878 Chr.10: 7787796-7787796 on Build GRCh38
rs777414024 Chr.10: 7787955-7787955 on Build GRCh38
rs775703505 Chr.10: 7787922-7787922 on Build GRCh38
rs754863631 Chr.10: 7787886-7787886 on Build GRCh38
rs1052382535 Chr.10: 7788122-7788122 on Build GRCh38
rs1051905616 Chr.10: 7787942-7787942 on Build GRCh38
rs758064650 Chr.10: 7787905-7787905 on Build GRCh38
rs770083886 Chr.10: 7787915-7787915 on Build GRCh38
rs773416264 Chr.10: 7787965-7787965 on Build GRCh38
rs772673260 Chr.10: 7788184-7788184 on Build GRCh38
rs375300847 Chr.10: 7788170-7788170 on Build GRCh38
rs1048218393 Chr.10: 7788027-7788027 on Build GRCh38
rs900702629 Chr.10: 7787927-7787927 on Build GRCh38
rs563412420 Chr.10: 7787939-7787939 on Build GRCh38
rs182940447 Chr.10: 7787761-7787761 on Build GRCh38
rs1032748639 Chr.10: 7788102-7788102 on Build GRCh38
rs748247172 Chr.10: 7787963-7787963 on Build GRCh38
rs747032888 Chr.10: 7788189-7788189 on Build GRCh38
rs946144230 Chr.10: 7788153-7788153 on Build GRCh38
rs985194575 Chr.10: 7788137-7788160 on Build GRCh38
rs370743676 Chr.10: 7787959-7787959 on Build GRCh38
rs771034590 Chr.10: 7787968-7787968 on Build GRCh38
rs149288495 Chr.10: 7787868-7787868 on Build GRCh38
rs756954603 Chr.10: 7788169-7788169 on Build GRCh38
rs755234623 Chr.10: 7788181-7788181 on Build GRCh38
rs959943489 Chr.10: 7788029-7788029 on Build GRCh38
rs759844284 Chr.10: 7787816-7787816 on Build GRCh38
rs747899557 Chr.10: 7787780-7787780 on Build GRCh38
rs774292015 Chr.10: 7787851-7787851 on Build GRCh38
rs769015070 Chr.10: 7787931-7787931 on Build GRCh38
rs775260431 Chr.10: 7788198-7788198 on Build GRCh38
rs762135681 Chr.10: 7787857-7787857 on Build GRCh38
rs187556202 Chr.10: 7787997-7787997 on Build GRCh38
rs537096204 Chr.10: 7787728-7787728 on Build GRCh38
rs138658111 Chr.10: 7787823-7787823 on Build GRCh38
rs928067409 Chr.10: 7787753-7787753 on Build GRCh38
rs201428790 Chr.10: 7787945-7787945 on Build GRCh38
rs766426045 Chr.10: 7787883-7787883 on Build GRCh38
rs767826253 Chr.10: 7787861-7787861 on Build GRCh38
rs771677941 Chr.10: 7787785-7787785 on Build GRCh38
rs202096192 Chr.10: 7787936-7787936 on Build GRCh38
rs776649148 Chr.10: 7788192-7788192 on Build GRCh38
rs769532551 Chr.10: 7788194-7788194 on Build GRCh38
rs938058467 Chr.10: 7788000-7788000 on Build GRCh38
rs563909572 Chr.10: 7787992-7787992 on Build GRCh38
rs41299054 Chr.10: 7787760-7787760 on Build GRCh38
rs776503902 Chr.10: 7787973-7787973 on Build GRCh38
rs749590685 Chr.10: 7788034-7788034 on Build GRCh38
rs772223404 Chr.10: 7787964-7787964 on Build GRCh38
rs892914779 Chr.10: 7788121-7788121 on Build GRCh38
rs370479576 Chr.10: 7788180-7788180 on Build GRCh38
rs1001537080 Chr.10: 7788172-7788172 on Build GRCh38
rs187843802 Chr.10: 7788088-7788088 on Build GRCh38
rs761015073 Chr.10: 7787966-7787966 on Build GRCh38
rs75813059 Chr.10: 7787930-7787930 on Build GRCh38
rs148041572 Chr.10: 7787910-7787910 on Build GRCh38
rs558152756 Chr.10: 7787723-7787723 on Build GRCh38
rs949900836 Chr.10: 7787711-7787711 on Build GRCh38
rs386740619 Chr.10: 7787919-7787921 on Build GRCh38
rs752574022 Chr.10: 7788158-7788158 on Build GRCh38
rs891888833 Chr.10: 7787993-7787993 on Build GRCh38
rs758643048 Chr.10: 7788192-7788192 on Build GRCh38
rs1021607486 Chr.10: 7787732-7787732 on Build GRCh38
rs545812235 Chr.10: 7787947-7787947 on Build GRCh38
rs762954328 Chr.10: 7787832-7787832 on Build GRCh38
rs568305158 Chr.10: 7788005-7788005 on Build GRCh38
rs770157983 Chr.10: 7788001-7788001 on Build GRCh38
rs781362469 Chr.10: 7787900-7787900 on Build GRCh38
rs374009374 Chr.10: 7787726-7787739 on Build GRCh38
rs374796717 Chr.10: 7787961-7787961 on Build GRCh38
rs766340242 Chr.10: 7787944-7787944 on Build GRCh38
rs553398219 Chr.10: 7788193-7788193 on Build GRCh38
rs534248012 Chr.10: 7788019-7788019 on Build GRCh38
rs771382610 Chr.10: 7788093-7788093 on Build GRCh38
rs183110135 Chr.10: 7788070-7788070 on Build GRCh38
rs1040504905 Chr.10: 7788077-7788077 on Build GRCh38
rs995928000 Chr.10: 7787925-7787925 on Build GRCh38
rs1009902339 Chr.10: 7788126-7788126 on Build GRCh38
rs773504230 Chr.10: 7787937-7787937 on Build GRCh38
rs942361519 Chr.10: 7788099-7788099 on Build GRCh38
rs748282216 Chr.10: 7787988-7787988 on Build GRCh38
rs537135190 Chr.10: 7788057-7788057 on Build GRCh38
rs983817194 Chr.10: 7788091-7788091 on Build GRCh38
rs1045997213 Chr.10: 7787746-7787746 on Build GRCh38
rs750268820 Chr.10: 7787864-7787864 on Build GRCh38
rs904565633 Chr.10: 7788171-7788171 on Build GRCh38
rs757835767 Chr.10: 7787953-7787953 on Build GRCh38
rs142039200 Chr.10: 7787932-7787932 on Build GRCh38
rs759208217 Chr.10: 7787975-7787975 on Build GRCh38
rs752997709 Chr.10: 7788162-7788176 on Build GRCh38
rs930770735 Chr.10: 7788196-7788196 on Build GRCh38
rs115872841 Chr.10: 7788040-7788040 on Build GRCh38
rs1020403703 Chr.10: 7788138-7788138 on Build GRCh38
rs753753404 Chr.10: 7787884-7787884 on Build GRCh38
rs765224621 Chr.10: 7787983-7787983 on Build GRCh38
rs774684994 Chr.10: 7787962-7787962 on Build GRCh38
rs144539815 Chr.10: 7787906-7787906 on Build GRCh38
rs879188207 Chr.10: 7787744-7787744 on Build GRCh38
rs957037677 Chr.10: 7787940-7787940 on Build GRCh38
rs981291076 Chr.10: 7787994-7787994 on Build GRCh38
rs2070594 Chr.10: 7788101-7788101 on Build GRCh38
rs546581434 Chr.10: 7788002-7788002 on Build GRCh38
rs748725502 Chr.10: 7788182-7788182 on Build GRCh38
rs780509174 Chr.10: 7787960-7787960 on Build GRCh38
rs920568904 Chr.10: 7788004-7788004 on Build GRCh38
rs778045277 Chr.10: 7788186-7788186 on Build GRCh38
rs192877993 Chr.10: 7788035-7788035 on Build GRCh38
rs900216480 Chr.10: 7787919-7787919 on Build GRCh38
rs763660211 Chr.10: 7788162-7788162 on Build GRCh38
rs267602587 Chr.10: 7787881-7787881 on Build GRCh38
rs751040206 Chr.10: 7788167-7788167 on Build GRCh38
rs558660560 Chr.10: 7787750-7787750 on Build GRCh38
rs764293669 Chr.10: 7787837-7787837 on Build GRCh38
rs762863135 Chr.10: 7788159-7788159 on Build GRCh38

View Full Product Details