Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 11749375-11749774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHDC3
Entrez Gene ID
79746
Gene Name
enoyl-CoA hydratase domain containing 3
Gene Aliases
-
UniGene
Hs.22242
Gene Chromosome Location
Chr.10: 11742357-11764070 on Build GRCh38
Transcripts
NM_024693.4 XM_011519689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5836302 c.361C>T p.H121Y Chr.10: 11749563-11749563 on Build GRCh38
5836303 c.520C>T p.H174Y Chr.10: 11749563-11749563 on Build GRCh38
5221478 c.502G>C p.E168Q Chr.10: 11749545-11749545 on Build GRCh38
915129 c.300G>A p.G100G Chr.10: 11749502-11749502 on Build GRCh38
427104 c.343G>C p.E115Q Chr.10: 11749545-11749545 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs192975215 Chr.10: 11749569-11749569 on Build GRCh38
rs116651922 Chr.10: 11749402-11749402 on Build GRCh38
rs762223225 Chr.10: 11749527-11749527 on Build GRCh38
rs751554267 Chr.10: 11749563-11749563 on Build GRCh38
rs371504407 Chr.10: 11749479-11749479 on Build GRCh38
rs758603382 Chr.10: 11749641-11749641 on Build GRCh38
rs537057464 Chr.10: 11749610-11749610 on Build GRCh38
rs192357074 Chr.10: 11749732-11749732 on Build GRCh38
rs760317056 Chr.10: 11749747-11749754 on Build GRCh38
rs141899048 Chr.10: 11749555-11749555 on Build GRCh38
rs534960821 Chr.10: 11749534-11749534 on Build GRCh38
rs1017403562 Chr.10: 11749728-11749728 on Build GRCh38
rs369047701 Chr.10: 11749584-11749584 on Build GRCh38
rs762666062 Chr.10: 11749598-11749598 on Build GRCh38
rs770766925 Chr.10: 11749568-11749568 on Build GRCh38
rs550522069 Chr.10: 11749413-11749413 on Build GRCh38
rs764726715 Chr.10: 11749522-11749522 on Build GRCh38
rs374358090 Chr.10: 11749745-11749746 on Build GRCh38
rs982260266 Chr.10: 11749616-11749616 on Build GRCh38
rs114256803 Chr.10: 11749619-11749619 on Build GRCh38
rs769596594 Chr.10: 11749582-11749582 on Build GRCh38
rs541973485 Chr.10: 11749733-11749733 on Build GRCh38
rs868444461 Chr.10: 11749472-11749472 on Build GRCh38
rs1027188617 Chr.10: 11749603-11749603 on Build GRCh38
rs978434355 Chr.10: 11749558-11749558 on Build GRCh38
rs752964581 Chr.10: 11749636-11749636 on Build GRCh38
rs756181405 Chr.10: 11749454-11749454 on Build GRCh38
rs753973368 Chr.10: 11749615-11749615 on Build GRCh38
rs768100155 Chr.10: 11749487-11749488 on Build GRCh38
rs1037684226 Chr.10: 11749627-11749627 on Build GRCh38
rs771771524 Chr.10: 11749745-11749752 on Build GRCh38
rs139837566 Chr.10: 11749420-11749420 on Build GRCh38
rs766730448 Chr.10: 11749614-11749614 on Build GRCh38
rs182993857 Chr.10: 11749656-11749656 on Build GRCh38
rs993351089 Chr.10: 11749456-11749456 on Build GRCh38
rs113825360 Chr.10: 11749674-11749674 on Build GRCh38
rs1009150495 Chr.10: 11749675-11749675 on Build GRCh38
rs201207580 Chr.10: 11749524-11749524 on Build GRCh38
rs745337827 Chr.10: 11749744-11749751 on Build GRCh38
rs376686816 Chr.10: 11749604-11749604 on Build GRCh38
rs1017768925 Chr.10: 11749513-11749513 on Build GRCh38
rs761617189 Chr.10: 11749528-11749529 on Build GRCh38
rs773700585 Chr.10: 11749605-11749605 on Build GRCh38
rs150630386 Chr.10: 11749622-11749622 on Build GRCh38
rs750556983 Chr.10: 11749453-11749453 on Build GRCh38
rs187672109 Chr.10: 11749711-11749711 on Build GRCh38
rs768197577 Chr.10: 11749748-11749755 on Build GRCh38
rs759879970 Chr.10: 11749575-11749575 on Build GRCh38
rs775989600 Chr.10: 11749520-11749520 on Build GRCh38
rs914106839 Chr.10: 11749611-11749611 on Build GRCh38
rs374956993 Chr.10: 11749565-11749565 on Build GRCh38
rs765540589 Chr.10: 11749587-11749588 on Build GRCh38
rs766540789 Chr.10: 11749540-11749540 on Build GRCh38
rs565727119 Chr.10: 11749533-11749533 on Build GRCh38
rs1040956766 Chr.10: 11749574-11749574 on Build GRCh38
rs747879768 Chr.10: 11749492-11749492 on Build GRCh38
rs533244535 Chr.10: 11749463-11749463 on Build GRCh38
rs943821762 Chr.10: 11749738-11749738 on Build GRCh38
rs757329801 Chr.10: 11749662-11749662 on Build GRCh38
rs369976445 Chr.10: 11749612-11749612 on Build GRCh38
rs774354221 Chr.10: 11749493-11749493 on Build GRCh38
rs768082307 Chr.10: 11749448-11749448 on Build GRCh38
rs746889195 Chr.10: 11749503-11749503 on Build GRCh38
rs770520205 Chr.10: 11749517-11749517 on Build GRCh38
rs779057310 Chr.10: 11749561-11749561 on Build GRCh38
rs926060007 Chr.10: 11749714-11749714 on Build GRCh38
rs913752268 Chr.10: 11749730-11749730 on Build GRCh38
rs772862832 Chr.10: 11749555-11749556 on Build GRCh38
rs146508606 Chr.10: 11749499-11749499 on Build GRCh38
rs760676507 Chr.10: 11749657-11749657 on Build GRCh38
rs747968074 Chr.10: 11749564-11749564 on Build GRCh38
rs745310969 Chr.10: 11749571-11749571 on Build GRCh38
rs202143598 Chr.10: 11749592-11749592 on Build GRCh38
rs552383167 Chr.10: 11749501-11749501 on Build GRCh38
rs926057492 Chr.10: 11749551-11749551 on Build GRCh38
rs749575731 Chr.10: 11749467-11749467 on Build GRCh38
rs562037117 Chr.10: 11749436-11749436 on Build GRCh38
rs777012488 Chr.10: 11749749-11749756 on Build GRCh38
rs775264655 Chr.10: 11749746-11749753 on Build GRCh38
rs755142554 Chr.10: 11749469-11749469 on Build GRCh38
rs768172158 Chr.10: 11749529-11749529 on Build GRCh38
rs141101500 Chr.10: 11749554-11749554 on Build GRCh38
rs934744205 Chr.10: 11749719-11749722 on Build GRCh38
rs759295596 Chr.10: 11749521-11749521 on Build GRCh38
rs58243400 Chr.10: 11749743-11749750 on Build GRCh38

View Full Product Details