Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12210076-12210592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC123
Entrez Gene ID
8872
Gene Name
cell division cycle 123
Gene Aliases
C10orf7, D123
UniGene
Hs.412842
Gene Chromosome Location
Chr.10: 12195962-12250590 on Build GRCh38
Transcripts
NM_006023.2 XM_005252638.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7175756 c.237G>A p.T79T Chr.10: 12210322-12210322 on Build GRCh38
6319978 c.80G>T p.W27L Chr.10: 12210291-12210291 on Build GRCh38
6319977 c.206G>T p.W69L Chr.10: 12210291-12210291 on Build GRCh38
7175757 c.111G>A p.T37T Chr.10: 12210322-12210322 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779537723 Chr.10: 12210328-12210328 on Build GRCh38
rs548744096 Chr.10: 12210266-12210266 on Build GRCh38
rs150151825 Chr.10: 12210515-12210515 on Build GRCh38
rs1025682071 Chr.10: 12210164-12210164 on Build GRCh38
rs748060572 Chr.10: 12210284-12210284 on Build GRCh38
rs972544157 Chr.10: 12210139-12210139 on Build GRCh38
rs769115291 Chr.10: 12210240-12210240 on Build GRCh38
rs771121636 Chr.10: 12210434-12210434 on Build GRCh38
rs776941616 Chr.10: 12210247-12210250 on Build GRCh38
rs758661336 Chr.10: 12210298-12210298 on Build GRCh38
rs199669441 Chr.10: 12210316-12210316 on Build GRCh38
rs1045462684 Chr.10: 12210191-12210191 on Build GRCh38
rs777759788 Chr.10: 12210304-12210304 on Build GRCh38
rs768468282 Chr.10: 12210401-12210401 on Build GRCh38
rs1012873509 Chr.10: 12210156-12210156 on Build GRCh38
rs182136185 Chr.10: 12210135-12210135 on Build GRCh38
rs762578600 Chr.10: 12210251-12210251 on Build GRCh38
rs552916590 Chr.10: 12210531-12210531 on Build GRCh38
rs770011895 Chr.10: 12210266-12210267 on Build GRCh38
rs935509051 Chr.10: 12210293-12210293 on Build GRCh38
rs745732754 Chr.10: 12210326-12210326 on Build GRCh38
rs201679420 Chr.10: 12210344-12210344 on Build GRCh38
rs754260354 Chr.10: 12210274-12210274 on Build GRCh38
rs748983765 Chr.10: 12210329-12210329 on Build GRCh38
rs1031063002 Chr.10: 12210221-12210221 on Build GRCh38
rs1001437347 Chr.10: 12210108-12210108 on Build GRCh38
rs771134969 Chr.10: 12210349-12210349 on Build GRCh38
rs759947064 Chr.10: 12210266-12210266 on Build GRCh38
rs138912400 Chr.10: 12210321-12210321 on Build GRCh38
rs781616054 Chr.10: 12210447-12210447 on Build GRCh38
rs908307653 Chr.10: 12210457-12210457 on Build GRCh38
rs534549936 Chr.10: 12210546-12210546 on Build GRCh38
rs2271804 Chr.10: 12210218-12210218 on Build GRCh38
rs758301266 Chr.10: 12210322-12210322 on Build GRCh38
rs576857381 Chr.10: 12210318-12210318 on Build GRCh38
rs140989496 Chr.10: 12210381-12210381 on Build GRCh38
rs768279741 Chr.10: 12210330-12210330 on Build GRCh38
rs765352004 Chr.10: 12210283-12210283 on Build GRCh38
rs766935539 Chr.10: 12210271-12210271 on Build GRCh38
rs922846550 Chr.10: 12210282-12210282 on Build GRCh38
rs1001617090 Chr.10: 12210405-12210405 on Build GRCh38
rs1036032840 Chr.10: 12210121-12210121 on Build GRCh38
rs754803978 Chr.10: 12210180-12210180 on Build GRCh38
rs759667495 Chr.10: 12210275-12210275 on Build GRCh38
rs572314414 Chr.10: 12210545-12210545 on Build GRCh38
rs145544440 Chr.10: 12210552-12210552 on Build GRCh38
rs371297474 Chr.10: 12210345-12210345 on Build GRCh38
rs956840827 Chr.10: 12210274-12210274 on Build GRCh38
rs570470197 Chr.10: 12210403-12210403 on Build GRCh38
rs775133872 Chr.10: 12210250-12210250 on Build GRCh38
rs376222610 Chr.10: 12210212-12210212 on Build GRCh38
rs1042848622 Chr.10: 12210380-12210380 on Build GRCh38
rs952833118 Chr.10: 12210179-12210179 on Build GRCh38
rs115088873 Chr.10: 12210551-12210551 on Build GRCh38
rs11257597 Chr.10: 12210464-12210464 on Build GRCh38
rs533143813 Chr.10: 12210445-12210445 on Build GRCh38
rs775303432 Chr.10: 12210362-12210363 on Build GRCh38
rs186070657 Chr.10: 12210261-12210261 on Build GRCh38
rs759949890 Chr.10: 12210362-12210362 on Build GRCh38

View Full Product Details