Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12238186-12238586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC123
Entrez Gene ID
8872
Gene Name
cell division cycle 123
Gene Aliases
C10orf7, D123
UniGene
Hs.412842
Gene Chromosome Location
Chr.10: 12195962-12250590 on Build GRCh38
Transcripts
NM_006023.2 XM_005252638.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012145 c.569T>G p.F190C Chr.10: 12238463-12238463 on Build GRCh38
4670592 c.697G>A p.D233N Chr.10: 12238465-12238465 on Build GRCh38
4670593 c.571G>A p.D191N Chr.10: 12238465-12238465 on Build GRCh38
3806591 c.570C>T p.F190F Chr.10: 12238464-12238464 on Build GRCh38
181020 c.696C>T p.F232F Chr.10: 12238464-12238464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114952366 Chr.10: 12238520-12238520 on Build GRCh38
rs373454189 Chr.10: 12238469-12238469 on Build GRCh38
rs924999070 Chr.10: 12238332-12238332 on Build GRCh38
rs190776856 Chr.10: 12238430-12238430 on Build GRCh38
rs747603830 Chr.10: 12238518-12238518 on Build GRCh38
rs937794993 Chr.10: 12238341-12238341 on Build GRCh38
rs917598530 Chr.10: 12238408-12238408 on Build GRCh38
rs376153867 Chr.10: 12238506-12238506 on Build GRCh38
rs989494677 Chr.10: 12238243-12238243 on Build GRCh38
rs775081005 Chr.10: 12238414-12238414 on Build GRCh38
rs755554198 Chr.10: 12238479-12238479 on Build GRCh38
rs746308913 Chr.10: 12238518-12238522 on Build GRCh38
rs972641886 Chr.10: 12238560-12238560 on Build GRCh38
rs56121235 Chr.10: 12238307-12238307 on Build GRCh38
rs377489031 Chr.10: 12238438-12238438 on Build GRCh38
rs200584206 Chr.10: 12238530-12238530 on Build GRCh38
rs531422619 Chr.10: 12238296-12238296 on Build GRCh38
rs186014836 Chr.10: 12238330-12238330 on Build GRCh38
rs758268340 Chr.10: 12238448-12238448 on Build GRCh38
rs1020546051 Chr.10: 12238275-12238275 on Build GRCh38
rs773828455 Chr.10: 12238426-12238426 on Build GRCh38
rs10906104 Chr.10: 12238297-12238297 on Build GRCh38
rs751551758 Chr.10: 12238466-12238466 on Build GRCh38
rs763790124 Chr.10: 12238452-12238452 on Build GRCh38
rs748882084 Chr.10: 12238485-12238485 on Build GRCh38
rs745412087 Chr.10: 12238474-12238474 on Build GRCh38
rs765390874 Chr.10: 12238535-12238535 on Build GRCh38
rs963540779 Chr.10: 12238542-12238542 on Build GRCh38
rs779695804 Chr.10: 12238482-12238482 on Build GRCh38
rs369720934 Chr.10: 12238458-12238458 on Build GRCh38
rs748515751 Chr.10: 12238419-12238419 on Build GRCh38
rs886378710 Chr.10: 12238464-12238464 on Build GRCh38
rs562394838 Chr.10: 12238278-12238278 on Build GRCh38
rs949319122 Chr.10: 12238422-12238422 on Build GRCh38
rs777240608 Chr.10: 12238525-12238525 on Build GRCh38
rs772555362 Chr.10: 12238504-12238504 on Build GRCh38
rs781059221 Chr.10: 12238470-12238470 on Build GRCh38
rs373772154 Chr.10: 12238563-12238563 on Build GRCh38
rs776833544 Chr.10: 12238433-12238433 on Build GRCh38
rs766520724 Chr.10: 12238431-12238431 on Build GRCh38
rs997489109 Chr.10: 12238468-12238468 on Build GRCh38
rs752903797 Chr.10: 12238440-12238440 on Build GRCh38
rs954963173 Chr.10: 12238214-12238214 on Build GRCh38
rs144785906 Chr.10: 12238347-12238347 on Build GRCh38
rs772269039 Chr.10: 12238425-12238425 on Build GRCh38

View Full Product Details