Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13656529-13657019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4A
Entrez Gene ID
55691
Gene Name
FERM domain containing 4A
Gene Aliases
CCAFCA, FRMD4, bA295P9.4
UniGene
Hs.330463
Gene Chromosome Location
Chr.10: 13643706-14330952 on Build GRCh38
Transcripts
NM_001318336.1 NM_001318337.1 NM_001318338.1 NM_018027.4 XM_005252489.2 XM_005252490.2 XM_005252495.4 XM_006717457.2 XM_006717458.3 XM_006717459.2 XM_006717460.2 XM_006717464.1 XM_011519539.1 XM_011519540.1 XM_011519541.1 XM_011519543.2 XM_017016393.1 XM_017016394.1 XM_017016395.1 XM_017016396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5507160 c.2695T>G p.S899A Chr.10: 13656894-13656894 on Build GRCh38
4685623 c.2573A>G p.E858G Chr.10: 13657016-13657016 on Build GRCh38
2024830 c.2692C>T p.P898S Chr.10: 13656897-13656897 on Build GRCh38
2024828 c.2921C>T p.T974I Chr.10: 13656668-13656668 on Build GRCh38
6666917 c.2837C>T p.S946L Chr.10: 13656752-13656752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750343072 Chr.10: 13656789-13656789 on Build GRCh38
rs755872986 Chr.10: 13656804-13656804 on Build GRCh38
rs1032427933 Chr.10: 13656962-13656962 on Build GRCh38
rs780371595 Chr.10: 13656977-13656977 on Build GRCh38
rs370267348 Chr.10: 13656756-13656756 on Build GRCh38
rs370459275 Chr.10: 13656703-13656703 on Build GRCh38
rs754169439 Chr.10: 13656660-13656660 on Build GRCh38
rs558385796 Chr.10: 13656584-13656584 on Build GRCh38
rs1033996442 Chr.10: 13656759-13656759 on Build GRCh38
rs775699436 Chr.10: 13656838-13656838 on Build GRCh38
rs146301035 Chr.10: 13656965-13656965 on Build GRCh38
rs779700778 Chr.10: 13656695-13656695 on Build GRCh38
rs769354778 Chr.10: 13656983-13656983 on Build GRCh38
rs777524534 Chr.10: 13656968-13656968 on Build GRCh38
rs756818243 Chr.10: 13656976-13656976 on Build GRCh38
rs774649049 Chr.10: 13656924-13656924 on Build GRCh38
rs759847969 Chr.10: 13656667-13656667 on Build GRCh38
rs966761633 Chr.10: 13656811-13656811 on Build GRCh38
rs756239572 Chr.10: 13656766-13656768 on Build GRCh38
rs932862761 Chr.10: 13656578-13656578 on Build GRCh38
rs1048783346 Chr.10: 13656914-13656914 on Build GRCh38
rs369869090 Chr.10: 13656711-13656711 on Build GRCh38
rs753168615 Chr.10: 13656729-13656729 on Build GRCh38
rs769499770 Chr.10: 13656694-13656694 on Build GRCh38
rs567836189 Chr.10: 13656891-13656891 on Build GRCh38
rs764374574 Chr.10: 13656848-13656848 on Build GRCh38
rs1012681461 Chr.10: 13656677-13656677 on Build GRCh38
rs753536105 Chr.10: 13656886-13656886 on Build GRCh38
rs1025184072 Chr.10: 13656700-13656700 on Build GRCh38
rs142082242 Chr.10: 13656836-13656836 on Build GRCh38
rs539819758 Chr.10: 13656610-13656610 on Build GRCh38
rs377346629 Chr.10: 13656889-13656889 on Build GRCh38
rs540835461 Chr.10: 13656607-13656607 on Build GRCh38
rs775039060 Chr.10: 13656624-13656624 on Build GRCh38
rs772283361 Chr.10: 13656924-13656929 on Build GRCh38
rs749818810 Chr.10: 13656982-13656982 on Build GRCh38
rs200846869 Chr.10: 13656887-13656887 on Build GRCh38
rs143274194 Chr.10: 13656719-13656719 on Build GRCh38
rs538597314 Chr.10: 13656568-13656568 on Build GRCh38
rs761352751 Chr.10: 13656714-13656714 on Build GRCh38
rs745639287 Chr.10: 13656690-13656690 on Build GRCh38
rs761928430 Chr.10: 13656852-13656852 on Build GRCh38
rs918939672 Chr.10: 13656912-13656912 on Build GRCh38
rs749166490 Chr.10: 13656820-13656820 on Build GRCh38
rs773642767 Chr.10: 13656656-13656656 on Build GRCh38
rs536647518 Chr.10: 13656923-13656924 on Build GRCh38
rs757243136 Chr.10: 13656688-13656688 on Build GRCh38
rs754697615 Chr.10: 13656952-13656952 on Build GRCh38
rs780095617 Chr.10: 13656876-13656902 on Build GRCh38
rs374074686 Chr.10: 13656710-13656710 on Build GRCh38
rs960695641 Chr.10: 13656765-13656765 on Build GRCh38
rs199847871 Chr.10: 13656788-13656788 on Build GRCh38
rs761166826 Chr.10: 13656658-13656658 on Build GRCh38
rs375022907 Chr.10: 13656937-13656937 on Build GRCh38
rs987552976 Chr.10: 13656791-13656791 on Build GRCh38
rs746575585 Chr.10: 13656898-13656898 on Build GRCh38
rs746051234 Chr.10: 13656629-13656630 on Build GRCh38
rs1032291954 Chr.10: 13656640-13656640 on Build GRCh38
rs747606415 Chr.10: 13656832-13656832 on Build GRCh38
rs983630296 Chr.10: 13656752-13656752 on Build GRCh38
rs201700015 Chr.10: 13656621-13656621 on Build GRCh38
rs368369588 Chr.10: 13656938-13656938 on Build GRCh38
rs754593288 Chr.10: 13656822-13656822 on Build GRCh38
rs746943117 Chr.10: 13656613-13656613 on Build GRCh38
rs750583045 Chr.10: 13656868-13656868 on Build GRCh38
rs913972349 Chr.10: 13656563-13656563 on Build GRCh38
rs750995961 Chr.10: 13656970-13656970 on Build GRCh38
rs748566561 Chr.10: 13656924-13656926 on Build GRCh38
rs527899634 Chr.10: 13656823-13656823 on Build GRCh38
rs774784218 Chr.10: 13656924-13656932 on Build GRCh38
rs758111344 Chr.10: 13656895-13656895 on Build GRCh38
rs543127114 Chr.10: 13656626-13656626 on Build GRCh38
rs779709485 Chr.10: 13656819-13656819 on Build GRCh38
rs973175969 Chr.10: 13656780-13656780 on Build GRCh38
rs774739073 Chr.10: 13656850-13656850 on Build GRCh38
rs931871427 Chr.10: 13656580-13656580 on Build GRCh38
rs755993250 Chr.10: 13656869-13656869 on Build GRCh38
rs370587290 Chr.10: 13656612-13656612 on Build GRCh38
rs531733605 Chr.10: 13656672-13656672 on Build GRCh38
rs890373618 Chr.10: 13656588-13656588 on Build GRCh38
rs769159019 Chr.10: 13656917-13656917 on Build GRCh38
rs534805903 Chr.10: 13656566-13656566 on Build GRCh38
rs573614192 Chr.10: 13656775-13656775 on Build GRCh38
rs547644775 Chr.10: 13656866-13656866 on Build GRCh38
rs751751390 Chr.10: 13656684-13656684 on Build GRCh38
rs765008817 Chr.10: 13656960-13656960 on Build GRCh38
rs777992765 Chr.10: 13656683-13656683 on Build GRCh38
rs929027969 Chr.10: 13656913-13656913 on Build GRCh38
rs148321299 Chr.10: 13656781-13656781 on Build GRCh38
rs771055887 Chr.10: 13656617-13656617 on Build GRCh38
rs765521655 Chr.10: 13656727-13656727 on Build GRCh38
rs759883779 Chr.10: 13656726-13656726 on Build GRCh38
rs773144615 Chr.10: 13656934-13656934 on Build GRCh38
rs777126735 Chr.10: 13656834-13656834 on Build GRCh38
rs749624928 Chr.10: 13656916-13656916 on Build GRCh38
rs574078315 Chr.10: 13656623-13656623 on Build GRCh38
rs202106361 Chr.10: 13656964-13656964 on Build GRCh38
rs746351922 Chr.10: 13656835-13656835 on Build GRCh38
rs180995308 Chr.10: 13656604-13656604 on Build GRCh38
rs763418844 Chr.10: 13656842-13656842 on Build GRCh38
rs370425803 Chr.10: 13656906-13656906 on Build GRCh38
rs562910285 Chr.10: 13656641-13656641 on Build GRCh38
rs370765891 Chr.10: 13656928-13656928 on Build GRCh38
rs777129980 Chr.10: 13656896-13656896 on Build GRCh38
rs762477064 Chr.10: 13656625-13656625 on Build GRCh38
rs763069203 Chr.10: 13656754-13656754 on Build GRCh38
rs748898441 Chr.10: 13656697-13656697 on Build GRCh38
rs765447123 Chr.10: 13656668-13656668 on Build GRCh38
rs368649469 Chr.10: 13656927-13656927 on Build GRCh38
rs921618294 Chr.10: 13656570-13656570 on Build GRCh38
rs191808513 Chr.10: 13656565-13656565 on Build GRCh38
rs529793716 Chr.10: 13656899-13656899 on Build GRCh38

View Full Product Details