Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14521677-14522191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM107B
Entrez Gene ID
83641
Gene Name
family with sequence similarity 107 member B
Gene Aliases
C10orf45
UniGene
Hs.446315
Gene Chromosome Location
Chr.10: 14518557-14774897 on Build GRCh38
Transcripts
NM_001282695.1 NM_001282696.1 NM_001282697.1 NM_001282698.1 NM_001282699.1 NM_001282700.1 NM_001282701.1 NM_001282702.1 NM_001282703.1 NM_001320735.1 NM_001320736.1 NM_001320737.1 NM_001320738.1 NM_001320739.1 NM_001320740.1 NM_001320741.1 NM_031453.3 XM_005252616.3 XM_011519735.1 XM_017016746.1 XM_017016747.1 XM_017016748.1 XM_017016749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014893 c.667C>T p.Q223* Chr.10: 14522006-14522006 on Build GRCh38
213947 c.784C>T p.R262W Chr.10: 14521889-14521889 on Build GRCh38
916589 c.756A>C p.K252N Chr.10: 14521917-14521917 on Build GRCh38
51592 c.710G>A p.R237Q Chr.10: 14521963-14521963 on Build GRCh38
916590 c.704G>T p.R235I Chr.10: 14521969-14521969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566258803 Chr.10: 14522110-14522110 on Build GRCh38
rs781471619 Chr.10: 14521825-14521825 on Build GRCh38
rs374430922 Chr.10: 14521855-14521855 on Build GRCh38
rs547748811 Chr.10: 14521743-14521743 on Build GRCh38
rs760370962 Chr.10: 14521868-14521868 on Build GRCh38
rs1032708005 Chr.10: 14522094-14522094 on Build GRCh38
rs970012398 Chr.10: 14522140-14522140 on Build GRCh38
rs150257845 Chr.10: 14522051-14522051 on Build GRCh38
rs928063099 Chr.10: 14521963-14521963 on Build GRCh38
rs756670200 Chr.10: 14522045-14522045 on Build GRCh38
rs536741710 Chr.10: 14521769-14521769 on Build GRCh38
rs957890732 Chr.10: 14522044-14522044 on Build GRCh38
rs751009216 Chr.10: 14521915-14521915 on Build GRCh38
rs576305397 Chr.10: 14521872-14521872 on Build GRCh38
rs199845848 Chr.10: 14521968-14521968 on Build GRCh38
rs770252379 Chr.10: 14521827-14521827 on Build GRCh38
rs749569396 Chr.10: 14521845-14521845 on Build GRCh38
rs958219373 Chr.10: 14521766-14521766 on Build GRCh38
rs772333412 Chr.10: 14521966-14521966 on Build GRCh38
rs199831281 Chr.10: 14521858-14521858 on Build GRCh38
rs748149906 Chr.10: 14521824-14521828 on Build GRCh38
rs780960343 Chr.10: 14521932-14521932 on Build GRCh38
rs567689227 Chr.10: 14521764-14521764 on Build GRCh38
rs776506034 Chr.10: 14522001-14522001 on Build GRCh38
rs779573547 Chr.10: 14522025-14522025 on Build GRCh38
rs752404794 Chr.10: 14522024-14522024 on Build GRCh38
rs769028004 Chr.10: 14521938-14521938 on Build GRCh38
rs989400801 Chr.10: 14521796-14521796 on Build GRCh38
rs781537708 Chr.10: 14521896-14521896 on Build GRCh38
rs958290982 Chr.10: 14521811-14521811 on Build GRCh38
rs1032655530 Chr.10: 14521802-14521802 on Build GRCh38
rs1056459614 Chr.10: 14522141-14522141 on Build GRCh38
rs757704900 Chr.10: 14521889-14521889 on Build GRCh38
rs1040197453 Chr.10: 14522066-14522066 on Build GRCh38
rs527966603 Chr.10: 14522157-14522157 on Build GRCh38
rs149948586 Chr.10: 14521887-14521887 on Build GRCh38
rs970558046 Chr.10: 14521742-14521742 on Build GRCh38
rs1050697954 Chr.10: 14521740-14521740 on Build GRCh38
rs888669823 Chr.10: 14521711-14521711 on Build GRCh38
rs773017959 Chr.10: 14521972-14521972 on Build GRCh38
rs756406965 Chr.10: 14521923-14521923 on Build GRCh38
rs989736033 Chr.10: 14521846-14521846 on Build GRCh38
rs1045785407 Chr.10: 14522162-14522162 on Build GRCh38
rs1046785885 Chr.10: 14522107-14522107 on Build GRCh38
rs892786127 Chr.10: 14522097-14522097 on Build GRCh38
rs772997958 Chr.10: 14521859-14521859 on Build GRCh38
rs780139442 Chr.10: 14521829-14521829 on Build GRCh38
rs998924519 Chr.10: 14522010-14522010 on Build GRCh38
rs772001031 Chr.10: 14521835-14521836 on Build GRCh38
rs770607673 Chr.10: 14521992-14521992 on Build GRCh38
rs993354971 Chr.10: 14521739-14521739 on Build GRCh38
rs189436902 Chr.10: 14522068-14522068 on Build GRCh38
rs779261052 Chr.10: 14522054-14522054 on Build GRCh38
rs73599366 Chr.10: 14522104-14522104 on Build GRCh38
rs1056042101 Chr.10: 14521931-14521933 on Build GRCh38
rs139987994 Chr.10: 14521961-14521961 on Build GRCh38
rs923875333 Chr.10: 14521760-14521760 on Build GRCh38
rs181075242 Chr.10: 14522069-14522069 on Build GRCh38
rs757141732 Chr.10: 14521903-14521903 on Build GRCh38
rs763702998 Chr.10: 14522034-14522034 on Build GRCh38
rs188805715 Chr.10: 14522138-14522138 on Build GRCh38
rs551669262 Chr.10: 14521714-14521714 on Build GRCh38
rs910074462 Chr.10: 14522127-14522127 on Build GRCh38
rs780971394 Chr.10: 14521774-14521775 on Build GRCh38
rs185220044 Chr.10: 14522076-14522076 on Build GRCh38
rs185602242 Chr.10: 14521735-14521735 on Build GRCh38
rs902884357 Chr.10: 14521772-14521772 on Build GRCh38
rs759124546 Chr.10: 14522023-14522023 on Build GRCh38
rs12254425 Chr.10: 14521715-14521715 on Build GRCh38
rs370182735 Chr.10: 14521826-14521826 on Build GRCh38
rs1035125097 Chr.10: 14522164-14522164 on Build GRCh38
rs531803911 Chr.10: 14522090-14522090 on Build GRCh38
rs758377692 Chr.10: 14522067-14522067 on Build GRCh38
rs752038569 Chr.10: 14521821-14521821 on Build GRCh38
rs200996851 Chr.10: 14521933-14521933 on Build GRCh38
rs377367079 Chr.10: 14522058-14522058 on Build GRCh38
rs998530118 Chr.10: 14521783-14521783 on Build GRCh38
rs773072184 Chr.10: 14521855-14521855 on Build GRCh38
rs769640912 Chr.10: 14521987-14521987 on Build GRCh38
rs894698637 Chr.10: 14522150-14522150 on Build GRCh38
rs779066984 Chr.10: 14521951-14521951 on Build GRCh38
rs958719080 Chr.10: 14521738-14521738 on Build GRCh38
rs1024409028 Chr.10: 14521988-14521988 on Build GRCh38
rs146524497 Chr.10: 14521893-14521893 on Build GRCh38
rs977893304 Chr.10: 14521749-14521749 on Build GRCh38
rs759233265 Chr.10: 14521878-14521878 on Build GRCh38
rs768837622 Chr.10: 14521847-14521847 on Build GRCh38
rs910913871 Chr.10: 14521801-14521801 on Build GRCh38
rs764676351 Chr.10: 14521882-14521882 on Build GRCh38
rs774379687 Chr.10: 14521728-14521728 on Build GRCh38
rs753401059 Chr.10: 14521873-14521873 on Build GRCh38
rs1024358108 Chr.10: 14521741-14521741 on Build GRCh38
rs940736953 Chr.10: 14521971-14521971 on Build GRCh38
rs540697893 Chr.10: 14522035-14522035 on Build GRCh38
rs1034786231 Chr.10: 14521724-14521724 on Build GRCh38
rs200211953 Chr.10: 14522050-14522050 on Build GRCh38
rs779291859 Chr.10: 14521745-14521745 on Build GRCh38
rs920565630 Chr.10: 14522002-14522002 on Build GRCh38
rs745864568 Chr.10: 14521974-14521974 on Build GRCh38

View Full Product Details