Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14903894-14904432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLRE1C
Entrez Gene ID
64421
Gene Name
DNA cross-link repair 1C
Gene Aliases
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
UniGene
Hs.655932
Gene Chromosome Location
Chr.10: 14897853-14954432 on Build GRCh38
Transcripts
XM_017016556.1 XM_017016559.1 XM_017016560.1 NM_001193424.1 NM_001193425.1 NM_001193426.1 NM_001193427.1 NM_024670.3 NR_034181.1 XM_011519662.2 XM_017016637.1

Gene Symbol
SUV39H2
Entrez Gene ID
79723
Gene Name
suppressor of variegation 3-9 homolog 2
Gene Aliases
KMT1B
UniGene
Hs.554883
Gene Chromosome Location
Chr.10: 14878783-14904315 on Build GRCh38
Transcripts
XM_017016556.1 XM_017016559.1 XM_017016560.1 NM_001193424.1 NM_001193425.1 NM_001193426.1 NM_001193427.1 NM_024670.3 NR_034181.1 XM_011519662.2 XM_017016637.1

SNP Detail

SNP ID Chromosome Location
rs1038168792 Chr.10: 14904369-14904369 on Build GRCh38
rs1003523490 Chr.10: 14904137-14904137 on Build GRCh38
rs1038538542 Chr.10: 14904067-14904067 on Build GRCh38
rs563622953 Chr.10: 14904058-14904058 on Build GRCh38
rs567373503 Chr.10: 14904213-14904213 on Build GRCh38
rs1023228529 Chr.10: 14903932-14903932 on Build GRCh38
rs1050973473 Chr.10: 14904055-14904055 on Build GRCh38
rs11259385 Chr.10: 14904138-14904138 on Build GRCh38
rs867899351 Chr.10: 14904295-14904295 on Build GRCh38
rs1044111336 Chr.10: 14904383-14904383 on Build GRCh38
rs35012000 Chr.10: 14904316-14904316 on Build GRCh38
rs534127559 Chr.10: 14904353-14904353 on Build GRCh38
rs898285535 Chr.10: 14904377-14904377 on Build GRCh38
rs183557434 Chr.10: 14904256-14904256 on Build GRCh38
rs548281322 Chr.10: 14904354-14904354 on Build GRCh38
rs571793715 Chr.10: 14904263-14904263 on Build GRCh38
rs191603474 Chr.10: 14904236-14904236 on Build GRCh38
rs542619141 Chr.10: 14903991-14903991 on Build GRCh38
rs575831121 Chr.10: 14904187-14904187 on Build GRCh38
rs35999949 Chr.10: 14904342-14904342 on Build GRCh38
rs1012556381 Chr.10: 14903927-14903927 on Build GRCh38
rs956177334 Chr.10: 14903974-14903974 on Build GRCh38
rs188840170 Chr.10: 14904385-14904385 on Build GRCh38
rs894205169 Chr.10: 14904317-14904317 on Build GRCh38
rs531526476 Chr.10: 14904036-14904036 on Build GRCh38
rs906849036 Chr.10: 14904397-14904397 on Build GRCh38
rs908429828 Chr.10: 14904262-14904262 on Build GRCh38
rs879884816 Chr.10: 14903975-14903975 on Build GRCh38
rs186353154 Chr.10: 14904218-14904218 on Build GRCh38
rs1012589844 Chr.10: 14904087-14904087 on Build GRCh38
rs371491790 Chr.10: 14904237-14904237 on Build GRCh38
rs150828159 Chr.10: 14903949-14903949 on Build GRCh38
rs527373001 Chr.10: 14904162-14904162 on Build GRCh38
rs772591940 Chr.10: 14904151-14904151 on Build GRCh38
rs79193913 Chr.10: 14904304-14904304 on Build GRCh38
rs1028588221 Chr.10: 14904362-14904372 on Build GRCh38
rs943382161 Chr.10: 14904029-14904029 on Build GRCh38
rs562405248 Chr.10: 14904033-14904033 on Build GRCh38
rs964762862 Chr.10: 14904198-14904198 on Build GRCh38
rs879463967 Chr.10: 14903961-14903961 on Build GRCh38
rs761613725 Chr.10: 14904038-14904038 on Build GRCh38
rs1022634417 Chr.10: 14904092-14904092 on Build GRCh38
rs750835995 Chr.10: 14904125-14904125 on Build GRCh38
rs868486262 Chr.10: 14904034-14904034 on Build GRCh38
rs956422486 Chr.10: 14904154-14904154 on Build GRCh38
rs971598982 Chr.10: 14903948-14903948 on Build GRCh38
rs868355888 Chr.10: 14904315-14904315 on Build GRCh38
rs560109337 Chr.10: 14903941-14903941 on Build GRCh38
rs767271946 Chr.10: 14904094-14904094 on Build GRCh38
rs923263877 Chr.10: 14904242-14904242 on Build GRCh38
rs914634953 Chr.10: 14904025-14904025 on Build GRCh38
rs948442666 Chr.10: 14904381-14904381 on Build GRCh38
rs538190738 Chr.10: 14904253-14904253 on Build GRCh38
rs756408667 Chr.10: 14904144-14904144 on Build GRCh38
rs997238694 Chr.10: 14904196-14904227 on Build GRCh38
rs939849255 Chr.10: 14904060-14904060 on Build GRCh38
rs139176971 Chr.10: 14904074-14904074 on Build GRCh38
rs775828475 Chr.10: 14904203-14904203 on Build GRCh38
rs540412361 Chr.10: 14903934-14903934 on Build GRCh38

View Full Product Details