Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15076357-15076893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD7
Entrez Gene ID
414149
Gene Name
acyl-CoA binding domain containing 7
Gene Aliases
bA455B2.2
UniGene
Hs.644598
Gene Chromosome Location
Chr.10: 15075475-15088776 on Build GRCh38
Transcripts
NM_001039844.2

SNP Detail

SNP ID Chromosome Location
rs954322660 Chr.10: 15076450-15076450 on Build GRCh38
rs61842449 Chr.10: 15076519-15076519 on Build GRCh38
rs758046831 Chr.10: 15076504-15076506 on Build GRCh38
rs924375824 Chr.10: 15076574-15076574 on Build GRCh38
rs560719742 Chr.10: 15076670-15076670 on Build GRCh38
rs1028547092 Chr.10: 15076446-15076446 on Build GRCh38
rs1019808477 Chr.10: 15076460-15076460 on Build GRCh38
rs1050684541 Chr.10: 15076795-15076795 on Build GRCh38
rs946202438 Chr.10: 15076719-15076720 on Build GRCh38
rs55908255 Chr.10: 15076549-15076549 on Build GRCh38
rs888744808 Chr.10: 15076804-15076804 on Build GRCh38
rs186383781 Chr.10: 15076542-15076542 on Build GRCh38
rs34941547 Chr.10: 15076520-15076520 on Build GRCh38
rs987372759 Chr.10: 15076455-15076455 on Build GRCh38
rs35897924 Chr.10: 15076519-15076519 on Build GRCh38
rs527342778 Chr.10: 15076606-15076606 on Build GRCh38
rs777512772 Chr.10: 15076659-15076660 on Build GRCh38
rs561058949 Chr.10: 15076782-15076782 on Build GRCh38
rs981112397 Chr.10: 15076615-15076615 on Build GRCh38
rs114892549 Chr.10: 15076746-15076746 on Build GRCh38
rs966168337 Chr.10: 15076485-15076485 on Build GRCh38
rs948697085 Chr.10: 15076656-15076656 on Build GRCh38
rs1028692546 Chr.10: 15076390-15076390 on Build GRCh38
rs776629288 Chr.10: 15076626-15076626 on Build GRCh38
rs977115531 Chr.10: 15076562-15076562 on Build GRCh38
rs1008602923 Chr.10: 15076809-15076809 on Build GRCh38
rs865830232 Chr.10: 15076525-15076525 on Build GRCh38
rs1018714366 Chr.10: 15076526-15076526 on Build GRCh38
rs955118078 Chr.10: 15076539-15076539 on Build GRCh38
rs935633046 Chr.10: 15076582-15076582 on Build GRCh38
rs541025470 Chr.10: 15076623-15076623 on Build GRCh38
rs775488606 Chr.10: 15076505-15076506 on Build GRCh38
rs397847726 Chr.10: 15076520-15076521 on Build GRCh38
rs376964150 Chr.10: 15076408-15076408 on Build GRCh38
rs961845432 Chr.10: 15076857-15076857 on Build GRCh38
rs190939308 Chr.10: 15076855-15076855 on Build GRCh38
rs35596040 Chr.10: 15076504-15076505 on Build GRCh38
rs869138442 Chr.10: 15076518-15076519 on Build GRCh38
rs780187773 Chr.10: 15076749-15076749 on Build GRCh38
rs756361214 Chr.10: 15076718-15076718 on Build GRCh38
rs868757232 Chr.10: 15076383-15076383 on Build GRCh38
rs764233655 Chr.10: 15076755-15076755 on Build GRCh38
rs1028367275 Chr.10: 15076822-15076822 on Build GRCh38
rs1029841895 Chr.10: 15076521-15076521 on Build GRCh38
rs962179158 Chr.10: 15076528-15076528 on Build GRCh38
rs937346773 Chr.10: 15076663-15076663 on Build GRCh38
rs1037091638 Chr.10: 15076723-15076723 on Build GRCh38
rs907061354 Chr.10: 15076707-15076707 on Build GRCh38
rs920489822 Chr.10: 15076581-15076581 on Build GRCh38
rs1045964278 Chr.10: 15076669-15076669 on Build GRCh38
rs555756110 Chr.10: 15076735-15076735 on Build GRCh38

View Full Product Details