Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15078795-15079224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD7
Entrez Gene ID
414149
Gene Name
acyl-CoA binding domain containing 7
Gene Aliases
bA455B2.2
UniGene
Hs.644598
Gene Chromosome Location
Chr.10: 15075475-15088776 on Build GRCh38
Transcripts
NM_001039844.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
916662 c.82G>C p.E28Q Chr.10: 15078971-15078971 on Build GRCh38
7160392 c.87C>G p.L29L Chr.10: 15078966-15078966 on Build GRCh38
1297129 c.51G>A p.L17L Chr.10: 15079002-15079002 on Build GRCh38
3436157 c.49C>T p.L17L Chr.10: 15079004-15079004 on Build GRCh38
6461032 c.37G>T p.D13Y Chr.10: 15079016-15079016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11259463 Chr.10: 15079050-15079050 on Build GRCh38
rs76424538 Chr.10: 15078920-15078920 on Build GRCh38
rs982686828 Chr.10: 15078847-15078847 on Build GRCh38
rs935512253 Chr.10: 15079189-15079189 on Build GRCh38
rs774879871 Chr.10: 15079078-15079078 on Build GRCh38
rs773978045 Chr.10: 15078925-15078925 on Build GRCh38
rs756028272 Chr.10: 15078904-15078904 on Build GRCh38
rs754762788 Chr.10: 15079013-15079013 on Build GRCh38
rs951293442 Chr.10: 15078839-15078839 on Build GRCh38
rs1023565559 Chr.10: 15079106-15079106 on Build GRCh38
rs760657236 Chr.10: 15079092-15079092 on Build GRCh38
rs776192960 Chr.10: 15078955-15078955 on Build GRCh38
rs916648553 Chr.10: 15078889-15078889 on Build GRCh38
rs768488252 Chr.10: 15078923-15078923 on Build GRCh38
rs779858331 Chr.10: 15078916-15078916 on Build GRCh38
rs991376387 Chr.10: 15079095-15079096 on Build GRCh38
rs1047796924 Chr.10: 15078871-15078871 on Build GRCh38
rs771541849 Chr.10: 15078935-15078935 on Build GRCh38
rs554900938 Chr.10: 15079190-15079190 on Build GRCh38
rs748294521 Chr.10: 15078889-15078890 on Build GRCh38
rs11259464 Chr.10: 15079196-15079196 on Build GRCh38
rs1009248031 Chr.10: 15079186-15079186 on Build GRCh38
rs72774373 Chr.10: 15079166-15079166 on Build GRCh38
rs750500301 Chr.10: 15078976-15078976 on Build GRCh38
rs751767832 Chr.10: 15078964-15078964 on Build GRCh38
rs375684007 Chr.10: 15079009-15079009 on Build GRCh38
rs7092227 Chr.10: 15078866-15078866 on Build GRCh38
rs926451328 Chr.10: 15078832-15078832 on Build GRCh38
rs371259194 Chr.10: 15078944-15078944 on Build GRCh38
rs11259462 Chr.10: 15079049-15079049 on Build GRCh38
rs745762174 Chr.10: 15078898-15078898 on Build GRCh38
rs767587219 Chr.10: 15078975-15078975 on Build GRCh38
rs539058526 Chr.10: 15079152-15079152 on Build GRCh38
rs770536536 Chr.10: 15079064-15079064 on Build GRCh38
rs140311786 Chr.10: 15078927-15078927 on Build GRCh38
rs929406121 Chr.10: 15079059-15079059 on Build GRCh38
rs778683102 Chr.10: 15079040-15079040 on Build GRCh38
rs776139720 Chr.10: 15079072-15079072 on Build GRCh38
rs751891917 Chr.10: 15078884-15078884 on Build GRCh38
rs997685371 Chr.10: 15079140-15079140 on Build GRCh38
rs568234071 Chr.10: 15079046-15079046 on Build GRCh38
rs544004912 Chr.10: 15079198-15079198 on Build GRCh38
rs146811375 Chr.10: 15078998-15078998 on Build GRCh38
rs763399462 Chr.10: 15078959-15078959 on Build GRCh38
rs988333252 Chr.10: 15079195-15079195 on Build GRCh38
rs781138251 Chr.10: 15078890-15078890 on Build GRCh38
rs982625442 Chr.10: 15078911-15078911 on Build GRCh38
rs937818670 Chr.10: 15079016-15079016 on Build GRCh38
rs375542858 Chr.10: 15079073-15079073 on Build GRCh38
rs4750594 Chr.10: 15078865-15078865 on Build GRCh38
rs773815944 Chr.10: 15078904-15078906 on Build GRCh38
rs1039254699 Chr.10: 15079124-15079124 on Build GRCh38
rs900755391 Chr.10: 15079139-15079139 on Build GRCh38
rs747454646 Chr.10: 15079065-15079066 on Build GRCh38
rs796250854 Chr.10: 15079049-15079050 on Build GRCh38
rs987752761 Chr.10: 15078930-15078930 on Build GRCh38
rs756258898 Chr.10: 15078994-15078994 on Build GRCh38
rs772607552 Chr.10: 15078938-15078938 on Build GRCh38
rs1006648068 Chr.10: 15079080-15079080 on Build GRCh38
rs888018699 Chr.10: 15078873-15078873 on Build GRCh38
rs572699423 Chr.10: 15079174-15079174 on Build GRCh38
rs76311113 Chr.10: 15078824-15078824 on Build GRCh38
rs997906286 Chr.10: 15079010-15079010 on Build GRCh38
rs762056312 Chr.10: 15079083-15079083 on Build GRCh38
rs11598719 Chr.10: 15078966-15078966 on Build GRCh38
rs887761832 Chr.10: 15079065-15079065 on Build GRCh38
rs964879042 Chr.10: 15078893-15078893 on Build GRCh38
rs993496123 Chr.10: 15078867-15078867 on Build GRCh38
rs541702431 Chr.10: 15079187-15079187 on Build GRCh38
rs750706877 Chr.10: 15079091-15079091 on Build GRCh38
rs769085367 Chr.10: 15079074-15079074 on Build GRCh38
rs764623952 Chr.10: 15078960-15078960 on Build GRCh38
rs937853330 Chr.10: 15078855-15078855 on Build GRCh38
rs140440611 Chr.10: 15079061-15079061 on Build GRCh38
rs767604150 Chr.10: 15079086-15079086 on Build GRCh38
rs772011565 Chr.10: 15078897-15078902 on Build GRCh38
rs151285977 Chr.10: 15078943-15078943 on Build GRCh38

View Full Product Details