Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15214074-15214584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM171A1
Entrez Gene ID
221061
Gene Name
family with sequence similarity 171 member A1
Gene Aliases
C10orf38
UniGene
Hs.66762
Gene Chromosome Location
Chr.10: 15211643-15374532 on Build GRCh38
Transcripts
NM_001010924.1 XM_011519378.2 XM_017015904.1 XM_017015905.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3436198 c.1179G>A p.L393L Chr.10: 15214409-15214409 on Build GRCh38
109766 c.1276A>C p.S426R Chr.10: 15214312-15214312 on Build GRCh38
2155962 c.1177C>A p.L393M Chr.10: 15214411-15214411 on Build GRCh38
3436202 c.1170G>A p.T390T Chr.10: 15214418-15214418 on Build GRCh38
427335 c.1044_1045insCA p.E349fs*46 Chr.10: 15214543-15214544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778300286 Chr.10: 15214257-15214257 on Build GRCh38
rs377674623 Chr.10: 15214143-15214143 on Build GRCh38
rs903825712 Chr.10: 15214252-15214252 on Build GRCh38
rs536840013 Chr.10: 15214360-15214360 on Build GRCh38
rs769274660 Chr.10: 15214478-15214478 on Build GRCh38
rs139806876 Chr.10: 15214442-15214442 on Build GRCh38
rs372338891 Chr.10: 15214422-15214422 on Build GRCh38
rs749334881 Chr.10: 15214288-15214288 on Build GRCh38
rs746180178 Chr.10: 15214373-15214373 on Build GRCh38
rs141457982 Chr.10: 15214205-15214205 on Build GRCh38
rs148829329 Chr.10: 15214234-15214234 on Build GRCh38
rs759176110 Chr.10: 15214430-15214430 on Build GRCh38
rs368577858 Chr.10: 15214350-15214350 on Build GRCh38
rs374113882 Chr.10: 15214186-15214186 on Build GRCh38
rs748453395 Chr.10: 15214387-15214388 on Build GRCh38
rs747441428 Chr.10: 15214259-15214259 on Build GRCh38
rs866633913 Chr.10: 15214294-15214294 on Build GRCh38
rs756728893 Chr.10: 15214438-15214438 on Build GRCh38
rs760473693 Chr.10: 15214339-15214339 on Build GRCh38
rs755810029 Chr.10: 15214152-15214152 on Build GRCh38
rs377177825 Chr.10: 15214197-15214197 on Build GRCh38
rs747350618 Chr.10: 15214172-15214172 on Build GRCh38
rs3814165 Chr.10: 15214195-15214195 on Build GRCh38
rs921168013 Chr.10: 15214221-15214221 on Build GRCh38
rs966797488 Chr.10: 15214431-15214431 on Build GRCh38
rs751168273 Chr.10: 15214484-15214484 on Build GRCh38
rs1025529695 Chr.10: 15214323-15214323 on Build GRCh38
rs758134504 Chr.10: 15214390-15214391 on Build GRCh38
rs573536367 Chr.10: 15214483-15214483 on Build GRCh38
rs143895945 Chr.10: 15214433-15214433 on Build GRCh38
rs201423446 Chr.10: 15214517-15214517 on Build GRCh38
rs370616970 Chr.10: 15214157-15214157 on Build GRCh38
rs150647132 Chr.10: 15214441-15214441 on Build GRCh38
rs898863593 Chr.10: 15214127-15214127 on Build GRCh38
rs529897109 Chr.10: 15214550-15214550 on Build GRCh38
rs779523563 Chr.10: 15214153-15214153 on Build GRCh38
rs541362246 Chr.10: 15214465-15214465 on Build GRCh38
rs371933282 Chr.10: 15214453-15214453 on Build GRCh38
rs189694579 Chr.10: 15214560-15214560 on Build GRCh38
rs748125768 Chr.10: 15214452-15214452 on Build GRCh38
rs768653441 Chr.10: 15214292-15214292 on Build GRCh38
rs987498164 Chr.10: 15214337-15214337 on Build GRCh38
rs777516441 Chr.10: 15214453-15214464 on Build GRCh38
rs368467593 Chr.10: 15214435-15214435 on Build GRCh38
rs774179594 Chr.10: 15214226-15214226 on Build GRCh38
rs746303349 Chr.10: 15214266-15214266 on Build GRCh38
rs771844084 Chr.10: 15214418-15214418 on Build GRCh38
rs200052994 Chr.10: 15214372-15214372 on Build GRCh38
rs763670993 Chr.10: 15214482-15214482 on Build GRCh38
rs1015228459 Chr.10: 15214361-15214361 on Build GRCh38
rs761256092 Chr.10: 15214485-15214485 on Build GRCh38
rs149346946 Chr.10: 15214460-15214460 on Build GRCh38
rs777815809 Chr.10: 15214544-15214544 on Build GRCh38
rs749494754 Chr.10: 15214377-15214377 on Build GRCh38
rs774533512 Chr.10: 15214297-15214297 on Build GRCh38
rs779206596 Chr.10: 15214450-15214450 on Build GRCh38
rs762538513 Chr.10: 15214434-15214434 on Build GRCh38
rs745602319 Chr.10: 15214557-15214557 on Build GRCh38
rs143772901 Chr.10: 15214369-15214369 on Build GRCh38
rs761631403 Chr.10: 15214229-15214229 on Build GRCh38
rs145909806 Chr.10: 15214419-15214419 on Build GRCh38
rs764629425 Chr.10: 15214366-15214366 on Build GRCh38
rs755085189 Chr.10: 15214444-15214444 on Build GRCh38
rs776538448 Chr.10: 15214461-15214461 on Build GRCh38
rs765007765 Chr.10: 15214432-15214432 on Build GRCh38
rs770682380 Chr.10: 15214423-15214423 on Build GRCh38
rs755754871 Chr.10: 15214233-15214233 on Build GRCh38
rs148056144 Chr.10: 15214309-15214309 on Build GRCh38
rs868591152 Chr.10: 15214156-15214156 on Build GRCh38
rs898241003 Chr.10: 15214531-15214531 on Build GRCh38
rs1013788044 Chr.10: 15214307-15214307 on Build GRCh38
rs1000170892 Chr.10: 15214468-15214468 on Build GRCh38
rs576625397 Chr.10: 15214370-15214370 on Build GRCh38
rs932539334 Chr.10: 15214223-15214223 on Build GRCh38
rs767191438 Chr.10: 15214232-15214232 on Build GRCh38
rs936640796 Chr.10: 15214295-15214295 on Build GRCh38
rs781122848 Chr.10: 15214554-15214554 on Build GRCh38
rs777868822 Chr.10: 15214454-15214454 on Build GRCh38
rs748262007 Chr.10: 15214401-15214401 on Build GRCh38
rs778961672 Chr.10: 15214512-15214512 on Build GRCh38
rs140852286 Chr.10: 15214183-15214183 on Build GRCh38
rs143460085 Chr.10: 15214262-15214262 on Build GRCh38
rs201274032 Chr.10: 15214394-15214394 on Build GRCh38
rs747160050 Chr.10: 15214548-15214548 on Build GRCh38
rs144392914 Chr.10: 15214115-15214115 on Build GRCh38
rs772498448 Chr.10: 15214162-15214162 on Build GRCh38
rs779252010 Chr.10: 15214277-15214277 on Build GRCh38
rs746927485 Chr.10: 15214457-15214457 on Build GRCh38
rs148482558 Chr.10: 15214312-15214312 on Build GRCh38
rs200789869 Chr.10: 15214151-15214151 on Build GRCh38
rs1042428606 Chr.10: 15214243-15214243 on Build GRCh38
rs780850714 Chr.10: 15214144-15214144 on Build GRCh38
rs139429894 Chr.10: 15214114-15214114 on Build GRCh38
rs751306255 Chr.10: 15214132-15214132 on Build GRCh38
rs767552901 Chr.10: 15214306-15214306 on Build GRCh38
rs751001284 Chr.10: 15214437-15214437 on Build GRCh38
rs956728885 Chr.10: 15214218-15214218 on Build GRCh38
rs757832880 Chr.10: 15214260-15214260 on Build GRCh38
rs779412516 Chr.10: 15214238-15214252 on Build GRCh38
rs756573118 Chr.10: 15214376-15214376 on Build GRCh38
rs746057503 Chr.10: 15214187-15214187 on Build GRCh38
rs911960365 Chr.10: 15214525-15214525 on Build GRCh38
rs151321027 Chr.10: 15214112-15214112 on Build GRCh38
rs371347314 Chr.10: 15214395-15214395 on Build GRCh38
rs1003896448 Chr.10: 15214355-15214355 on Build GRCh38
rs906956662 Chr.10: 15214346-15214346 on Build GRCh38
rs145662406 Chr.10: 15214170-15214170 on Build GRCh38
rs138837950 Chr.10: 15214427-15214427 on Build GRCh38
rs962236898 Chr.10: 15214536-15214536 on Build GRCh38
rs754195448 Chr.10: 15214504-15214504 on Build GRCh38
rs776738874 Chr.10: 15214105-15214105 on Build GRCh38
rs762970660 Chr.10: 15214118-15214118 on Build GRCh38
rs754506637 Chr.10: 15214235-15214235 on Build GRCh38
rs755388845 Chr.10: 15214505-15214505 on Build GRCh38
rs754664838 Chr.10: 15214356-15214356 on Build GRCh38
rs770076811 Chr.10: 15214273-15214273 on Build GRCh38
rs750077585 Chr.10: 15214149-15214149 on Build GRCh38
rs764195006 Chr.10: 15214128-15214128 on Build GRCh38
rs545151535 Chr.10: 15214429-15214429 on Build GRCh38
rs374082293 Chr.10: 15214126-15214126 on Build GRCh38
rs775234345 Chr.10: 15214479-15214479 on Build GRCh38
rs746109500 Chr.10: 15214486-15214486 on Build GRCh38
rs369503900 Chr.10: 15214518-15214518 on Build GRCh38
rs374423460 Chr.10: 15214305-15214305 on Build GRCh38
rs145441467 Chr.10: 15214215-15214215 on Build GRCh38

View Full Product Details