Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15786480-15786983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188A
Entrez Gene ID
80013
Gene Name
family with sequence similarity 188 member A
Gene Aliases
C10orf97, CARP, DERP5, MST126, MSTP126, my042
UniGene
Hs.158870
Gene Chromosome Location
Chr.10: 15778169-15860533 on Build GRCh38
Transcripts
NM_001318330.1 NM_024948.3 XM_005252600.2 XM_005252602.2 XM_006717508.2 XM_011519690.2 XM_011519691.2 XM_011519693.1 XM_011519694.1 XM_017016671.1 XM_017016672.1 XM_017016673.1 XM_017016674.1 XM_017016675.1 XM_017016676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682802 c.1069G>A p.G357R Chr.10: 15786608-15786608 on Build GRCh38
427343 c.1041G>A p.M347I Chr.10: 15786636-15786636 on Build GRCh38
3436251 c.1074C>T p.I358I Chr.10: 15786603-15786603 on Build GRCh38
5034578 c.1116+9G>C p.? Chr.10: 15786552-15786552 on Build GRCh38
7203074 c.1074C>G p.I358M Chr.10: 15786603-15786603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145776875 Chr.10: 15786718-15786718 on Build GRCh38
rs770573249 Chr.10: 15786681-15786681 on Build GRCh38
rs778997815 Chr.10: 15786551-15786551 on Build GRCh38
rs959371742 Chr.10: 15786743-15786743 on Build GRCh38
rs777757716 Chr.10: 15786678-15786678 on Build GRCh38
rs576152931 Chr.10: 15786687-15786687 on Build GRCh38
rs780231673 Chr.10: 15786534-15786534 on Build GRCh38
rs534481290 Chr.10: 15786686-15786687 on Build GRCh38
rs946034019 Chr.10: 15786920-15786920 on Build GRCh38
rs141296028 Chr.10: 15786873-15786873 on Build GRCh38
rs961246024 Chr.10: 15786658-15786658 on Build GRCh38
rs536206669 Chr.10: 15786603-15786603 on Build GRCh38
rs565795738 Chr.10: 15786580-15786580 on Build GRCh38
rs547388496 Chr.10: 15786508-15786508 on Build GRCh38
rs913307979 Chr.10: 15786890-15786890 on Build GRCh38
rs143762840 Chr.10: 15786642-15786642 on Build GRCh38
rs767911740 Chr.10: 15786629-15786629 on Build GRCh38
rs78547190 Chr.10: 15786653-15786653 on Build GRCh38
rs200421665 Chr.10: 15786646-15786646 on Build GRCh38
rs1020841614 Chr.10: 15786703-15786703 on Build GRCh38
rs778979220 Chr.10: 15786655-15786655 on Build GRCh38
rs753365702 Chr.10: 15786575-15786576 on Build GRCh38
rs922132665 Chr.10: 15786821-15786821 on Build GRCh38
rs950374676 Chr.10: 15786763-15786763 on Build GRCh38
rs138443885 Chr.10: 15786742-15786742 on Build GRCh38
rs56392801 Chr.10: 15786601-15786601 on Build GRCh38
rs974006487 Chr.10: 15786667-15786667 on Build GRCh38
rs769363791 Chr.10: 15786686-15786686 on Build GRCh38
rs757107904 Chr.10: 15786872-15786872 on Build GRCh38
rs762162882 Chr.10: 15786621-15786621 on Build GRCh38
rs945638080 Chr.10: 15786505-15786505 on Build GRCh38
rs933498203 Chr.10: 15786841-15786841 on Build GRCh38
rs758234455 Chr.10: 15786674-15786674 on Build GRCh38
rs754441535 Chr.10: 15786663-15786666 on Build GRCh38
rs752746552 Chr.10: 15786671-15786671 on Build GRCh38
rs200060239 Chr.10: 15786652-15786652 on Build GRCh38
rs1043968553 Chr.10: 15786874-15786874 on Build GRCh38
rs574004993 Chr.10: 15786765-15786765 on Build GRCh38
rs764061938 Chr.10: 15786712-15786712 on Build GRCh38
rs374623464 Chr.10: 15786823-15786823 on Build GRCh38
rs909462090 Chr.10: 15786769-15786769 on Build GRCh38
rs377243258 Chr.10: 15786596-15786596 on Build GRCh38
rs761629034 Chr.10: 15786876-15786876 on Build GRCh38
rs1025165439 Chr.10: 15786760-15786760 on Build GRCh38
rs541046543 Chr.10: 15786857-15786857 on Build GRCh38
rs747893028 Chr.10: 15786553-15786553 on Build GRCh38
rs764531471 Chr.10: 15786602-15786602 on Build GRCh38
rs1013310521 Chr.10: 15786744-15786744 on Build GRCh38
rs932690302 Chr.10: 15786913-15786913 on Build GRCh38
rs77694765 Chr.10: 15786696-15786696 on Build GRCh38
rs762358852 Chr.10: 15786700-15786700 on Build GRCh38
rs774899055 Chr.10: 15786613-15786613 on Build GRCh38
rs746799257 Chr.10: 15786679-15786679 on Build GRCh38
rs980240936 Chr.10: 15786701-15786701 on Build GRCh38
rs1039211987 Chr.10: 15786619-15786619 on Build GRCh38
rs780725464 Chr.10: 15786683-15786683 on Build GRCh38
rs749351166 Chr.10: 15786535-15786535 on Build GRCh38
rs372473285 Chr.10: 15786687-15786687 on Build GRCh38
rs551349777 Chr.10: 15786928-15786928 on Build GRCh38
rs963638061 Chr.10: 15786797-15786797 on Build GRCh38
rs903778490 Chr.10: 15786711-15786711 on Build GRCh38
rs141903607 Chr.10: 15786637-15786637 on Build GRCh38
rs772992864 Chr.10: 15786568-15786568 on Build GRCh38
rs1043668231 Chr.10: 15786520-15786520 on Build GRCh38
rs745374220 Chr.10: 15786685-15786685 on Build GRCh38
rs746712630 Chr.10: 15786571-15786571 on Build GRCh38
rs754905294 Chr.10: 15786536-15786536 on Build GRCh38
rs974863248 Chr.10: 15786806-15786806 on Build GRCh38
rs945608367 Chr.10: 15786840-15786840 on Build GRCh38

View Full Product Details