Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15837126-15837641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188A
Entrez Gene ID
80013
Gene Name
family with sequence similarity 188 member A
Gene Aliases
C10orf97, CARP, DERP5, MST126, MSTP126, my042
UniGene
Hs.158870
Gene Chromosome Location
Chr.10: 15778169-15860533 on Build GRCh38
Transcripts
NM_001318330.1 NM_024948.3 XM_005252600.2 XM_005252602.2 XM_006717508.2 XM_011519690.2 XM_011519691.2 XM_011519693.1 XM_011519694.1 XM_017016671.1 XM_017016672.1 XM_017016673.1 XM_017016674.1 XM_017016675.1 XM_017016676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6421245 c.85G>T p.G29* Chr.10: 15837254-15837254 on Build GRCh38
6421244 c.526G>T p.G176* Chr.10: 15837254-15837254 on Build GRCh38
465495 c.536T>C p.F179S Chr.10: 15837244-15837244 on Build GRCh38
4013056 c.79A>G p.M27V Chr.10: 15837260-15837260 on Build GRCh38
3806851 c.474C>G p.F158L Chr.10: 15837306-15837306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992517944 Chr.10: 15837441-15837441 on Build GRCh38
rs1039967660 Chr.10: 15837568-15837568 on Build GRCh38
rs765251866 Chr.10: 15837316-15837316 on Build GRCh38
rs531167159 Chr.10: 15837528-15837528 on Build GRCh38
rs889887224 Chr.10: 15837490-15837490 on Build GRCh38
rs780838769 Chr.10: 15837333-15837333 on Build GRCh38
rs145286799 Chr.10: 15837263-15837263 on Build GRCh38
rs771530354 Chr.10: 15837384-15837384 on Build GRCh38
rs759476856 Chr.10: 15837555-15837555 on Build GRCh38
rs772363234 Chr.10: 15837292-15837292 on Build GRCh38
rs760779729 Chr.10: 15837225-15837225 on Build GRCh38
rs1006947638 Chr.10: 15837508-15837508 on Build GRCh38
rs151207830 Chr.10: 15837160-15837162 on Build GRCh38
rs541618558 Chr.10: 15837340-15837340 on Build GRCh38
rs753986883 Chr.10: 15837232-15837232 on Build GRCh38
rs747403863 Chr.10: 15837534-15837534 on Build GRCh38
rs141542599 Chr.10: 15837423-15837423 on Build GRCh38
rs779362503 Chr.10: 15837352-15837352 on Build GRCh38
rs777236595 Chr.10: 15837401-15837401 on Build GRCh38
rs969000956 Chr.10: 15837358-15837358 on Build GRCh38
rs752638854 Chr.10: 15837325-15837325 on Build GRCh38
rs984230040 Chr.10: 15837542-15837542 on Build GRCh38
rs754924301 Chr.10: 15837250-15837250 on Build GRCh38
rs138124154 Chr.10: 15837296-15837296 on Build GRCh38
rs759701593 Chr.10: 15837313-15837313 on Build GRCh38
rs762101990 Chr.10: 15837180-15837180 on Build GRCh38
rs756794863 Chr.10: 15837329-15837329 on Build GRCh38
rs766419407 Chr.10: 15837228-15837228 on Build GRCh38
rs533685050 Chr.10: 15837398-15837398 on Build GRCh38
rs918509501 Chr.10: 15837341-15837341 on Build GRCh38
rs929636879 Chr.10: 15837447-15837447 on Build GRCh38
rs764730854 Chr.10: 15837320-15837321 on Build GRCh38
rs199663495 Chr.10: 15837260-15837260 on Build GRCh38
rs778209018 Chr.10: 15837325-15837326 on Build GRCh38
rs141513453 Chr.10: 15837217-15837217 on Build GRCh38
rs949662154 Chr.10: 15837425-15837425 on Build GRCh38
rs901325220 Chr.10: 15837613-15837613 on Build GRCh38
rs748653459 Chr.10: 15837353-15837353 on Build GRCh38
rs937066827 Chr.10: 15837385-15837385 on Build GRCh38
rs758181937 Chr.10: 15837261-15837261 on Build GRCh38
rs111280365 Chr.10: 15837293-15837293 on Build GRCh38
rs1021540381 Chr.10: 15837355-15837355 on Build GRCh38
rs751390196 Chr.10: 15837327-15837327 on Build GRCh38
rs1032320245 Chr.10: 15837461-15837461 on Build GRCh38
rs1055231094 Chr.10: 15837391-15837391 on Build GRCh38
rs142665795 Chr.10: 15837309-15837309 on Build GRCh38
rs769097934 Chr.10: 15837275-15837275 on Build GRCh38
rs927071694 Chr.10: 15837379-15837379 on Build GRCh38
rs774900741 Chr.10: 15837283-15837283 on Build GRCh38
rs547989620 Chr.10: 15837318-15837318 on Build GRCh38
rs368071018 Chr.10: 15837186-15837186 on Build GRCh38
rs772657825 Chr.10: 15837360-15837360 on Build GRCh38
rs765443510 Chr.10: 15837587-15837587 on Build GRCh38
rs748204806 Chr.10: 15837169-15837172 on Build GRCh38
rs959882242 Chr.10: 15837402-15837402 on Build GRCh38
rs770580917 Chr.10: 15837380-15837381 on Build GRCh38
rs778997753 Chr.10: 15837258-15837258 on Build GRCh38
rs983741785 Chr.10: 15837368-15837368 on Build GRCh38
rs771219612 Chr.10: 15837546-15837546 on Build GRCh38
rs767767324 Chr.10: 15837197-15837197 on Build GRCh38
rs776784319 Chr.10: 15837310-15837310 on Build GRCh38
rs909550996 Chr.10: 15837550-15837550 on Build GRCh38
rs759681075 Chr.10: 15837565-15837565 on Build GRCh38
rs138505011 Chr.10: 15837266-15837266 on Build GRCh38
rs994009403 Chr.10: 15837573-15837573 on Build GRCh38
rs745389552 Chr.10: 15837274-15837274 on Build GRCh38
rs902756400 Chr.10: 15837432-15837432 on Build GRCh38
rs7903095 Chr.10: 15837442-15837442 on Build GRCh38
rs1043070728 Chr.10: 15837426-15837426 on Build GRCh38
rs780644394 Chr.10: 15837268-15837268 on Build GRCh38
rs931009069 Chr.10: 15837343-15837343 on Build GRCh38
rs1026013678 Chr.10: 15837605-15837605 on Build GRCh38
rs113653394 Chr.10: 15837376-15837376 on Build GRCh38
rs758178221 Chr.10: 15837326-15837326 on Build GRCh38
rs748542130 Chr.10: 15837286-15837286 on Build GRCh38

View Full Product Details