Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 18400895-18401393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB2
Entrez Gene ID
783
Gene Name
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases
CACNLB2, CAVB2, MYSB
UniGene
Hs.59093
Gene Chromosome Location
Chr.10: 18140560-18545881 on Build GRCh38
Transcripts
NM_000724.3 NM_001167945.1 NM_201570.2 NM_201571.3 NM_201572.3 NM_201590.2 NM_201593.2 NM_201596.2 NM_201597.2 XM_005252588.3 XM_006717502.3 XM_011519659.2 XM_011519660.2 XM_017016625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3741596 c.34G>C p.A12P Chr.10: 18401065-18401065 on Build GRCh38
6499377 c.18C>T p.I6I Chr.10: 18401049-18401049 on Build GRCh38
5434782 c.43G>T p.G15* Chr.10: 18401074-18401074 on Build GRCh38
6254829 c.39G>T p.K13N Chr.10: 18401070-18401070 on Build GRCh38
3806883 c.54G>A p.K18K Chr.10: 18401085-18401085 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs903429861 Chr.10: 18401251-18401251 on Build GRCh38
rs868024775 Chr.10: 18401034-18401034 on Build GRCh38
rs554528439 Chr.10: 18401313-18401313 on Build GRCh38
rs772055505 Chr.10: 18401093-18401093 on Build GRCh38
rs189137873 Chr.10: 18401349-18401349 on Build GRCh38
rs965662799 Chr.10: 18400965-18400965 on Build GRCh38
rs200266419 Chr.10: 18401088-18401088 on Build GRCh38
rs771974029 Chr.10: 18400981-18400981 on Build GRCh38
rs544269614 Chr.10: 18401205-18401205 on Build GRCh38
rs868547270 Chr.10: 18401190-18401190 on Build GRCh38
rs917297571 Chr.10: 18401099-18401099 on Build GRCh38
rs763518752 Chr.10: 18401011-18401011 on Build GRCh38
rs574230561 Chr.10: 18401311-18401311 on Build GRCh38
rs565383119 Chr.10: 18401007-18401007 on Build GRCh38
rs962539095 Chr.10: 18401301-18401301 on Build GRCh38
rs370250270 Chr.10: 18400971-18400971 on Build GRCh38
rs757667676 Chr.10: 18401023-18401023 on Build GRCh38
rs117306738 Chr.10: 18400963-18400963 on Build GRCh38
rs762295397 Chr.10: 18401124-18401124 on Build GRCh38
rs941164121 Chr.10: 18400950-18400950 on Build GRCh38
rs569402536 Chr.10: 18400964-18400964 on Build GRCh38
rs374910593 Chr.10: 18401069-18401069 on Build GRCh38
rs1010367398 Chr.10: 18400995-18400995 on Build GRCh38
rs972013146 Chr.10: 18400966-18400966 on Build GRCh38
rs981764686 Chr.10: 18401357-18401357 on Build GRCh38
rs953354951 Chr.10: 18401247-18401247 on Build GRCh38
rs973065942 Chr.10: 18400953-18400953 on Build GRCh38
rs1041983163 Chr.10: 18401248-18401248 on Build GRCh38
rs746422386 Chr.10: 18401333-18401333 on Build GRCh38
rs953471949 Chr.10: 18400925-18400925 on Build GRCh38
rs139184330 Chr.10: 18401090-18401090 on Build GRCh38
rs751942206 Chr.10: 18401020-18401020 on Build GRCh38
rs541619207 Chr.10: 18401319-18401319 on Build GRCh38
rs758919099 Chr.10: 18401118-18401118 on Build GRCh38
rs771094562 Chr.10: 18401025-18401025 on Build GRCh38
rs909754206 Chr.10: 18400927-18400927 on Build GRCh38
rs937132277 Chr.10: 18400984-18400984 on Build GRCh38
rs1029017352 Chr.10: 18401240-18401240 on Build GRCh38
rs1016321131 Chr.10: 18401266-18401266 on Build GRCh38
rs936231349 Chr.10: 18401255-18401255 on Build GRCh38
rs950048560 Chr.10: 18401223-18401223 on Build GRCh38
rs752100216 Chr.10: 18401123-18401123 on Build GRCh38
rs370040046 Chr.10: 18401050-18401050 on Build GRCh38
rs972569136 Chr.10: 18401304-18401304 on Build GRCh38
rs756233244 Chr.10: 18401028-18401028 on Build GRCh38
rs184756750 Chr.10: 18401094-18401094 on Build GRCh38
rs776234515 Chr.10: 18401108-18401108 on Build GRCh38
rs760236241 Chr.10: 18400994-18400994 on Build GRCh38
rs534229682 Chr.10: 18401006-18401006 on Build GRCh38
rs555967843 Chr.10: 18401263-18401263 on Build GRCh38
rs555244679 Chr.10: 18400974-18400974 on Build GRCh38
rs577177654 Chr.10: 18400926-18400926 on Build GRCh38
rs554006918 Chr.10: 18401183-18401183 on Build GRCh38
rs764703043 Chr.10: 18401119-18401119 on Build GRCh38
rs536133287 Chr.10: 18401212-18401212 on Build GRCh38
rs201189633 Chr.10: 18400993-18400993 on Build GRCh38
rs370191541 Chr.10: 18401132-18401132 on Build GRCh38
rs373457804 Chr.10: 18400991-18400991 on Build GRCh38
rs750702461 Chr.10: 18401027-18401027 on Build GRCh38
rs786205265 Chr.10: 18401085-18401087 on Build GRCh38
rs780188247 Chr.10: 18401049-18401049 on Build GRCh38
rs779046436 Chr.10: 18401070-18401070 on Build GRCh38
rs377382858 Chr.10: 18401012-18401012 on Build GRCh38
rs375436063 Chr.10: 18400980-18400980 on Build GRCh38
rs760386247 Chr.10: 18401096-18401096 on Build GRCh38
rs984905257 Chr.10: 18401257-18401257 on Build GRCh38

View Full Product Details