Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 21904380-21904884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC1
Entrez Gene ID
64215
Gene Name
DnaJ heat shock protein family (Hsp40) member C1
Gene Aliases
DNAJL1, ERdj1, HTJ1, MTJ1
UniGene
Hs.499000
Gene Chromosome Location
Chr.10: 21756548-22003768 on Build GRCh38
Transcripts
NM_022365.3 XM_011519614.2 XM_017016536.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5703542 c.785A>T p.D262V Chr.10: 21904557-21904557 on Build GRCh38
365609 c.730G>T p.D244Y Chr.10: 21904612-21904612 on Build GRCh38
3806914 c.735T>C p.A245A Chr.10: 21904607-21904607 on Build GRCh38
7341235 c.808G>A p.E270K Chr.10: 21904534-21904534 on Build GRCh38
6415260 c.767G>T p.R256I Chr.10: 21904575-21904575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371440320 Chr.10: 21904496-21904496 on Build GRCh38
rs868718821 Chr.10: 21904826-21904826 on Build GRCh38
rs748444568 Chr.10: 21904487-21904487 on Build GRCh38
rs577612115 Chr.10: 21904744-21904744 on Build GRCh38
rs1020857956 Chr.10: 21904699-21904699 on Build GRCh38
rs749879261 Chr.10: 21904563-21904563 on Build GRCh38
rs919471326 Chr.10: 21904781-21904781 on Build GRCh38
rs80315655 Chr.10: 21904419-21904420 on Build GRCh38
rs199829427 Chr.10: 21904582-21904582 on Build GRCh38
rs908541683 Chr.10: 21904441-21904441 on Build GRCh38
rs61851898 Chr.10: 21904405-21904405 on Build GRCh38
rs925809394 Chr.10: 21904855-21904855 on Build GRCh38
rs549184605 Chr.10: 21904572-21904572 on Build GRCh38
rs949033521 Chr.10: 21904542-21904542 on Build GRCh38
rs147466228 Chr.10: 21904579-21904579 on Build GRCh38
rs779361897 Chr.10: 21904771-21904771 on Build GRCh38
rs767091919 Chr.10: 21904649-21904649 on Build GRCh38
rs961320962 Chr.10: 21904447-21904447 on Build GRCh38
rs545492166 Chr.10: 21904596-21904596 on Build GRCh38
rs776773994 Chr.10: 21904523-21904523 on Build GRCh38
rs200289319 Chr.10: 21904568-21904568 on Build GRCh38
rs141409128 Chr.10: 21904571-21904571 on Build GRCh38
rs745872865 Chr.10: 21904604-21904604 on Build GRCh38
rs542818538 Chr.10: 21904849-21904849 on Build GRCh38
rs775518370 Chr.10: 21904613-21904613 on Build GRCh38
rs192441556 Chr.10: 21904545-21904545 on Build GRCh38
rs376199935 Chr.10: 21904817-21904818 on Build GRCh38
rs997481225 Chr.10: 21904713-21904713 on Build GRCh38
rs762941364 Chr.10: 21904653-21904654 on Build GRCh38
rs775503833 Chr.10: 21904625-21904642 on Build GRCh38
rs537089517 Chr.10: 21904775-21904775 on Build GRCh38
rs775426214 Chr.10: 21904536-21904536 on Build GRCh38
rs760130454 Chr.10: 21904656-21904656 on Build GRCh38
rs557348081 Chr.10: 21904800-21904800 on Build GRCh38
rs1036603425 Chr.10: 21904504-21904504 on Build GRCh38
rs778099656 Chr.10: 21904575-21904575 on Build GRCh38
rs141362773 Chr.10: 21904710-21904710 on Build GRCh38
rs985733113 Chr.10: 21904819-21904819 on Build GRCh38
rs1046484680 Chr.10: 21904816-21904816 on Build GRCh38
rs758672435 Chr.10: 21904574-21904574 on Build GRCh38
rs781192184 Chr.10: 21904484-21904485 on Build GRCh38
rs773262003 Chr.10: 21904495-21904495 on Build GRCh38
rs765204602 Chr.10: 21904526-21904526 on Build GRCh38
rs759530138 Chr.10: 21904525-21904525 on Build GRCh38
rs765139210 Chr.10: 21904619-21904620 on Build GRCh38
rs574296467 Chr.10: 21904847-21904847 on Build GRCh38
rs774119328 Chr.10: 21904639-21904639 on Build GRCh38
rs766925463 Chr.10: 21904558-21904558 on Build GRCh38
rs763828735 Chr.10: 21904555-21904555 on Build GRCh38
rs79248107 Chr.10: 21904509-21904509 on Build GRCh38
rs373207012 Chr.10: 21904557-21904557 on Build GRCh38
rs1030369452 Chr.10: 21904717-21904717 on Build GRCh38
rs986699427 Chr.10: 21904419-21904419 on Build GRCh38
rs186713248 Chr.10: 21904489-21904489 on Build GRCh38
rs369587800 Chr.10: 21904615-21904615 on Build GRCh38
rs755704263 Chr.10: 21904556-21904556 on Build GRCh38
rs781432147 Chr.10: 21904583-21904583 on Build GRCh38
rs77439330 Chr.10: 21904408-21904408 on Build GRCh38
rs949645418 Chr.10: 21904806-21904806 on Build GRCh38
rs953975868 Chr.10: 21904818-21904818 on Build GRCh38
rs976698329 Chr.10: 21904787-21904787 on Build GRCh38
rs769879922 Chr.10: 21904611-21904611 on Build GRCh38
rs527396473 Chr.10: 21904445-21904445 on Build GRCh38
rs759356365 Chr.10: 21904579-21904582 on Build GRCh38
rs953586178 Chr.10: 21904727-21904727 on Build GRCh38
rs779064107 Chr.10: 21904408-21904408 on Build GRCh38
rs792455 Chr.10: 21904493-21904493 on Build GRCh38
rs777549432 Chr.10: 21904650-21904650 on Build GRCh38
rs986257094 Chr.10: 21904735-21904735 on Build GRCh38
rs965290184 Chr.10: 21904705-21904705 on Build GRCh38

View Full Product Details