Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22317579-22318088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD3
Entrez Gene ID
23412
Gene Name
COMM domain containing 3
Gene Aliases
BUP, C10orf8
UniGene
Hs.534398
Gene Chromosome Location
Chr.10: 22316383-22320317 on Build GRCh38
Transcripts
NM_012071.3 NM_001204062.1

Gene Symbol
COMMD3-BMI1
Entrez Gene ID
100532731
Gene Name
COMMD3-BMI1 readthrough
Gene Aliases
-
UniGene
Hs.380403
Gene Chromosome Location
Chr.10: 22316383-22331485 on Build GRCh38
Transcripts
NM_012071.3 NM_001204062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5050098 c.229C>T p.R77* Chr.10: 22317973-22317973 on Build GRCh38
2133094 c.189T>C p.H63H Chr.10: 22317933-22317933 on Build GRCh38
4912459 c.180A>G p.K60K Chr.10: 22317926-22317926 on Build GRCh38
4965917 c.161T>C p.I54T Chr.10: 22317905-22317905 on Build GRCh38
1474554 c.153G>A p.L51L Chr.10: 22317897-22317897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747041337 Chr.10: 22318037-22318037 on Build GRCh38
rs554824204 Chr.10: 22317804-22317804 on Build GRCh38
rs201107890 Chr.10: 22318009-22318009 on Build GRCh38
rs762505507 Chr.10: 22317886-22317886 on Build GRCh38
rs141169357 Chr.10: 22317631-22317631 on Build GRCh38
rs949466015 Chr.10: 22317622-22317622 on Build GRCh38
rs746955114 Chr.10: 22317948-22317948 on Build GRCh38
rs545385153 Chr.10: 22317993-22317993 on Build GRCh38
rs750837618 Chr.10: 22317819-22317819 on Build GRCh38
rs1047536636 Chr.10: 22317695-22317695 on Build GRCh38
rs577865273 Chr.10: 22317691-22317691 on Build GRCh38
rs755177169 Chr.10: 22317910-22317910 on Build GRCh38
rs557175586 Chr.10: 22317750-22317750 on Build GRCh38
rs774889597 Chr.10: 22317885-22317885 on Build GRCh38
rs780323408 Chr.10: 22317841-22317841 on Build GRCh38
rs758546043 Chr.10: 22317935-22317935 on Build GRCh38
rs1035753974 Chr.10: 22317731-22317731 on Build GRCh38
rs543113343 Chr.10: 22317958-22317958 on Build GRCh38
rs766292147 Chr.10: 22317969-22317969 on Build GRCh38
rs778863839 Chr.10: 22317847-22317847 on Build GRCh38
rs201564339 Chr.10: 22317875-22317875 on Build GRCh38
rs773252260 Chr.10: 22317853-22317853 on Build GRCh38
rs748208878 Chr.10: 22317850-22317850 on Build GRCh38
rs1037174413 Chr.10: 22317786-22317786 on Build GRCh38
rs372028300 Chr.10: 22318035-22318035 on Build GRCh38
rs770921306 Chr.10: 22317951-22317951 on Build GRCh38
rs766744221 Chr.10: 22317905-22317905 on Build GRCh38
rs756180400 Chr.10: 22317817-22317817 on Build GRCh38
rs765443558 Chr.10: 22318006-22318006 on Build GRCh38
rs769385334 Chr.10: 22317960-22317960 on Build GRCh38
rs866975977 Chr.10: 22317970-22317970 on Build GRCh38
rs933066874 Chr.10: 22317604-22317604 on Build GRCh38
rs781419791 Chr.10: 22317960-22317961 on Build GRCh38
rs536887900 Chr.10: 22317726-22317726 on Build GRCh38
rs773794310 Chr.10: 22317974-22317974 on Build GRCh38
rs1037333660 Chr.10: 22317657-22317657 on Build GRCh38
rs910317648 Chr.10: 22317652-22317652 on Build GRCh38
rs937986703 Chr.10: 22317698-22317698 on Build GRCh38
rs1048455318 Chr.10: 22317724-22317724 on Build GRCh38
rs779173780 Chr.10: 22317927-22317927 on Build GRCh38
rs997233291 Chr.10: 22317809-22317809 on Build GRCh38
rs762802964 Chr.10: 22317863-22317863 on Build GRCh38
rs754165911 Chr.10: 22317907-22317907 on Build GRCh38
rs899078500 Chr.10: 22317679-22317679 on Build GRCh38
rs202116321 Chr.10: 22317963-22317963 on Build GRCh38
rs377587517 Chr.10: 22317882-22317882 on Build GRCh38
rs943017304 Chr.10: 22317656-22317656 on Build GRCh38
rs536294304 Chr.10: 22317609-22317609 on Build GRCh38
rs756442192 Chr.10: 22317831-22317831 on Build GRCh38
rs940272968 Chr.10: 22317768-22317768 on Build GRCh38
rs758632811 Chr.10: 22318016-22318016 on Build GRCh38
rs746865139 Chr.10: 22317856-22317856 on Build GRCh38
rs761175014 Chr.10: 22317981-22317981 on Build GRCh38
rs1012887605 Chr.10: 22317779-22317779 on Build GRCh38
rs755337996 Chr.10: 22318003-22318003 on Build GRCh38
rs906385379 Chr.10: 22317709-22317709 on Build GRCh38
rs768865449 Chr.10: 22317845-22317845 on Build GRCh38
rs763582726 Chr.10: 22317965-22317965 on Build GRCh38
rs748214105 Chr.10: 22317932-22317932 on Build GRCh38
rs375698089 Chr.10: 22318000-22318000 on Build GRCh38
rs1021670853 Chr.10: 22317789-22317789 on Build GRCh38
rs757342931 Chr.10: 22317845-22317847 on Build GRCh38
rs926687895 Chr.10: 22318038-22318038 on Build GRCh38
rs376846812 Chr.10: 22318044-22318044 on Build GRCh38
rs374103867 Chr.10: 22317881-22317881 on Build GRCh38
rs973039125 Chr.10: 22317984-22317984 on Build GRCh38
rs1031826703 Chr.10: 22317838-22317838 on Build GRCh38
rs929252140 Chr.10: 22317680-22317680 on Build GRCh38
rs750927222 Chr.10: 22317893-22317893 on Build GRCh38
rs777913128 Chr.10: 22317945-22317945 on Build GRCh38
rs371112741 Chr.10: 22317955-22317955 on Build GRCh38
rs745692233 Chr.10: 22318057-22318057 on Build GRCh38
rs371087282 Chr.10: 22317843-22317843 on Build GRCh38
rs756531761 Chr.10: 22317902-22317902 on Build GRCh38
rs559797623 Chr.10: 22317962-22317962 on Build GRCh38
rs978101721 Chr.10: 22317829-22317829 on Build GRCh38
rs768439024 Chr.10: 22318014-22318014 on Build GRCh38
rs780022629 Chr.10: 22317891-22317891 on Build GRCh38

View Full Product Details