Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22327353-22327887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMI1
Entrez Gene ID
648
Gene Name
BMI1 proto-oncogene, polycomb ring finger
Gene Aliases
FLVI2/BMI1, PCGF4, RNF51, flvi-2/bmi-1
UniGene
Hs.380403
Gene Chromosome Location
Chr.10: 22321210-22331485 on Build GRCh38
Transcripts
NM_005180.8 NM_001204062.1

Gene Symbol
COMMD3-BMI1
Entrez Gene ID
100532731
Gene Name
COMMD3-BMI1 readthrough
Gene Aliases
-
UniGene
Hs.380403
Gene Chromosome Location
Chr.10: 22316383-22331485 on Build GRCh38
Transcripts
NM_005180.8 NM_001204062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
296072 c.221C>G p.T74S Chr.10: 22327606-22327606 on Build GRCh38
5798393 c.268G>A p.E90K Chr.10: 22327744-22327744 on Build GRCh38
1603318 c.249T>A p.V83V Chr.10: 22327634-22327634 on Build GRCh38
6128934 c.301C>T p.H101Y Chr.10: 22327777-22327777 on Build GRCh38
427389 c.229G>C p.D77H Chr.10: 22327614-22327614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746957758 Chr.10: 22327818-22327822 on Build GRCh38
rs776625889 Chr.10: 22327715-22327715 on Build GRCh38
rs775196246 Chr.10: 22327730-22327730 on Build GRCh38
rs547121387 Chr.10: 22327513-22327513 on Build GRCh38
rs773326187 Chr.10: 22327549-22327549 on Build GRCh38
rs765740411 Chr.10: 22327815-22327815 on Build GRCh38
rs758744806 Chr.10: 22327826-22327826 on Build GRCh38
rs376472987 Chr.10: 22327832-22327832 on Build GRCh38
rs145972442 Chr.10: 22327790-22327790 on Build GRCh38
rs770971835 Chr.10: 22327826-22327827 on Build GRCh38
rs766134368 Chr.10: 22327536-22327536 on Build GRCh38
rs779179193 Chr.10: 22327699-22327699 on Build GRCh38
rs888939192 Chr.10: 22327497-22327497 on Build GRCh38
rs777965132 Chr.10: 22327708-22327708 on Build GRCh38
rs763818630 Chr.10: 22327619-22327619 on Build GRCh38
rs144025407 Chr.10: 22327788-22327788 on Build GRCh38
rs747145981 Chr.10: 22327709-22327709 on Build GRCh38
rs760711068 Chr.10: 22327555-22327555 on Build GRCh38
rs777629774 Chr.10: 22327784-22327786 on Build GRCh38
rs536353146 Chr.10: 22327505-22327505 on Build GRCh38
rs375478127 Chr.10: 22327663-22327663 on Build GRCh38
rs139371320 Chr.10: 22327784-22327784 on Build GRCh38
rs780765384 Chr.10: 22327685-22327685 on Build GRCh38
rs538115790 Chr.10: 22327828-22327828 on Build GRCh38
rs200727365 Chr.10: 22327846-22327846 on Build GRCh38
rs113858902 Chr.10: 22327526-22327527 on Build GRCh38
rs386741810 Chr.10: 22327479-22327486 on Build GRCh38
rs757430223 Chr.10: 22327831-22327831 on Build GRCh38
rs1009535677 Chr.10: 22327529-22327529 on Build GRCh38
rs557061825 Chr.10: 22327855-22327855 on Build GRCh38
rs149125612 Chr.10: 22327395-22327395 on Build GRCh38
rs945449139 Chr.10: 22327863-22327863 on Build GRCh38
rs764815710 Chr.10: 22327829-22327829 on Build GRCh38
rs1010243884 Chr.10: 22327461-22327461 on Build GRCh38
rs150886028 Chr.10: 22327616-22327616 on Build GRCh38
rs754319725 Chr.10: 22327692-22327692 on Build GRCh38
rs752495933 Chr.10: 22327589-22327589 on Build GRCh38
rs966418090 Chr.10: 22327478-22327478 on Build GRCh38
rs1019031824 Chr.10: 22327462-22327462 on Build GRCh38
rs113127516 Chr.10: 22327773-22327773 on Build GRCh38
rs772220427 Chr.10: 22327706-22327706 on Build GRCh38
rs755465324 Chr.10: 22327798-22327798 on Build GRCh38
rs114192056 Chr.10: 22327570-22327570 on Build GRCh38
rs761499549 Chr.10: 22327786-22327786 on Build GRCh38
rs763906949 Chr.10: 22327750-22327750 on Build GRCh38
rs774618892 Chr.10: 22327527-22327527 on Build GRCh38
rs568562897 Chr.10: 22327524-22327525 on Build GRCh38
rs758213472 Chr.10: 22327567-22327568 on Build GRCh38
rs759459168 Chr.10: 22327576-22327576 on Build GRCh38
rs774135266 Chr.10: 22327761-22327761 on Build GRCh38
rs893856148 Chr.10: 22327860-22327860 on Build GRCh38
rs566845639 Chr.10: 22327560-22327560 on Build GRCh38
rs1036452340 Chr.10: 22327378-22327378 on Build GRCh38
rs891875352 Chr.10: 22327415-22327415 on Build GRCh38
rs762779689 Chr.10: 22327736-22327736 on Build GRCh38
rs745823038 Chr.10: 22327717-22327717 on Build GRCh38
rs769721002 Chr.10: 22327720-22327720 on Build GRCh38
rs984954299 Chr.10: 22327525-22327525 on Build GRCh38
rs771007673 Chr.10: 22327714-22327714 on Build GRCh38
rs369564383 Chr.10: 22327807-22327807 on Build GRCh38
rs770917839 Chr.10: 22327567-22327567 on Build GRCh38
rs756824541 Chr.10: 22327684-22327684 on Build GRCh38
rs1052260626 Chr.10: 22327401-22327401 on Build GRCh38
rs748499058 Chr.10: 22327703-22327703 on Build GRCh38
rs1053625076 Chr.10: 22327856-22327856 on Build GRCh38
rs755450201 Chr.10: 22327696-22327696 on Build GRCh38
rs201168430 Chr.10: 22327827-22327827 on Build GRCh38
rs996205856 Chr.10: 22327502-22327502 on Build GRCh38
rs751156303 Chr.10: 22327678-22327678 on Build GRCh38
rs753067653 Chr.10: 22327825-22327825 on Build GRCh38
rs1021376536 Chr.10: 22327532-22327532 on Build GRCh38
rs764945458 Chr.10: 22327577-22327577 on Build GRCh38

View Full Product Details